Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 37850523-37851030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NME8
Entrez Gene ID
51314
Gene Name
NME/NM23 family member 8
Gene Aliases
CILD6, HEL-S-99, NM23-H8, SPTRX2, TXNDC3, sptrx-2
UniGene
Hs.723454
Gene Chromosome Location
Chr.7: 37848597-37900401 on Build GRCh38
Transcripts
NM_016616.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4707392 c.140C>A p.P47H Chr.7: 37850677-37850677 on Build GRCh38
1450587 c.177C>T p.D59D Chr.7: 37850714-37850714 on Build GRCh38
1635298 c.198+10T>C p.? Chr.7: 37850745-37850745 on Build GRCh38
5003652 c.129A>C p.R43S Chr.7: 37850666-37850666 on Build GRCh38
6489576 c.130G>A p.A44T Chr.7: 37850667-37850667 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765556103 Chr.7: 37850693-37850694 on Build GRCh38
rs2722372 Chr.7: 37850665-37850665 on Build GRCh38
rs975133477 Chr.7: 37851004-37851004 on Build GRCh38
rs547522924 Chr.7: 37850907-37850908 on Build GRCh38
rs751539349 Chr.7: 37850721-37850721 on Build GRCh38
rs760905416 Chr.7: 37850682-37850682 on Build GRCh38
rs781008114 Chr.7: 37850724-37850724 on Build GRCh38
rs369447226 Chr.7: 37850593-37850593 on Build GRCh38
rs754089157 Chr.7: 37850699-37850699 on Build GRCh38
rs752971479 Chr.7: 37850779-37850779 on Build GRCh38
rs3213977 Chr.7: 37851005-37851005 on Build GRCh38
rs763302949 Chr.7: 37850855-37850855 on Build GRCh38
rs140951161 Chr.7: 37850847-37850850 on Build GRCh38
rs200758390 Chr.7: 37850632-37850632 on Build GRCh38
rs369542497 Chr.7: 37850709-37850709 on Build GRCh38
rs965024180 Chr.7: 37850994-37850994 on Build GRCh38
rs758333100 Chr.7: 37850604-37850604 on Build GRCh38
rs777532091 Chr.7: 37850715-37850715 on Build GRCh38
rs375751041 Chr.7: 37850580-37850580 on Build GRCh38
rs373621005 Chr.7: 37850596-37850596 on Build GRCh38
rs149640541 Chr.7: 37850810-37850810 on Build GRCh38
rs202008099 Chr.7: 37850603-37850603 on Build GRCh38
rs753041716 Chr.7: 37850698-37850699 on Build GRCh38
rs577648663 Chr.7: 37850934-37850934 on Build GRCh38
rs766441690 Chr.7: 37850582-37850582 on Build GRCh38
rs138311368 Chr.7: 37850663-37850663 on Build GRCh38
rs868708838 Chr.7: 37850700-37850700 on Build GRCh38
rs1012991749 Chr.7: 37850835-37850835 on Build GRCh38
rs765497376 Chr.7: 37850776-37850776 on Build GRCh38
rs756982461 Chr.7: 37850723-37850723 on Build GRCh38
rs759817309 Chr.7: 37850774-37850774 on Build GRCh38
rs906242840 Chr.7: 37850851-37850851 on Build GRCh38
rs140124846 Chr.7: 37850922-37850922 on Build GRCh38
rs895773947 Chr.7: 37850821-37850821 on Build GRCh38
rs1022658463 Chr.7: 37850850-37850850 on Build GRCh38
rs2598043 Chr.7: 37850979-37850979 on Build GRCh38
rs779697883 Chr.7: 37850734-37850734 on Build GRCh38
rs540938955 Chr.7: 37850636-37850636 on Build GRCh38
rs768239983 Chr.7: 37850736-37850736 on Build GRCh38
rs1031773229 Chr.7: 37850933-37850933 on Build GRCh38
rs200038015 Chr.7: 37850725-37850725 on Build GRCh38
rs777200123 Chr.7: 37850771-37850771 on Build GRCh38
rs999921846 Chr.7: 37850929-37850929 on Build GRCh38
rs986529297 Chr.7: 37850728-37850728 on Build GRCh38
rs766786157 Chr.7: 37850685-37850685 on Build GRCh38
rs769884433 Chr.7: 37850733-37850733 on Build GRCh38
rs913747487 Chr.7: 37850807-37850807 on Build GRCh38
rs745571114 Chr.7: 37850653-37850653 on Build GRCh38
rs150406326 Chr.7: 37850599-37850599 on Build GRCh38
rs746723740 Chr.7: 37850617-37850617 on Build GRCh38
rs113388202 Chr.7: 37850620-37850620 on Build GRCh38
rs772405642 Chr.7: 37850671-37850671 on Build GRCh38
rs180763598 Chr.7: 37850708-37850708 on Build GRCh38
rs2598042 Chr.7: 37850987-37850987 on Build GRCh38
rs769263780 Chr.7: 37850659-37850659 on Build GRCh38
rs1002238773 Chr.7: 37850587-37850587 on Build GRCh38
rs748684004 Chr.7: 37850735-37850735 on Build GRCh38
rs773539561 Chr.7: 37850747-37850747 on Build GRCh38
rs1001913720 Chr.7: 37850875-37850875 on Build GRCh38
rs372505788 Chr.7: 37850762-37850762 on Build GRCh38
rs199848335 Chr.7: 37850681-37850681 on Build GRCh38
rs146109508 Chr.7: 37850833-37850833 on Build GRCh38
rs759876007 Chr.7: 37850706-37850706 on Build GRCh38
rs780775214 Chr.7: 37850648-37850648 on Build GRCh38
rs113161807 Chr.7: 37850937-37850937 on Build GRCh38
rs763629984 Chr.7: 37850913-37850913 on Build GRCh38
rs761290030 Chr.7: 37850758-37850758 on Build GRCh38
rs777610363 Chr.7: 37850610-37850610 on Build GRCh38
rs545103083 Chr.7: 37850936-37850936 on Build GRCh38
rs2598044 Chr.7: 37850714-37850714 on Build GRCh38

View Full Product Details