Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28858485-28859012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OPRD1
Entrez Gene ID
4985
Gene Name
opioid receptor delta 1
Gene Aliases
OPRD
UniGene
Hs.372
Gene Chromosome Location
Chr.1: 28812142-28863696 on Build GRCh38
Transcripts
NM_000911.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5936917 c.254A>G p.N85S Chr.1: 28858980-28858980 on Build GRCh38
4891436 c.229T>C p.Y77H Chr.1: 28858955-28858955 on Build GRCh38
163090 c.258C>G p.I86M Chr.1: 28858984-28858984 on Build GRCh38
4031218 c.263T>C p.I88T Chr.1: 28858989-28858989 on Build GRCh38
39537 c.245C>T p.T82M Chr.1: 28858971-28858971 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750294449 Chr.1: 28858910-28858910 on Build GRCh38
rs964663230 Chr.1: 28858887-28858887 on Build GRCh38
rs766147800 Chr.1: 28858937-28858937 on Build GRCh38
rs934478575 Chr.1: 28858615-28858615 on Build GRCh38
rs545758877 Chr.1: 28858657-28858657 on Build GRCh38
rs985019433 Chr.1: 28858726-28858726 on Build GRCh38
rs754696931 Chr.1: 28858954-28858954 on Build GRCh38
rs775151883 Chr.1: 28858972-28858972 on Build GRCh38
rs896653076 Chr.1: 28858743-28858743 on Build GRCh38
rs1002868947 Chr.1: 28858705-28858705 on Build GRCh38
rs922073241 Chr.1: 28858699-28858699 on Build GRCh38
rs1017260945 Chr.1: 28858870-28858870 on Build GRCh38
rs573564683 Chr.1: 28858653-28858653 on Build GRCh38
rs142467790 Chr.1: 28858990-28858990 on Build GRCh38
rs553127302 Chr.1: 28858763-28858763 on Build GRCh38
rs751288367 Chr.1: 28858938-28858938 on Build GRCh38
rs1016670798 Chr.1: 28858529-28858529 on Build GRCh38
rs552160832 Chr.1: 28858543-28858543 on Build GRCh38
rs972853855 Chr.1: 28858582-28858582 on Build GRCh38
rs762686120 Chr.1: 28858911-28858911 on Build GRCh38
rs892820142 Chr.1: 28858879-28858879 on Build GRCh38
rs558990527 Chr.1: 28858731-28858731 on Build GRCh38
rs892528147 Chr.1: 28858650-28858650 on Build GRCh38
rs866650725 Chr.1: 28858953-28858953 on Build GRCh38
rs144634501 Chr.1: 28858605-28858605 on Build GRCh38
rs537013412 Chr.1: 28858585-28858585 on Build GRCh38
rs972214074 Chr.1: 28858512-28858512 on Build GRCh38
rs765180888 Chr.1: 28858909-28858909 on Build GRCh38
rs976396298 Chr.1: 28858901-28858901 on Build GRCh38
rs749756833 Chr.1: 28858973-28858973 on Build GRCh38
rs905477830 Chr.1: 28858683-28858683 on Build GRCh38
rs951694266 Chr.1: 28858530-28858530 on Build GRCh38
rs112411290 Chr.1: 28858801-28858801 on Build GRCh38
rs923094672 Chr.1: 28858607-28858607 on Build GRCh38
rs755623365 Chr.1: 28858962-28858962 on Build GRCh38
rs984430464 Chr.1: 28858544-28858544 on Build GRCh38
rs558944910 Chr.1: 28858820-28858820 on Build GRCh38
rs1045599980 Chr.1: 28858819-28858819 on Build GRCh38
rs940693540 Chr.1: 28858735-28858735 on Build GRCh38
rs760721041 Chr.1: 28858907-28858907 on Build GRCh38
rs374517967 Chr.1: 28858635-28858635 on Build GRCh38
rs905656881 Chr.1: 28858854-28858854 on Build GRCh38
rs897010098 Chr.1: 28858521-28858521 on Build GRCh38
rs1035230760 Chr.1: 28858736-28858736 on Build GRCh38
rs867386525 Chr.1: 28858614-28858614 on Build GRCh38
rs1009884487 Chr.1: 28858900-28858900 on Build GRCh38
rs780984314 Chr.1: 28858957-28858957 on Build GRCh38
rs918976848 Chr.1: 28858513-28858513 on Build GRCh38
rs1045568321 Chr.1: 28858576-28858576 on Build GRCh38
rs16837737 Chr.1: 28858865-28858865 on Build GRCh38
rs910484005 Chr.1: 28858600-28858600 on Build GRCh38
rs966203229 Chr.1: 28858671-28858671 on Build GRCh38
rs975965055 Chr.1: 28858606-28858606 on Build GRCh38
rs1036878804 Chr.1: 28858758-28858758 on Build GRCh38
rs1026499694 Chr.1: 28858762-28858762 on Build GRCh38
rs12404612 Chr.1: 28858581-28858581 on Build GRCh38
rs566693362 Chr.1: 28858793-28858793 on Build GRCh38
rs575897727 Chr.1: 28858829-28858829 on Build GRCh38
rs536379262 Chr.1: 28858701-28858701 on Build GRCh38
rs752309169 Chr.1: 28858961-28858961 on Build GRCh38
rs147870831 Chr.1: 28858641-28858641 on Build GRCh38
rs918025878 Chr.1: 28858771-28858771 on Build GRCh38
rs993639597 Chr.1: 28858750-28858750 on Build GRCh38
rs923174398 Chr.1: 28858914-28858914 on Build GRCh38

View Full Product Details