Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56162513-56163025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMARCC2
Entrez Gene ID
6601
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 2
Gene Aliases
BAF170, CRACC2, Rsc8
UniGene
Hs.236030
Gene Chromosome Location
Chr.12: 56161852-56189567 on Build GRCh38
Transcripts
NM_001130420.1 NM_003075.3 NM_139067.2 XM_005269101.1 XM_005269102.1 XM_005269103.1 XM_005269104.1 XM_011538693.2 XM_017019884.1 XM_017019885.1 XM_017019886.1 XM_017019887.1

SNP Detail

SNP ID Chromosome Location
rs76618553 Chr.12: 56162849-56162849 on Build GRCh38
rs552827280 Chr.12: 56162614-56162614 on Build GRCh38
rs1042310936 Chr.12: 56162692-56162692 on Build GRCh38
rs986874327 Chr.12: 56162599-56162599 on Build GRCh38
rs746757371 Chr.12: 56162545-56162545 on Build GRCh38
rs973172422 Chr.12: 56162588-56162588 on Build GRCh38
rs926149424 Chr.12: 56162718-56162718 on Build GRCh38
rs757273275 Chr.12: 56162989-56162989 on Build GRCh38
rs986780229 Chr.12: 56162967-56162967 on Build GRCh38
rs946337863 Chr.12: 56162639-56162639 on Build GRCh38
rs890370837 Chr.12: 56162833-56162833 on Build GRCh38
rs151015752 Chr.12: 56162947-56162947 on Build GRCh38
rs768299050 Chr.12: 56162743-56162743 on Build GRCh38
rs575182959 Chr.12: 56162796-56162796 on Build GRCh38
rs78451485 Chr.12: 56162683-56162683 on Build GRCh38
rs758207608 Chr.12: 56162570-56162570 on Build GRCh38
rs745785785 Chr.12: 56162602-56162602 on Build GRCh38
rs998158456 Chr.12: 56162823-56162823 on Build GRCh38
rs1058342 Chr.12: 56162603-56162603 on Build GRCh38
rs558530398 Chr.12: 56162556-56162556 on Build GRCh38
rs893596660 Chr.12: 56162676-56162676 on Build GRCh38
rs1012378068 Chr.12: 56162837-56162837 on Build GRCh38
rs34102859 Chr.12: 56162605-56162605 on Build GRCh38
rs963120287 Chr.12: 56162579-56162579 on Build GRCh38
rs781151522 Chr.12: 56162595-56162595 on Build GRCh38
rs972980812 Chr.12: 56162910-56162910 on Build GRCh38
rs918623602 Chr.12: 56162566-56162566 on Build GRCh38
rs565303262 Chr.12: 56162830-56162830 on Build GRCh38
rs1034077553 Chr.12: 56162938-56162938 on Build GRCh38
rs543364875 Chr.12: 56162824-56162824 on Build GRCh38
rs541669700 Chr.12: 56162735-56162735 on Build GRCh38
rs532463222 Chr.12: 56162847-56162847 on Build GRCh38
rs902210391 Chr.12: 56162709-56162709 on Build GRCh38
rs1004685028 Chr.12: 56162555-56162555 on Build GRCh38
rs933972839 Chr.12: 56162580-56162580 on Build GRCh38
rs117911625 Chr.12: 56162791-56162791 on Build GRCh38
rs1025837456 Chr.12: 56162937-56162937 on Build GRCh38
rs994685002 Chr.12: 56162905-56162905 on Build GRCh38
rs899025792 Chr.12: 56162868-56162868 on Build GRCh38
rs964169507 Chr.12: 56162994-56162995 on Build GRCh38
rs936076233 Chr.12: 56162719-56162719 on Build GRCh38
rs570428512 Chr.12: 56162592-56162592 on Build GRCh38
rs899935307 Chr.12: 56162785-56162785 on Build GRCh38
rs992470091 Chr.12: 56162970-56162970 on Build GRCh38
rs917466084 Chr.12: 56162615-56162615 on Build GRCh38

View Full Product Details