Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 120254003-120254497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC649133
Entrez Gene ID
649133
Gene Name
uncharacterized LOC649133
Gene Aliases
-
UniGene
Hs.568902
Gene Chromosome Location
Chr.11: 120249759-120265932 on Build GRCh38
Transcripts
NR_038829.1 NM_001244682.1 NM_014352.3 XM_011542739.2 XM_011542740.2 XM_011542741.2 XM_011542742.2 XM_011542743.2 XM_017017487.1

Gene Symbol
POU2F3
Entrez Gene ID
25833
Gene Name
POU class 2 homeobox 3
Gene Aliases
Epoc-1, OCT-11, OCT11, OTF-11, PLA-1, PLA1, Skn-1a
UniGene
Hs.227115
Gene Chromosome Location
Chr.11: 120236640-120319944 on Build GRCh38
Transcripts
NR_038829.1 NM_001244682.1 NM_014352.3 XM_011542739.2 XM_011542740.2 XM_011542741.2 XM_011542742.2 XM_011542743.2 XM_017017487.1

SNP Detail

SNP ID Chromosome Location
rs569480822 Chr.11: 120254212-120254212 on Build GRCh38
rs1048084221 Chr.11: 120254034-120254034 on Build GRCh38
rs113621906 Chr.11: 120254136-120254136 on Build GRCh38
rs991706938 Chr.11: 120254243-120254243 on Build GRCh38
rs577640497 Chr.11: 120254344-120254344 on Build GRCh38
rs1040785013 Chr.11: 120254050-120254050 on Build GRCh38
rs35460702 Chr.11: 120254044-120254044 on Build GRCh38
rs185337054 Chr.11: 120254408-120254408 on Build GRCh38
rs1050347220 Chr.11: 120254330-120254330 on Build GRCh38
rs186088613 Chr.11: 120254192-120254192 on Build GRCh38
rs942369899 Chr.11: 120254420-120254420 on Build GRCh38
rs544664476 Chr.11: 120254091-120254091 on Build GRCh38
rs889579786 Chr.11: 120254039-120254039 on Build GRCh38
rs906303471 Chr.11: 120254268-120254268 on Build GRCh38
rs778939090 Chr.11: 120254213-120254213 on Build GRCh38
rs1034200070 Chr.11: 120254307-120254307 on Build GRCh38
rs555194420 Chr.11: 120254228-120254228 on Build GRCh38
rs368282821 Chr.11: 120254309-120254309 on Build GRCh38
rs1046129218 Chr.11: 120254266-120254266 on Build GRCh38
rs904314356 Chr.11: 120254275-120254275 on Build GRCh38
rs867958259 Chr.11: 120254084-120254084 on Build GRCh38
rs958716138 Chr.11: 120254308-120254308 on Build GRCh38
rs998950318 Chr.11: 120254413-120254413 on Build GRCh38
rs952157558 Chr.11: 120254378-120254378 on Build GRCh38
rs1010102773 Chr.11: 120254246-120254246 on Build GRCh38
rs962259770 Chr.11: 120254183-120254183 on Build GRCh38
rs977855293 Chr.11: 120254210-120254210 on Build GRCh38
rs967901992 Chr.11: 120254365-120254365 on Build GRCh38
rs562886139 Chr.11: 120254140-120254140 on Build GRCh38
rs879588649 Chr.11: 120254354-120254355 on Build GRCh38
rs930879535 Chr.11: 120254202-120254202 on Build GRCh38
rs1050628354 Chr.11: 120254208-120254208 on Build GRCh38
rs976813871 Chr.11: 120254442-120254442 on Build GRCh38
rs1036070675 Chr.11: 120254279-120254279 on Build GRCh38
rs904376056 Chr.11: 120254471-120254471 on Build GRCh38
rs141908317 Chr.11: 120254453-120254453 on Build GRCh38
rs547015821 Chr.11: 120254092-120254092 on Build GRCh38
rs1046584408 Chr.11: 120254029-120254029 on Build GRCh38
rs994722863 Chr.11: 120254318-120254318 on Build GRCh38
rs1041431662 Chr.11: 120254467-120254467 on Build GRCh38
rs899115910 Chr.11: 120254075-120254075 on Build GRCh38
rs750550359 Chr.11: 120254222-120254222 on Build GRCh38
rs1032938852 Chr.11: 120254444-120254444 on Build GRCh38
rs958662218 Chr.11: 120254117-120254117 on Build GRCh38
rs922413336 Chr.11: 120254450-120254450 on Build GRCh38
rs1000075814 Chr.11: 120254284-120254284 on Build GRCh38
rs957378650 Chr.11: 120254447-120254447 on Build GRCh38
rs148504611 Chr.11: 120254349-120254349 on Build GRCh38
rs34859262 Chr.11: 120254051-120254051 on Build GRCh38
rs181806445 Chr.11: 120254325-120254325 on Build GRCh38
rs536856528 Chr.11: 120254226-120254226 on Build GRCh38
rs1006648290 Chr.11: 120254045-120254045 on Build GRCh38
rs753617934 Chr.11: 120254101-120254101 on Build GRCh38
rs1010463511 Chr.11: 120254062-120254062 on Build GRCh38
rs1019016988 Chr.11: 120254350-120254350 on Build GRCh38
rs539151643 Chr.11: 120254148-120254148 on Build GRCh38
rs2715888 Chr.11: 120254244-120254244 on Build GRCh38
rs901021388 Chr.11: 120254061-120254061 on Build GRCh38
rs1024422211 Chr.11: 120254320-120254320 on Build GRCh38
rs920890149 Chr.11: 120254196-120254196 on Build GRCh38
rs867522250 Chr.11: 120254380-120254380 on Build GRCh38
rs1034312972 Chr.11: 120254109-120254109 on Build GRCh38
rs191202559 Chr.11: 120254270-120254270 on Build GRCh38

View Full Product Details