Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30921850-30922372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL19-AS1
Entrez Gene ID
283932
Gene Name
FBXL19 antisense RNA 1 (head to head)
Gene Aliases
NCRNA00095
UniGene
Hs.711535
Gene Chromosome Location
Chr.16: 30919319-30923269 on Build GRCh38
Transcripts
NR_024348.1

SNP Detail

SNP ID Chromosome Location
rs995250981 Chr.16: 30922008-30922008 on Build GRCh38
rs900588495 Chr.16: 30922052-30922052 on Build GRCh38
rs944849309 Chr.16: 30922000-30922000 on Build GRCh38
rs144512749 Chr.16: 30922225-30922225 on Build GRCh38
rs140636137 Chr.16: 30921918-30921918 on Build GRCh38
rs573156685 Chr.16: 30922025-30922025 on Build GRCh38
rs944938220 Chr.16: 30922319-30922319 on Build GRCh38
rs983738353 Chr.16: 30922308-30922308 on Build GRCh38
rs900214681 Chr.16: 30921991-30921991 on Build GRCh38
rs907417328 Chr.16: 30922044-30922052 on Build GRCh38
rs1027062308 Chr.16: 30922329-30922329 on Build GRCh38
rs369160802 Chr.16: 30921904-30921904 on Build GRCh38
rs77088799 Chr.16: 30922226-30922226 on Build GRCh38
rs931645400 Chr.16: 30921996-30921996 on Build GRCh38
rs918274175 Chr.16: 30922249-30922249 on Build GRCh38
rs765447318 Chr.16: 30922263-30922263 on Build GRCh38
rs983368347 Chr.16: 30921900-30921900 on Build GRCh38
rs930530592 Chr.16: 30921875-30921875 on Build GRCh38
rs1042404337 Chr.16: 30922251-30922251 on Build GRCh38
rs543749158 Chr.16: 30922195-30922195 on Build GRCh38
rs376688152 Chr.16: 30922004-30922007 on Build GRCh38
rs1055772416 Chr.16: 30922111-30922111 on Build GRCh38
rs931697839 Chr.16: 30922221-30922221 on Build GRCh38
rs1010134871 Chr.16: 30922174-30922174 on Build GRCh38
rs907427168 Chr.16: 30922259-30922259 on Build GRCh38
rs1010931356 Chr.16: 30922027-30922027 on Build GRCh38
rs562116652 Chr.16: 30922104-30922104 on Build GRCh38
rs1054500661 Chr.16: 30922002-30922005 on Build GRCh38
rs929610323 Chr.16: 30922303-30922303 on Build GRCh38
rs11644389 Chr.16: 30921938-30921938 on Build GRCh38
rs1034454920 Chr.16: 30922265-30922265 on Build GRCh38
rs899226983 Chr.16: 30922004-30922004 on Build GRCh38
rs921693629 Chr.16: 30922210-30922210 on Build GRCh38
rs1039619512 Chr.16: 30922338-30922338 on Build GRCh38
rs1033442089 Chr.16: 30922090-30922090 on Build GRCh38
rs944525336 Chr.16: 30922188-30922188 on Build GRCh38
rs945725806 Chr.16: 30922237-30922237 on Build GRCh38
rs973819574 Chr.16: 30922232-30922232 on Build GRCh38
rs1020524784 Chr.16: 30922035-30922035 on Build GRCh38
rs561159392 Chr.16: 30922017-30922017 on Build GRCh38
rs983806334 Chr.16: 30922150-30922150 on Build GRCh38
rs912147151 Chr.16: 30922310-30922310 on Build GRCh38
rs894627828 Chr.16: 30922286-30922286 on Build GRCh38
rs189601154 Chr.16: 30922055-30922055 on Build GRCh38
rs1027422034 Chr.16: 30922100-30922100 on Build GRCh38
rs951501143 Chr.16: 30922345-30922345 on Build GRCh38
rs890657166 Chr.16: 30921964-30921964 on Build GRCh38
rs182250378 Chr.16: 30922248-30922248 on Build GRCh38
rs944513940 Chr.16: 30921944-30921945 on Build GRCh38
rs980338901 Chr.16: 30922212-30922212 on Build GRCh38
rs747798285 Chr.16: 30922095-30922096 on Build GRCh38
rs1040185982 Chr.16: 30921979-30921979 on Build GRCh38
rs1024161247 Chr.16: 30922189-30922189 on Build GRCh38
rs543085415 Chr.16: 30922011-30922011 on Build GRCh38
rs1047985137 Chr.16: 30921950-30921950 on Build GRCh38
rs1034402672 Chr.16: 30922079-30922079 on Build GRCh38

View Full Product Details