Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30922592-30923100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL19
Entrez Gene ID
54620
Gene Name
F-box and leucine rich repeat protein 19
Gene Aliases
CXXC11, Fbl19, JHDM1C
UniGene
Hs.152149
Gene Chromosome Location
Chr.16: 30923055-30948783 on Build GRCh38
Transcripts
NM_001282351.1 NR_024348.1

Gene Symbol
FBXL19-AS1
Entrez Gene ID
283932
Gene Name
FBXL19 antisense RNA 1 (head to head)
Gene Aliases
NCRNA00095
UniGene
Hs.711535
Gene Chromosome Location
Chr.16: 30919319-30923269 on Build GRCh38
Transcripts
NM_001282351.1 NR_024348.1

SNP Detail

SNP ID Chromosome Location
rs780548372 Chr.16: 30923027-30923027 on Build GRCh38
rs549534156 Chr.16: 30922845-30922845 on Build GRCh38
rs977172769 Chr.16: 30922800-30922800 on Build GRCh38
rs984424931 Chr.16: 30922992-30922992 on Build GRCh38
rs777106439 Chr.16: 30923061-30923061 on Build GRCh38
rs908786491 Chr.16: 30922999-30922999 on Build GRCh38
rs946147763 Chr.16: 30922829-30922829 on Build GRCh38
rs185275374 Chr.16: 30922988-30922988 on Build GRCh38
rs1019363069 Chr.16: 30922945-30922945 on Build GRCh38
rs1011518560 Chr.16: 30922910-30922910 on Build GRCh38
rs1021480268 Chr.16: 30922795-30922795 on Build GRCh38
rs1055697201 Chr.16: 30922727-30922727 on Build GRCh38
rs1020427214 Chr.16: 30923044-30923044 on Build GRCh38
rs1054501411 Chr.16: 30922632-30922632 on Build GRCh38
rs748652745 Chr.16: 30922809-30922809 on Build GRCh38
rs975431897 Chr.16: 30922807-30922807 on Build GRCh38
rs774074045 Chr.16: 30922949-30922949 on Build GRCh38
rs879194009 Chr.16: 30922801-30922801 on Build GRCh38
rs878933996 Chr.16: 30922734-30922734 on Build GRCh38
rs893778400 Chr.16: 30922904-30922904 on Build GRCh38
rs984761835 Chr.16: 30922844-30922844 on Build GRCh38
rs886918658 Chr.16: 30922915-30922915 on Build GRCh38
rs759490863 Chr.16: 30922630-30922631 on Build GRCh38
rs1056155607 Chr.16: 30922851-30922851 on Build GRCh38
rs532001310 Chr.16: 30923001-30923001 on Build GRCh38
rs922697802 Chr.16: 30923020-30923020 on Build GRCh38
rs371268352 Chr.16: 30922773-30922773 on Build GRCh38
rs990332442 Chr.16: 30923026-30923026 on Build GRCh38
rs755603269 Chr.16: 30922695-30922695 on Build GRCh38
rs913313972 Chr.16: 30922633-30922633 on Build GRCh38
rs903054149 Chr.16: 30922933-30922933 on Build GRCh38
rs898415561 Chr.16: 30922860-30922860 on Build GRCh38
rs201034706 Chr.16: 30922813-30922813 on Build GRCh38
rs907183160 Chr.16: 30922694-30922694 on Build GRCh38
rs747303358 Chr.16: 30923028-30923028 on Build GRCh38
rs931481027 Chr.16: 30922666-30922666 on Build GRCh38
rs959753550 Chr.16: 30922948-30922948 on Build GRCh38
rs866380454 Chr.16: 30922731-30922731 on Build GRCh38
rs561462134 Chr.16: 30922868-30922868 on Build GRCh38
rs1035409223 Chr.16: 30922947-30922947 on Build GRCh38
rs898351371 Chr.16: 30922742-30922742 on Build GRCh38
rs887209414 Chr.16: 30922683-30922683 on Build GRCh38
rs1056011445 Chr.16: 30922651-30922651 on Build GRCh38
rs951120379 Chr.16: 30922772-30922772 on Build GRCh38
rs573217038 Chr.16: 30922806-30922806 on Build GRCh38
rs146780755 Chr.16: 30923045-30923045 on Build GRCh38
rs922202390 Chr.16: 30922808-30922808 on Build GRCh38
rs758891549 Chr.16: 30923018-30923018 on Build GRCh38
rs188206247 Chr.16: 30922638-30922638 on Build GRCh38
rs931558882 Chr.16: 30922830-30922830 on Build GRCh38
rs140973394 Chr.16: 30922797-30922797 on Build GRCh38
rs903064275 Chr.16: 30923069-30923069 on Build GRCh38
rs1045768454 Chr.16: 30922843-30922843 on Build GRCh38
rs150202164 Chr.16: 30922972-30922972 on Build GRCh38
rs967646440 Chr.16: 30922886-30922886 on Build GRCh38
rs1011841026 Chr.16: 30922792-30922792 on Build GRCh38
rs192762891 Chr.16: 30922728-30922728 on Build GRCh38
rs946094029 Chr.16: 30922636-30922636 on Build GRCh38
rs1042980350 Chr.16: 30923065-30923065 on Build GRCh38
rs967499934 Chr.16: 30923046-30923046 on Build GRCh38
rs751593794 Chr.16: 30922992-30922994 on Build GRCh38
rs879064436 Chr.16: 30922877-30922877 on Build GRCh38
rs932744814 Chr.16: 30923023-30923025 on Build GRCh38
rs769034594 Chr.16: 30923050-30923050 on Build GRCh38
rs1048527856 Chr.16: 30922667-30922667 on Build GRCh38
rs1041191096 Chr.16: 30922861-30922861 on Build GRCh38
rs866045321 Chr.16: 30922620-30922620 on Build GRCh38
rs916162035 Chr.16: 30922814-30922814 on Build GRCh38
rs995349735 Chr.16: 30922751-30922751 on Build GRCh38
rs3813020 Chr.16: 30922754-30922754 on Build GRCh38
rs536729049 Chr.16: 30922786-30922786 on Build GRCh38
rs1046945333 Chr.16: 30922881-30922881 on Build GRCh38
rs1045714636 Chr.16: 30922693-30922693 on Build GRCh38
rs916125091 Chr.16: 30922663-30922663 on Build GRCh38
rs1004324585 Chr.16: 30922685-30922685 on Build GRCh38
rs932649997 Chr.16: 30922878-30922878 on Build GRCh38
rs990278335 Chr.16: 30922811-30922811 on Build GRCh38
rs778158319 Chr.16: 30922958-30922958 on Build GRCh38
rs893839296 Chr.16: 30923036-30923036 on Build GRCh38
rs937344321 Chr.16: 30922847-30922847 on Build GRCh38
rs938340195 Chr.16: 30922642-30922642 on Build GRCh38
rs1021952325 Chr.16: 30922911-30922911 on Build GRCh38
rs1008006050 Chr.16: 30922922-30922922 on Build GRCh38
rs151069683 Chr.16: 30922692-30922692 on Build GRCh38

View Full Product Details