Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 85285310-85285842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSE1
Entrez Gene ID
23199
Gene Name
Gse1 coiled-coil protein
Gene Aliases
CRHSP24, KIAA0182
UniGene
Hs.461647
Gene Chromosome Location
Chr.16: 85169512-85676206 on Build GRCh38
Transcripts
XM_005255859.4 XM_005255860.3 XM_005255861.4 NR_038859.1

Gene Symbol
LINC00311
Entrez Gene ID
197196
Gene Name
long intergenic non-protein coding RNA 311
Gene Aliases
NCRNA00311, TMEM148
UniGene
Hs.679002
Gene Chromosome Location
Chr.16: 85282958-85288082 on Build GRCh38
Transcripts
XM_005255859.4 XM_005255860.3 XM_005255861.4 NR_038859.1

SNP Detail

SNP ID Chromosome Location
rs1041619211 Chr.16: 85285747-85285747 on Build GRCh38
rs142123505 Chr.16: 85285510-85285510 on Build GRCh38
rs539078548 Chr.16: 85285669-85285669 on Build GRCh38
rs907306700 Chr.16: 85285380-85285380 on Build GRCh38
rs1026926106 Chr.16: 85285607-85285607 on Build GRCh38
rs574849275 Chr.16: 85285386-85285386 on Build GRCh38
rs752906341 Chr.16: 85285571-85285571 on Build GRCh38
rs555278595 Chr.16: 85285766-85285766 on Build GRCh38
rs563531488 Chr.16: 85285445-85285445 on Build GRCh38
rs942111241 Chr.16: 85285352-85285352 on Build GRCh38
rs989836562 Chr.16: 85285740-85285740 on Build GRCh38
rs552824340 Chr.16: 85285676-85285676 on Build GRCh38
rs907356947 Chr.16: 85285575-85285575 on Build GRCh38
rs1002966400 Chr.16: 85285576-85285576 on Build GRCh38
rs963811835 Chr.16: 85285808-85285808 on Build GRCh38
rs777735401 Chr.16: 85285595-85285595 on Build GRCh38
rs946313777 Chr.16: 85285754-85285754 on Build GRCh38
rs1056242565 Chr.16: 85285583-85285583 on Build GRCh38
rs1027504143 Chr.16: 85285440-85285440 on Build GRCh38
rs886564850 Chr.16: 85285339-85285339 on Build GRCh38
rs1003365259 Chr.16: 85285392-85285392 on Build GRCh38
rs1010339100 Chr.16: 85285498-85285498 on Build GRCh38
rs1037715368 Chr.16: 85285362-85285362 on Build GRCh38
rs894498577 Chr.16: 85285588-85285588 on Build GRCh38
rs995496876 Chr.16: 85285606-85285606 on Build GRCh38
rs921619094 Chr.16: 85285549-85285549 on Build GRCh38
rs941996401 Chr.16: 85285625-85285625 on Build GRCh38
rs990699012 Chr.16: 85285422-85285422 on Build GRCh38
rs543755984 Chr.16: 85285418-85285418 on Build GRCh38
rs1034388381 Chr.16: 85285406-85285406 on Build GRCh38
rs758192852 Chr.16: 85285573-85285573 on Build GRCh38
rs36070534 Chr.16: 85285700-85285700 on Build GRCh38
rs190816595 Chr.16: 85285555-85285555 on Build GRCh38
rs144141235 Chr.16: 85285775-85285775 on Build GRCh38
rs946260309 Chr.16: 85285572-85285572 on Build GRCh38
rs921649288 Chr.16: 85285731-85285731 on Build GRCh38
rs759592515 Chr.16: 85285534-85285534 on Build GRCh38
rs944506352 Chr.16: 85285538-85285538 on Build GRCh38
rs552891003 Chr.16: 85285524-85285524 on Build GRCh38
rs975876865 Chr.16: 85285717-85285717 on Build GRCh38
rs536750157 Chr.16: 85285612-85285612 on Build GRCh38
rs535600780 Chr.16: 85285707-85285707 on Build GRCh38
rs183042827 Chr.16: 85285556-85285556 on Build GRCh38
rs561402164 Chr.16: 85285355-85285355 on Build GRCh38
rs140331264 Chr.16: 85285637-85285637 on Build GRCh38
rs1004749549 Chr.16: 85285628-85285628 on Build GRCh38
rs905706775 Chr.16: 85285389-85285389 on Build GRCh38
rs983450569 Chr.16: 85285601-85285601 on Build GRCh38
rs753967083 Chr.16: 85285515-85285515 on Build GRCh38
rs904209858 Chr.16: 85285764-85285764 on Build GRCh38
rs958922880 Chr.16: 85285419-85285419 on Build GRCh38
rs757656300 Chr.16: 85285545-85285545 on Build GRCh38
rs560117935 Chr.16: 85285543-85285543 on Build GRCh38
rs927350446 Chr.16: 85285643-85285643 on Build GRCh38
rs550387899 Chr.16: 85285697-85285698 on Build GRCh38
rs914235732 Chr.16: 85285742-85285742 on Build GRCh38
rs928821891 Chr.16: 85285769-85285769 on Build GRCh38
rs570103096 Chr.16: 85285660-85285660 on Build GRCh38
rs572820333 Chr.16: 85285697-85285697 on Build GRCh38
rs983272473 Chr.16: 85285511-85285511 on Build GRCh38
rs976205030 Chr.16: 85285542-85285542 on Build GRCh38
rs953093508 Chr.16: 85285732-85285732 on Build GRCh38
rs920577134 Chr.16: 85285682-85285682 on Build GRCh38
rs12447206 Chr.16: 85285793-85285793 on Build GRCh38
rs151188941 Chr.16: 85285559-85285559 on Build GRCh38
rs954981604 Chr.16: 85285599-85285599 on Build GRCh38

View Full Product Details