Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46676731-46677273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DACT3-AS1
Entrez Gene ID
100506068
Gene Name
DACT3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.434327
Gene Chromosome Location
Chr.19: 46660364-46677456 on Build GRCh38
Transcripts
NR_040042.1 NM_001079880.1 NM_001079881.1 NM_001079882.1 NM_016457.4 XM_005258716.2

Gene Symbol
PRKD2
Entrez Gene ID
25865
Gene Name
protein kinase D2
Gene Aliases
HSPC187, PKD2, nPKC-D2
UniGene
Hs.466987
Gene Chromosome Location
Chr.19: 46674316-46717127 on Build GRCh38
Transcripts
NR_040042.1 NM_001079880.1 NM_001079881.1 NM_001079882.1 NM_016457.4 XM_005258716.2

SNP Detail

SNP ID Chromosome Location
rs535016550 Chr.19: 46676975-46676975 on Build GRCh38
rs775041250 Chr.19: 46677093-46677093 on Build GRCh38
rs565774610 Chr.19: 46676925-46676925 on Build GRCh38
rs929317687 Chr.19: 46677170-46677170 on Build GRCh38
rs888007806 Chr.19: 46676780-46676780 on Build GRCh38
rs891514526 Chr.19: 46677235-46677235 on Build GRCh38
rs889287421 Chr.19: 46677058-46677058 on Build GRCh38
rs991308245 Chr.19: 46676823-46676823 on Build GRCh38
rs189032685 Chr.19: 46676847-46676847 on Build GRCh38
rs372081534 Chr.19: 46677183-46677183 on Build GRCh38
rs397772495 Chr.19: 46677075-46677076 on Build GRCh38
rs1033105632 Chr.19: 46677177-46677177 on Build GRCh38
rs533108328 Chr.19: 46676945-46676946 on Build GRCh38
rs570281945 Chr.19: 46676833-46676833 on Build GRCh38
rs149456811 Chr.19: 46677047-46677047 on Build GRCh38
rs545925677 Chr.19: 46677095-46677095 on Build GRCh38
rs757665348 Chr.19: 46676998-46676998 on Build GRCh38
rs889451150 Chr.19: 46677056-46677056 on Build GRCh38
rs966149291 Chr.19: 46677116-46677116 on Build GRCh38
rs909477029 Chr.19: 46677190-46677190 on Build GRCh38
rs577466901 Chr.19: 46677090-46677090 on Build GRCh38
rs115005728 Chr.19: 46677198-46677198 on Build GRCh38
rs998385254 Chr.19: 46677076-46677076 on Build GRCh38
rs906650865 Chr.19: 46676872-46676872 on Build GRCh38
rs779354313 Chr.19: 46677147-46677147 on Build GRCh38
rs1050522290 Chr.19: 46677052-46677052 on Build GRCh38
rs1036339635 Chr.19: 46677248-46677248 on Build GRCh38
rs559482907 Chr.19: 46677159-46677159 on Build GRCh38
rs576016900 Chr.19: 46677209-46677209 on Build GRCh38
rs375459727 Chr.19: 46677186-46677186 on Build GRCh38
rs1053297769 Chr.19: 46677230-46677230 on Build GRCh38
rs534544646 Chr.19: 46677229-46677229 on Build GRCh38
rs1049762576 Chr.19: 46676755-46676755 on Build GRCh38
rs1009053557 Chr.19: 46677082-46677082 on Build GRCh38
rs948904560 Chr.19: 46676829-46676829 on Build GRCh38
rs1046516031 Chr.19: 46676866-46676866 on Build GRCh38
rs953907837 Chr.19: 46676994-46676994 on Build GRCh38
rs575064201 Chr.19: 46677046-46677046 on Build GRCh38
rs530651970 Chr.19: 46676779-46676779 on Build GRCh38
rs11397034 Chr.19: 46677060-46677061 on Build GRCh38
rs769322248 Chr.19: 46676831-46676831 on Build GRCh38
rs762065031 Chr.19: 46677124-46677124 on Build GRCh38
rs998505424 Chr.19: 46677171-46677171 on Build GRCh38
rs1031363953 Chr.19: 46676897-46676897 on Build GRCh38
rs928799550 Chr.19: 46677214-46677214 on Build GRCh38
rs752905714 Chr.19: 46676886-46676886 on Build GRCh38
rs991406654 Chr.19: 46677187-46677187 on Build GRCh38
rs906965663 Chr.19: 46676880-46676880 on Build GRCh38
rs113240194 Chr.19: 46677050-46677050 on Build GRCh38
rs940690044 Chr.19: 46676883-46676883 on Build GRCh38
rs1003033563 Chr.19: 46676881-46676881 on Build GRCh38
rs919290614 Chr.19: 46677166-46677166 on Build GRCh38
rs554993914 Chr.19: 46677034-46677034 on Build GRCh38
rs749596746 Chr.19: 46676984-46676984 on Build GRCh38
rs73563509 Chr.19: 46676924-46676924 on Build GRCh38
rs550766121 Chr.19: 46676830-46676830 on Build GRCh38

View Full Product Details