Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53751099-53751382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR522
Entrez Gene ID
574495
Gene Name
microRNA 522
Gene Aliases
MIRN522, hsa-mir-522, mir-522
UniGene
Gene Chromosome Location
Chr.19: 53751211-53751297 on Build GRCh38
Transcripts
NR_030217.1

SNP Detail

SNP ID Chromosome Location
rs767590469 Chr.19: 53751342-53751342 on Build GRCh38
rs549273588 Chr.19: 53751141-53751141 on Build GRCh38
rs377484755 Chr.19: 53751289-53751289 on Build GRCh38
rs546743311 Chr.19: 53751192-53751192 on Build GRCh38
rs769241317 Chr.19: 53751296-53751296 on Build GRCh38
rs146976572 Chr.19: 53751240-53751240 on Build GRCh38
rs369117827 Chr.19: 53751164-53751164 on Build GRCh38
rs770673662 Chr.19: 53751214-53751214 on Build GRCh38
rs1032971079 Chr.19: 53751357-53751357 on Build GRCh38
rs781379833 Chr.19: 53751288-53751288 on Build GRCh38
rs996166035 Chr.19: 53751348-53751348 on Build GRCh38
rs780536107 Chr.19: 53751196-53751196 on Build GRCh38
rs776468139 Chr.19: 53751217-53751217 on Build GRCh38
rs749809776 Chr.19: 53751201-53751201 on Build GRCh38
rs1048386926 Chr.19: 53751300-53751300 on Build GRCh38
rs567473384 Chr.19: 53751159-53751159 on Build GRCh38
rs557368006 Chr.19: 53751328-53751328 on Build GRCh38
rs762959743 Chr.19: 53751242-53751242 on Build GRCh38
rs757377349 Chr.19: 53751274-53751274 on Build GRCh38
rs370360227 Chr.19: 53751135-53751135 on Build GRCh38
rs373829370 Chr.19: 53751252-53751252 on Build GRCh38
rs896368184 Chr.19: 53751199-53751199 on Build GRCh38
rs1039835788 Chr.19: 53751131-53751131 on Build GRCh38
rs981184928 Chr.19: 53751166-53751166 on Build GRCh38
rs762757708 Chr.19: 53751337-53751337 on Build GRCh38
rs976084203 Chr.19: 53751330-53751330 on Build GRCh38
rs988642327 Chr.19: 53751123-53751123 on Build GRCh38
rs751691225 Chr.19: 53751254-53751254 on Build GRCh38
rs770620730 Chr.19: 53751306-53751306 on Build GRCh38
rs931450736 Chr.19: 53751351-53751351 on Build GRCh38
rs778841097 Chr.19: 53751243-53751243 on Build GRCh38
rs775377924 Chr.19: 53751336-53751336 on Build GRCh38
rs930351564 Chr.19: 53751147-53751147 on Build GRCh38
rs764071637 Chr.19: 53751338-53751338 on Build GRCh38
rs772804973 Chr.19: 53751203-53751203 on Build GRCh38
rs749713294 Chr.19: 53751294-53751294 on Build GRCh38
rs75928006 Chr.19: 53751134-53751134 on Build GRCh38
rs754096494 Chr.19: 53751331-53751331 on Build GRCh38
rs776415174 Chr.19: 53751316-53751316 on Build GRCh38
rs765188254 Chr.19: 53751239-53751239 on Build GRCh38
rs1012578303 Chr.19: 53751235-53751235 on Build GRCh38
rs371123365 Chr.19: 53751293-53751293 on Build GRCh38
rs780407189 Chr.19: 53751345-53751345 on Build GRCh38
rs760209489 Chr.19: 53751209-53751209 on Build GRCh38
rs753180158 Chr.19: 53751309-53751309 on Build GRCh38
rs532981267 Chr.19: 53751355-53751355 on Build GRCh38
rs376051027 Chr.19: 53751202-53751202 on Build GRCh38
rs759389170 Chr.19: 53751222-53751222 on Build GRCh38
rs746210089 Chr.19: 53751193-53751193 on Build GRCh38
rs761878285 Chr.19: 53751340-53751340 on Build GRCh38
rs745519209 Chr.19: 53751327-53751327 on Build GRCh38
rs373849524 Chr.19: 53751179-53751179 on Build GRCh38
rs117785241 Chr.19: 53751339-53751339 on Build GRCh38
rs779324237 Chr.19: 53751301-53751301 on Build GRCh38
rs1009411980 Chr.19: 53751303-53751303 on Build GRCh38
rs372562860 Chr.19: 53751188-53751188 on Build GRCh38
rs766776440 Chr.19: 53751285-53751334 on Build GRCh38
rs746501954 Chr.19: 53751305-53751305 on Build GRCh38

View Full Product Details