Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 72044736-72045285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NHSL2
Entrez Gene ID
340527
Gene Name
NHS like 2
Gene Aliases
-
UniGene
Hs.397836
Gene Chromosome Location
Chr.X: 71911073-72153286 on Build GRCh38
Transcripts
NM_001013627.2 XM_011530932.1 XM_011530933.1 XM_011530934.2 XM_017029477.1 NR_002309.1

Gene Symbol
RPS26P11
Entrez Gene ID
441502
Gene Name
ribosomal protein S26 pseudogene 11
Gene Aliases
RPS26L1, RPS26_22_1784, bA366E13.1
UniGene
Hs.449523
Gene Chromosome Location
Chr.X: 72044409-72044974 on Build GRCh38
Transcripts
NM_001013627.2 XM_011530932.1 XM_011530933.1 XM_011530934.2 XM_017029477.1 NR_002309.1

SNP Detail

SNP ID Chromosome Location
rs921290524 Chr.X: 72045035-72045035 on Build GRCh38
rs907591586 Chr.X: 72044810-72044810 on Build GRCh38
rs982949459 Chr.X: 72044823-72044823 on Build GRCh38
rs199981428 Chr.X: 72044821-72044821 on Build GRCh38
rs762632650 Chr.X: 72044960-72044960 on Build GRCh38
rs951564147 Chr.X: 72044816-72044816 on Build GRCh38
rs961834454 Chr.X: 72044941-72044941 on Build GRCh38
rs369212673 Chr.X: 72044991-72044991 on Build GRCh38
rs975921793 Chr.X: 72044981-72044981 on Build GRCh38
rs1037740407 Chr.X: 72044833-72044833 on Build GRCh38
rs781098415 Chr.X: 72045130-72045130 on Build GRCh38
rs756232050 Chr.X: 72044843-72044843 on Build GRCh38
rs754043168 Chr.X: 72044933-72044933 on Build GRCh38
rs60336217 Chr.X: 72045053-72045053 on Build GRCh38
rs902561246 Chr.X: 72045234-72045234 on Build GRCh38
rs1023370133 Chr.X: 72045152-72045152 on Build GRCh38
rs777763104 Chr.X: 72044869-72044869 on Build GRCh38
rs772632262 Chr.X: 72045210-72045210 on Build GRCh38
rs184617081 Chr.X: 72045119-72045119 on Build GRCh38
rs769117752 Chr.X: 72044854-72044854 on Build GRCh38
rs557165568 Chr.X: 72045028-72045028 on Build GRCh38
rs746817219 Chr.X: 72045089-72045089 on Build GRCh38
rs73635612 Chr.X: 72045131-72045131 on Build GRCh38
rs999361094 Chr.X: 72045092-72045092 on Build GRCh38
rs947399279 Chr.X: 72045042-72045042 on Build GRCh38
rs189586061 Chr.X: 72045194-72045194 on Build GRCh38
rs931678170 Chr.X: 72045043-72045043 on Build GRCh38
rs867541932 Chr.X: 72044770-72044770 on Build GRCh38
rs1052228825 Chr.X: 72045093-72045093 on Build GRCh38
rs1043660181 Chr.X: 72045051-72045051 on Build GRCh38
rs753324212 Chr.X: 72044809-72044809 on Build GRCh38
rs879042092 Chr.X: 72044952-72044952 on Build GRCh38

View Full Product Details