Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 6899447-6900009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX12P2
Entrez Gene ID
245
Gene Name
arachidonate 12-lipoxygenase pseudogene 2
Gene Aliases
ALOX12E
UniGene
Hs.378695
Gene Chromosome Location
Chr.17: 6853576-6900349 on Build GRCh38
Transcripts
NR_002710.2 NR_120453.1

SNP Detail

SNP ID Chromosome Location
rs79546968 Chr.17: 6899819-6899819 on Build GRCh38
rs311757 Chr.17: 6899497-6899497 on Build GRCh38
rs554936664 Chr.17: 6899747-6899747 on Build GRCh38
rs1043824323 Chr.17: 6899914-6899914 on Build GRCh38
rs1056219401 Chr.17: 6899872-6899872 on Build GRCh38
rs978808828 Chr.17: 6899491-6899491 on Build GRCh38
rs1000363937 Chr.17: 6899708-6899708 on Build GRCh38
rs894769883 Chr.17: 6899885-6899885 on Build GRCh38
rs975029211 Chr.17: 6899758-6899758 on Build GRCh38
rs1036331522 Chr.17: 6899682-6899682 on Build GRCh38
rs1031816859 Chr.17: 6899960-6899960 on Build GRCh38
rs1047587521 Chr.17: 6899595-6899595 on Build GRCh38
rs188195725 Chr.17: 6899818-6899818 on Build GRCh38
rs930484495 Chr.17: 6899594-6899594 on Build GRCh38
rs924263408 Chr.17: 6899787-6899787 on Build GRCh38
rs1000481328 Chr.17: 6899954-6899954 on Build GRCh38
rs894437762 Chr.17: 6899596-6899596 on Build GRCh38
rs758342985 Chr.17: 6899695-6899695 on Build GRCh38
rs930600536 Chr.17: 6899763-6899763 on Build GRCh38
rs911928231 Chr.17: 6899527-6899527 on Build GRCh38
rs987564796 Chr.17: 6899740-6899740 on Build GRCh38
rs540465417 Chr.17: 6899815-6899815 on Build GRCh38
rs571296464 Chr.17: 6899786-6899786 on Build GRCh38
rs77797852 Chr.17: 6899508-6899508 on Build GRCh38
rs552969908 Chr.17: 6899871-6899871 on Build GRCh38
rs904812671 Chr.17: 6899696-6899696 on Build GRCh38
rs749713169 Chr.17: 6899590-6899590 on Build GRCh38
rs1027852781 Chr.17: 6899713-6899713 on Build GRCh38
rs957261595 Chr.17: 6899798-6899798 on Build GRCh38
rs746152344 Chr.17: 6899670-6899670 on Build GRCh38
rs528541151 Chr.17: 6899588-6899588 on Build GRCh38
rs184095181 Chr.17: 6899710-6899710 on Build GRCh38
rs61744237 Chr.17: 6899650-6899650 on Build GRCh38
rs769429288 Chr.17: 6899903-6899903 on Build GRCh38
rs977585249 Chr.17: 6899780-6899780 on Build GRCh38
rs1003269469 Chr.17: 6899622-6899622 on Build GRCh38
rs964742712 Chr.17: 6899751-6899751 on Build GRCh38
rs968918184 Chr.17: 6899476-6899476 on Build GRCh38
rs1038966836 Chr.17: 6899574-6899574 on Build GRCh38
rs915818986 Chr.17: 6899898-6899898 on Build GRCh38
rs920513816 Chr.17: 6899759-6899759 on Build GRCh38
rs956097003 Chr.17: 6899726-6899726 on Build GRCh38
rs943250437 Chr.17: 6899571-6899571 on Build GRCh38
rs531237469 Chr.17: 6899976-6899976 on Build GRCh38
rs61746805 Chr.17: 6899677-6899677 on Build GRCh38
rs956149328 Chr.17: 6899511-6899511 on Build GRCh38
rs574674936 Chr.17: 6899784-6899784 on Build GRCh38
rs530757810 Chr.17: 6899985-6899985 on Build GRCh38
rs544495584 Chr.17: 6899746-6899746 on Build GRCh38
rs891985494 Chr.17: 6899962-6899962 on Build GRCh38
rs311755 Chr.17: 6899646-6899646 on Build GRCh38
rs759100379 Chr.17: 6899689-6899689 on Build GRCh38
rs983228882 Chr.17: 6899809-6899809 on Build GRCh38
rs1020662520 Chr.17: 6899631-6899631 on Build GRCh38
rs61746793 Chr.17: 6899605-6899605 on Build GRCh38
rs781722107 Chr.17: 6899482-6899482 on Build GRCh38
rs562479224 Chr.17: 6899953-6899953 on Build GRCh38
rs311756 Chr.17: 6899515-6899515 on Build GRCh38
rs1045566971 Chr.17: 6899961-6899961 on Build GRCh38
rs753095916 Chr.17: 6899484-6899484 on Build GRCh38

View Full Product Details