Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 10716035-10716590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGOH2P
Entrez Gene ID
100506898
Gene Name
mago homolog 2, pseudogene
Gene Aliases
MAGOH2
UniGene
Hs.674373
Gene Chromosome Location
Chr.17: 10715943-10716582 on Build GRCh38
Transcripts
NR_049723.1 NM_001004313.1 XM_005256634.4 XM_011523835.2 XM_011523836.2 XM_017024616.1 XM_017024617.1

Gene Symbol
TMEM220
Entrez Gene ID
388335
Gene Name
transmembrane protein 220
Gene Aliases
-
UniGene
Hs.462230
Gene Chromosome Location
Chr.17: 10711084-10730329 on Build GRCh38
Transcripts
NR_049723.1 NM_001004313.1 XM_005256634.4 XM_011523835.2 XM_011523836.2 XM_017024616.1 XM_017024617.1

SNP Detail

SNP ID Chromosome Location
rs544281820 Chr.17: 10716213-10716213 on Build GRCh38
rs1038019683 Chr.17: 10716566-10716566 on Build GRCh38
rs9674829 Chr.17: 10716534-10716534 on Build GRCh38
rs971628034 Chr.17: 10716428-10716428 on Build GRCh38
rs568425091 Chr.17: 10716360-10716360 on Build GRCh38
rs956180874 Chr.17: 10716484-10716484 on Build GRCh38
rs1000969017 Chr.17: 10716061-10716061 on Build GRCh38
rs549033215 Chr.17: 10716356-10716356 on Build GRCh38
rs923452347 Chr.17: 10716351-10716351 on Build GRCh38
rs112424748 Chr.17: 10716181-10716181 on Build GRCh38
rs1044165807 Chr.17: 10716561-10716561 on Build GRCh38
rs9675034 Chr.17: 10716555-10716555 on Build GRCh38
rs192896641 Chr.17: 10716073-10716073 on Build GRCh38
rs879129029 Chr.17: 10716349-10716349 on Build GRCh38
rs755939891 Chr.17: 10716260-10716260 on Build GRCh38
rs1056897083 Chr.17: 10716290-10716290 on Build GRCh38
rs878925506 Chr.17: 10716319-10716319 on Build GRCh38
rs1022778379 Chr.17: 10716427-10716427 on Build GRCh38
rs886788372 Chr.17: 10716096-10716096 on Build GRCh38
rs1032564658 Chr.17: 10716367-10716367 on Build GRCh38
rs916986325 Chr.17: 10716546-10716546 on Build GRCh38
rs879250050 Chr.17: 10716328-10716328 on Build GRCh38
rs923575765 Chr.17: 10716550-10716550 on Build GRCh38
rs879031303 Chr.17: 10716273-10716273 on Build GRCh38
rs569208246 Chr.17: 10716403-10716403 on Build GRCh38
rs534201030 Chr.17: 10716426-10716426 on Build GRCh38
rs1003907151 Chr.17: 10716149-10716149 on Build GRCh38
rs879206855 Chr.17: 10716156-10716156 on Build GRCh38
rs924750745 Chr.17: 10716240-10716240 on Build GRCh38
rs1012569221 Chr.17: 10716306-10716306 on Build GRCh38
rs1051269793 Chr.17: 10716387-10716387 on Build GRCh38
rs112948764 Chr.17: 10716425-10716425 on Build GRCh38
rs404399 Chr.17: 10716060-10716060 on Build GRCh38
rs403817 Chr.17: 10716266-10716266 on Build GRCh38
rs879108123 Chr.17: 10716243-10716243 on Build GRCh38
rs12452583 Chr.17: 10716198-10716198 on Build GRCh38
rs140520651 Chr.17: 10716491-10716491 on Build GRCh38
rs1044477621 Chr.17: 10716325-10716325 on Build GRCh38
rs79277973 Chr.17: 10716493-10716493 on Build GRCh38
rs879157926 Chr.17: 10716274-10716274 on Build GRCh38
rs192564117 Chr.17: 10716404-10716404 on Build GRCh38
rs878996744 Chr.17: 10716289-10716289 on Build GRCh38
rs148702156 Chr.17: 10716234-10716234 on Build GRCh38
rs142272873 Chr.17: 10716394-10716394 on Build GRCh38
rs774168760 Chr.17: 10716187-10716187 on Build GRCh38
rs939755725 Chr.17: 10716562-10716562 on Build GRCh38
rs770765379 Chr.17: 10716238-10716240 on Build GRCh38
rs1023880264 Chr.17: 10716056-10716056 on Build GRCh38
rs878975799 Chr.17: 10716140-10716140 on Build GRCh38
rs188709860 Chr.17: 10716364-10716364 on Build GRCh38
rs183777926 Chr.17: 10716160-10716160 on Build GRCh38
rs9675020 Chr.17: 10716327-10716327 on Build GRCh38
rs768615526 Chr.17: 10716175-10716175 on Build GRCh38
rs878867948 Chr.17: 10716241-10716241 on Build GRCh38
rs528659598 Chr.17: 10716533-10716533 on Build GRCh38
rs1047927810 Chr.17: 10716558-10716558 on Build GRCh38
rs533083511 Chr.17: 10716239-10716239 on Build GRCh38
rs878864470 Chr.17: 10716182-10716182 on Build GRCh38
rs878971130 Chr.17: 10716128-10716128 on Build GRCh38
rs560425650 Chr.17: 10716292-10716292 on Build GRCh38
rs908256042 Chr.17: 10716560-10716560 on Build GRCh38
rs548806787 Chr.17: 10716068-10716068 on Build GRCh38
rs183679161 Chr.17: 10716477-10716477 on Build GRCh38
rs1016646853 Chr.17: 10716516-10716516 on Build GRCh38
rs879202336 Chr.17: 10716348-10716348 on Build GRCh38
rs9674753 Chr.17: 10716456-10716456 on Build GRCh38
rs879081859 Chr.17: 10716286-10716286 on Build GRCh38
rs767546488 Chr.17: 10716062-10716065 on Build GRCh38
rs775706996 Chr.17: 10716065-10716065 on Build GRCh38
rs990146889 Chr.17: 10716276-10716276 on Build GRCh38
rs760984336 Chr.17: 10716321-10716321 on Build GRCh38
rs759314995 Chr.17: 10716301-10716302 on Build GRCh38
rs1039138461 Chr.17: 10716340-10716340 on Build GRCh38
rs991288682 Chr.17: 10716176-10716176 on Build GRCh38

View Full Product Details