Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75589648-75590225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO15B
Entrez Gene ID
80022
Gene Name
myosin XVB
Gene Aliases
MYO15BP
UniGene
Hs.390817
Gene Chromosome Location
Chr.17: 75587545-75626850 on Build GRCh38
Transcripts
NM_001309242.1 XM_017025120.1 XM_017025121.1 XM_017025122.1 XM_017025123.1 XM_017025124.1 XM_017025125.1 XM_017025126.1 XM_017025127.1 XM_017025128.1 XM_017025141.1 XM_017025142.1 XM_017025144.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3820554 c.381C>G p.R127R Chr.17: 75589965-75589965 on Build GRCh38
84415 c.606G>A p.K202K Chr.17: 75590190-75590190 on Build GRCh38
7226319 c.519C>G p.P173P Chr.17: 75590103-75590103 on Build GRCh38
5573572 c.214G>T p.G72W Chr.17: 75589798-75589798 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs915272486 Chr.17: 75589930-75589930 on Build GRCh38
rs570509344 Chr.17: 75589758-75589758 on Build GRCh38
rs186512072 Chr.17: 75590107-75590107 on Build GRCh38
rs565406713 Chr.17: 75590121-75590121 on Build GRCh38
rs982739623 Chr.17: 75589910-75589910 on Build GRCh38
rs575578227 Chr.17: 75589961-75589961 on Build GRCh38
rs751890508 Chr.17: 75590140-75590140 on Build GRCh38
rs533274601 Chr.17: 75590002-75590002 on Build GRCh38
rs182828502 Chr.17: 75590005-75590005 on Build GRCh38
rs992906371 Chr.17: 75590136-75590136 on Build GRCh38
rs978674042 Chr.17: 75589979-75589979 on Build GRCh38
rs567117658 Chr.17: 75590166-75590166 on Build GRCh38
rs952496232 Chr.17: 75589885-75589885 on Build GRCh38
rs888176289 Chr.17: 75590091-75590091 on Build GRCh38
rs889705836 Chr.17: 75589689-75589689 on Build GRCh38
rs528259589 Chr.17: 75590035-75590035 on Build GRCh38
rs1047392393 Chr.17: 75589977-75589977 on Build GRCh38
rs965216208 Chr.17: 75589781-75589781 on Build GRCh38
rs933792962 Chr.17: 75590125-75590125 on Build GRCh38
rs796442782 Chr.17: 75589915-75589916 on Build GRCh38
rs975507611 Chr.17: 75589954-75589954 on Build GRCh38
rs971420627 Chr.17: 75590147-75590147 on Build GRCh38
rs80175090 Chr.17: 75589964-75589964 on Build GRCh38
rs820241 Chr.17: 75590103-75590103 on Build GRCh38
rs1025355833 Chr.17: 75589841-75589841 on Build GRCh38
rs749590281 Chr.17: 75589700-75589700 on Build GRCh38
rs1038298821 Chr.17: 75590170-75590170 on Build GRCh38
rs1003733793 Chr.17: 75590063-75590063 on Build GRCh38
rs62089224 Chr.17: 75589899-75589899 on Build GRCh38
rs769135244 Chr.17: 75590074-75590074 on Build GRCh38
rs1005229615 Chr.17: 75589801-75589801 on Build GRCh38
rs924130556 Chr.17: 75590157-75590157 on Build GRCh38
rs975434135 Chr.17: 75590120-75590120 on Build GRCh38
rs79599625 Chr.17: 75589916-75589916 on Build GRCh38
rs994427867 Chr.17: 75589803-75589803 on Build GRCh38
rs80017587 Chr.17: 75589915-75589915 on Build GRCh38
rs991057678 Chr.17: 75589958-75589958 on Build GRCh38
rs559134040 Chr.17: 75590027-75590027 on Build GRCh38
rs139077523 Chr.17: 75589672-75589672 on Build GRCh38
rs771040887 Chr.17: 75589702-75589702 on Build GRCh38
rs889821973 Chr.17: 75590014-75590014 on Build GRCh38
rs970889890 Chr.17: 75589900-75589900 on Build GRCh38
rs564292129 Chr.17: 75589971-75589971 on Build GRCh38
rs920081414 Chr.17: 75590022-75590022 on Build GRCh38
rs1040945708 Chr.17: 75590044-75590044 on Build GRCh38
rs781380175 Chr.17: 75590161-75590161 on Build GRCh38
rs28488097 Chr.17: 75590046-75590046 on Build GRCh38
rs983748322 Chr.17: 75589888-75589888 on Build GRCh38
rs62089225 Chr.17: 75590152-75590152 on Build GRCh38
rs372818002 Chr.17: 75590066-75590066 on Build GRCh38
rs79229026 Chr.17: 75590178-75590178 on Build GRCh38
rs1031610303 Chr.17: 75590070-75590070 on Build GRCh38
rs530454803 Chr.17: 75590089-75590089 on Build GRCh38
rs1053770597 Chr.17: 75590008-75590008 on Build GRCh38
rs926984437 Chr.17: 75589936-75589936 on Build GRCh38
rs1022249015 Chr.17: 75590094-75590094 on Build GRCh38
rs539506418 Chr.17: 75589774-75589774 on Build GRCh38
rs911330248 Chr.17: 75590167-75590167 on Build GRCh38
rs145925770 Chr.17: 75590055-75590055 on Build GRCh38
rs72854924 Chr.17: 75589947-75589947 on Build GRCh38
rs1025191937 Chr.17: 75590142-75590142 on Build GRCh38
rs1025936790 Chr.17: 75590081-75590081 on Build GRCh38
rs1012698779 Chr.17: 75589873-75589873 on Build GRCh38
rs556804611 Chr.17: 75589692-75589692 on Build GRCh38
rs1038027674 Chr.17: 75590013-75590013 on Build GRCh38

View Full Product Details