Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75594331-75594880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO15B
Entrez Gene ID
80022
Gene Name
myosin XVB
Gene Aliases
MYO15BP
UniGene
Hs.390817
Gene Chromosome Location
Chr.17: 75587545-75626850 on Build GRCh38
Transcripts
NM_001309242.1 XM_017025120.1 XM_017025121.1 XM_017025122.1 XM_017025123.1 XM_017025124.1 XM_017025125.1 XM_017025126.1 XM_017025127.1 XM_017025128.1 XM_017025129.1 XM_017025130.1 XM_017025131.1 XM_017025132.1 XM_017025133.1 XM_017025134.1 XM_017025135.1 XM_017025141.1 XM_017025142.1 XM_017025144.1

SNP Detail

SNP ID Chromosome Location
rs879024164 Chr.17: 75594819-75594819 on Build GRCh38
rs539506406 Chr.17: 75594827-75594827 on Build GRCh38
rs12602035 Chr.17: 75594774-75594774 on Build GRCh38
rs879067580 Chr.17: 75594823-75594823 on Build GRCh38
rs779183458 Chr.17: 75594760-75594760 on Build GRCh38
rs73998365 Chr.17: 75594745-75594745 on Build GRCh38
rs919525453 Chr.17: 75594707-75594707 on Build GRCh38
rs985139893 Chr.17: 75594681-75594681 on Build GRCh38
rs540016003 Chr.17: 75594564-75594564 on Build GRCh38
rs535745584 Chr.17: 75594619-75594619 on Build GRCh38
rs1001120906 Chr.17: 75594638-75594638 on Build GRCh38
rs963652892 Chr.17: 75594412-75594412 on Build GRCh38
rs1000687236 Chr.17: 75594778-75594778 on Build GRCh38
rs1007016308 Chr.17: 75594803-75594803 on Build GRCh38
rs1019273740 Chr.17: 75594704-75594704 on Build GRCh38
rs182533374 Chr.17: 75594542-75594542 on Build GRCh38
rs747042961 Chr.17: 75594695-75594695 on Build GRCh38
rs73366119 Chr.17: 75594380-75594380 on Build GRCh38
rs965021426 Chr.17: 75594705-75594705 on Build GRCh38
rs555741318 Chr.17: 75594663-75594663 on Build GRCh38
rs768291037 Chr.17: 75594856-75594856 on Build GRCh38
rs377015919 Chr.17: 75594795-75594795 on Build GRCh38
rs537149086 Chr.17: 75594404-75594404 on Build GRCh38
rs820254 Chr.17: 75594355-75594355 on Build GRCh38
rs1012465785 Chr.17: 75594644-75594644 on Build GRCh38
rs886580470 Chr.17: 75594362-75594362 on Build GRCh38
rs548141550 Chr.17: 75594748-75594748 on Build GRCh38
rs820255 Chr.17: 75594441-75594441 on Build GRCh38
rs1030995944 Chr.17: 75594709-75594709 on Build GRCh38
rs922265989 Chr.17: 75594573-75594573 on Build GRCh38
rs924009322 Chr.17: 75594533-75594533 on Build GRCh38
rs73366121 Chr.17: 75594667-75594667 on Build GRCh38
rs1049118604 Chr.17: 75594432-75594432 on Build GRCh38
rs915100290 Chr.17: 75594555-75594555 on Build GRCh38
rs902324435 Chr.17: 75594635-75594635 on Build GRCh38
rs2305524 Chr.17: 75594736-75594736 on Build GRCh38
rs989266610 Chr.17: 75594552-75594552 on Build GRCh38
rs573497101 Chr.17: 75594596-75594596 on Build GRCh38
rs820256 Chr.17: 75594749-75594749 on Build GRCh38
rs929476739 Chr.17: 75594417-75594417 on Build GRCh38
rs989528847 Chr.17: 75594726-75594726 on Build GRCh38
rs1005339474 Chr.17: 75594631-75594631 on Build GRCh38
rs559256984 Chr.17: 75594740-75594740 on Build GRCh38
rs886843244 Chr.17: 75594597-75594597 on Build GRCh38
rs550723313 Chr.17: 75594792-75594792 on Build GRCh38
rs757788925 Chr.17: 75594816-75594816 on Build GRCh38
rs76361517 Chr.17: 75594807-75594807 on Build GRCh38
rs971144225 Chr.17: 75594476-75594476 on Build GRCh38
rs140580829 Chr.17: 75594724-75594724 on Build GRCh38
rs749254421 Chr.17: 75594541-75594541 on Build GRCh38
rs1042300828 Chr.17: 75594773-75594773 on Build GRCh38
rs576663140 Chr.17: 75594725-75594725 on Build GRCh38
rs889533899 Chr.17: 75594797-75594797 on Build GRCh38
rs933661136 Chr.17: 75594587-75594587 on Build GRCh38
rs73366123 Chr.17: 75594706-75594706 on Build GRCh38
rs759038233 Chr.17: 75594802-75594802 on Build GRCh38
rs761237274 Chr.17: 75594661-75594661 on Build GRCh38
rs548192257 Chr.17: 75594379-75594379 on Build GRCh38
rs1012950854 Chr.17: 75594424-75594424 on Build GRCh38
rs1019325600 Chr.17: 75594804-75594804 on Build GRCh38
rs993804334 Chr.17: 75594843-75594843 on Build GRCh38
rs1040036841 Chr.17: 75594565-75594565 on Build GRCh38
rs1055576265 Chr.17: 75594532-75594532 on Build GRCh38
rs938011724 Chr.17: 75594482-75594482 on Build GRCh38
rs34390451 Chr.17: 75594816-75594816 on Build GRCh38
rs909298889 Chr.17: 75594737-75594737 on Build GRCh38
rs948202331 Chr.17: 75594834-75594834 on Build GRCh38
rs553735826 Chr.17: 75594588-75594588 on Build GRCh38

View Full Product Details