Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 35307917-35308485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF271P
Entrez Gene ID
10778
Gene Name
zinc finger protein 271, pseudogene
Gene Aliases
CT-ZFP48, HZF7, ZNF-EB, ZNF-dp, ZNF271, ZNFEB
UniGene
Hs.314246
Gene Chromosome Location
Chr.18: 35290272-35310766 on Build GRCh38
Transcripts
NR_024565.1 NR_024566.1

SNP Detail

SNP ID Chromosome Location
rs758929128 Chr.18: 35308433-35308434 on Build GRCh38
rs63352060 Chr.18: 35308433-35308433 on Build GRCh38
rs1012612709 Chr.18: 35308431-35308433 on Build GRCh38
rs899240837 Chr.18: 35307983-35307983 on Build GRCh38
rs573948669 Chr.18: 35308412-35308412 on Build GRCh38
rs897682195 Chr.18: 35308443-35308443 on Build GRCh38
rs1051690900 Chr.18: 35308175-35308175 on Build GRCh38
rs143499122 Chr.18: 35308132-35308132 on Build GRCh38
rs67506491 Chr.18: 35308438-35308439 on Build GRCh38
rs569507405 Chr.18: 35308122-35308122 on Build GRCh38
rs547644524 Chr.18: 35308029-35308029 on Build GRCh38
rs991534214 Chr.18: 35308111-35308111 on Build GRCh38
rs9963614 Chr.18: 35307969-35307969 on Build GRCh38
rs1044789359 Chr.18: 35308293-35308293 on Build GRCh38
rs543036798 Chr.18: 35308428-35308428 on Build GRCh38
rs966695446 Chr.18: 35308060-35308060 on Build GRCh38
rs914646275 Chr.18: 35308232-35308232 on Build GRCh38
rs375931134 Chr.18: 35308012-35308012 on Build GRCh38
rs543670772 Chr.18: 35308313-35308313 on Build GRCh38
rs993262078 Chr.18: 35307981-35307981 on Build GRCh38
rs1054492126 Chr.18: 35308197-35308197 on Build GRCh38
rs559845098 Chr.18: 35308005-35308005 on Build GRCh38
rs1039106565 Chr.18: 35307982-35307982 on Build GRCh38
rs985782859 Chr.18: 35308011-35308011 on Build GRCh38
rs943410412 Chr.18: 35308215-35308215 on Build GRCh38
rs915692958 Chr.18: 35308122-35308123 on Build GRCh38
rs531363820 Chr.18: 35308396-35308396 on Build GRCh38
rs905102174 Chr.18: 35307960-35307960 on Build GRCh38
rs1027383658 Chr.18: 35307991-35307991 on Build GRCh38
rs758435017 Chr.18: 35307943-35307943 on Build GRCh38
rs143732419 Chr.18: 35308252-35308252 on Build GRCh38
rs577821906 Chr.18: 35308292-35308292 on Build GRCh38
rs140575083 Chr.18: 35308153-35308153 on Build GRCh38
rs968491040 Chr.18: 35308434-35308434 on Build GRCh38
rs1035268774 Chr.18: 35307976-35307976 on Build GRCh38
rs1022636264 Chr.18: 35308432-35308432 on Build GRCh38
rs751727446 Chr.18: 35307946-35307946 on Build GRCh38
rs557272911 Chr.18: 35308352-35308352 on Build GRCh38
rs1131709 Chr.18: 35307950-35307950 on Build GRCh38
rs1003805811 Chr.18: 35308073-35308073 on Build GRCh38
rs1047768998 Chr.18: 35308319-35308319 on Build GRCh38
rs199707435 Chr.18: 35307978-35307978 on Build GRCh38
rs1026436366 Chr.18: 35308019-35308021 on Build GRCh38
rs12965288 Chr.18: 35308126-35308126 on Build GRCh38
rs994872156 Chr.18: 35308317-35308317 on Build GRCh38
rs549200273 Chr.18: 35307980-35307980 on Build GRCh38
rs982934948 Chr.18: 35308416-35308416 on Build GRCh38
rs764184166 Chr.18: 35307945-35307945 on Build GRCh38
rs537706605 Chr.18: 35308431-35308432 on Build GRCh38
rs768296502 Chr.18: 35308275-35308275 on Build GRCh38
rs951718743 Chr.18: 35307994-35307994 on Build GRCh38
rs74831732 Chr.18: 35308437-35308437 on Build GRCh38
rs1035291112 Chr.18: 35308427-35308427 on Build GRCh38
rs776696346 Chr.18: 35308110-35308111 on Build GRCh38
rs539715726 Chr.18: 35308086-35308086 on Build GRCh38
rs778856006 Chr.18: 35308368-35308368 on Build GRCh38
rs572131632 Chr.18: 35308139-35308139 on Build GRCh38
rs911777101 Chr.18: 35308193-35308193 on Build GRCh38
rs550101881 Chr.18: 35308093-35308093 on Build GRCh38
rs34303074 Chr.18: 35308445-35308446 on Build GRCh38
rs143423913 Chr.18: 35308437-35308438 on Build GRCh38
rs146487298 Chr.18: 35308042-35308042 on Build GRCh38
rs79828126 Chr.18: 35308461-35308461 on Build GRCh38
rs533973173 Chr.18: 35308084-35308084 on Build GRCh38
rs886611105 Chr.18: 35308337-35308337 on Build GRCh38
rs899293418 Chr.18: 35308315-35308315 on Build GRCh38
rs563299111 Chr.18: 35307967-35307967 on Build GRCh38
rs201643077 Chr.18: 35308126-35308126 on Build GRCh38
rs978517663 Chr.18: 35308434-35308435 on Build GRCh38
rs12607752 Chr.18: 35308431-35308431 on Build GRCh38
rs747796263 Chr.18: 35308342-35308342 on Build GRCh38

View Full Product Details