Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 108561687-108561841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXO3
Entrez Gene ID
2309
Gene Name
forkhead box O3
Gene Aliases
AF6q21, FKHRL1, FKHRL1P2, FOXO2, FOXO3A
UniGene
Hs.220950
Gene Chromosome Location
Chr.6: 108559823-108684769 on Build GRCh38
Transcripts
NM_001455.3 NM_201559.2 XM_005266867.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5702574 c.583A>T p.K195* Chr.6: 108561791-108561791 on Build GRCh38
5898881 c.520C>T p.P174S Chr.6: 108561728-108561728 on Build GRCh38
4769171 c.553G>A p.E185K Chr.6: 108561761-108561761 on Build GRCh38
35779 c.492C>T p.D164D Chr.6: 108561700-108561700 on Build GRCh38
6401622 c.503G>A p.R168H Chr.6: 108561711-108561711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777525804 Chr.6: 108561781-108561781 on Build GRCh38
rs142429317 Chr.6: 108561784-108561784 on Build GRCh38
rs754777172 Chr.6: 108561772-108561772 on Build GRCh38
rs369304108 Chr.6: 108561773-108561773 on Build GRCh38
rs765632505 Chr.6: 108561735-108561735 on Build GRCh38
rs749997186 Chr.6: 108561736-108561736 on Build GRCh38
rs764495520 Chr.6: 108561727-108561727 on Build GRCh38
rs759514852 Chr.6: 108561711-108561711 on Build GRCh38
rs756083158 Chr.6: 108561778-108561778 on Build GRCh38
rs567562629 Chr.6: 108561798-108561798 on Build GRCh38
rs761129538 Chr.6: 108561718-108561718 on Build GRCh38
rs766086478 Chr.6: 108561760-108561760 on Build GRCh38
rs536376745 Chr.6: 108561724-108561724 on Build GRCh38
rs550645016 Chr.6: 108561715-108561715 on Build GRCh38
rs961995451 Chr.6: 108561814-108561814 on Build GRCh38
rs149189425 Chr.6: 108561730-108561730 on Build GRCh38
rs150320900 Chr.6: 108561712-108561712 on Build GRCh38
rs747929839 Chr.6: 108561774-108561774 on Build GRCh38
rs373503752 Chr.6: 108561802-108561802 on Build GRCh38
rs747292650 Chr.6: 108561820-108561820 on Build GRCh38
rs751106449 Chr.6: 108561770-108561770 on Build GRCh38
rs757795843 Chr.6: 108561742-108561742 on Build GRCh38
rs771982912 Chr.6: 108561787-108561787 on Build GRCh38
rs375610127 Chr.6: 108561746-108561746 on Build GRCh38

View Full Product Details