Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 47287002-47287127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGB3
Entrez Gene ID
3690
Gene Name
integrin subunit beta 3
Gene Aliases
BDPLT16, BDPLT2, CD61, GP3A, GPIIIa, GT
UniGene
Hs.218040
Gene Chromosome Location
Chr.17: 47253842-47312711 on Build GRCh38
Transcripts
NM_000212.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
980418 c.783G>A p.K261K Chr.17: 47287075-47287075 on Build GRCh38
1588953 c.642G>A p.K214K Chr.17: 47287075-47287075 on Build GRCh38
5551975 c.688G>A p.D230N Chr.17: 47287121-47287121 on Build GRCh38
7204966 c.788G>T p.G263V Chr.17: 47287080-47287080 on Build GRCh38
980419 c.801T>C p.D267D Chr.17: 47287093-47287093 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765922492 Chr.17: 47287102-47287102 on Build GRCh38
rs779788908 Chr.17: 47287060-47287060 on Build GRCh38
rs751784488 Chr.17: 47287033-47287033 on Build GRCh38
rs769762003 Chr.17: 47287055-47287055 on Build GRCh38
rs780737952 Chr.17: 47287106-47287106 on Build GRCh38
rs781395375 Chr.17: 47287048-47287048 on Build GRCh38
rs753092292 Chr.17: 47287106-47287106 on Build GRCh38
rs749261962 Chr.17: 47287068-47287068 on Build GRCh38
rs758627480 Chr.17: 47287028-47287028 on Build GRCh38
rs745999570 Chr.17: 47287053-47287053 on Build GRCh38
rs757586917 Chr.17: 47287039-47287039 on Build GRCh38
rs761425776 Chr.17: 47287085-47287085 on Build GRCh38
rs397752941 Chr.17: 47287030-47287034 on Build GRCh38
rs56197296 Chr.17: 47287025-47287029 on Build GRCh38
rs760245142 Chr.17: 47287100-47287100 on Build GRCh38
rs772696780 Chr.17: 47287094-47287094 on Build GRCh38
rs771763693 Chr.17: 47287088-47287088 on Build GRCh38
rs200636313 Chr.17: 47287074-47287074 on Build GRCh38
rs368829511 Chr.17: 47287079-47287079 on Build GRCh38

View Full Product Details