Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16231871-16232140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RSG1
Entrez Gene ID
79363
Gene Name
REM2 and RAB like small GTPase 1
Gene Aliases
C1orf89
UniGene
Hs.546430
Gene Chromosome Location
Chr.1: 16231687-16237445 on Build GRCh38
Transcripts
NM_030907.3 XM_011542126.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6218398 c.693C>A p.L231L Chr.1: 16232132-16232132 on Build GRCh38
6770042 c.687C>T p.H229H Chr.1: 16232138-16232138 on Build GRCh38
2088980 c.758delC p.P253fs*>6 Chr.1: 16232067-16232067 on Build GRCh38
1687068 c.739G>A p.G247S Chr.1: 16232086-16232086 on Build GRCh38
7416756 c.758C>A p.P253Q Chr.1: 16232067-16232067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747336256 Chr.1: 16232030-16232030 on Build GRCh38
rs916285193 Chr.1: 16231948-16231948 on Build GRCh38
rs386628905 Chr.1: 16231903-16231905 on Build GRCh38
rs1036914533 Chr.1: 16232059-16232060 on Build GRCh38
rs7512391 Chr.1: 16231998-16231998 on Build GRCh38
rs778226251 Chr.1: 16232026-16232026 on Build GRCh38
rs200415347 Chr.1: 16232016-16232016 on Build GRCh38
rs761587443 Chr.1: 16231999-16231999 on Build GRCh38
rs774313119 Chr.1: 16232084-16232084 on Build GRCh38
rs374755961 Chr.1: 16232105-16232105 on Build GRCh38
rs768436972 Chr.1: 16232080-16232080 on Build GRCh38
rs766009 Chr.1: 16232036-16232036 on Build GRCh38
rs371168864 Chr.1: 16232067-16232067 on Build GRCh38
rs755740011 Chr.1: 16232003-16232003 on Build GRCh38
rs748395836 Chr.1: 16232012-16232019 on Build GRCh38
rs200866605 Chr.1: 16232090-16232090 on Build GRCh38
rs555605675 Chr.1: 16232005-16232005 on Build GRCh38
rs1025770352 Chr.1: 16231896-16231896 on Build GRCh38
rs557518995 Chr.1: 16232118-16232118 on Build GRCh38
rs769774905 Chr.1: 16232016-16232018 on Build GRCh38
rs368799603 Chr.1: 16232019-16232019 on Build GRCh38
rs927949175 Chr.1: 16231991-16231991 on Build GRCh38
rs745616121 Chr.1: 16232012-16232012 on Build GRCh38
rs112722264 Chr.1: 16231903-16231903 on Build GRCh38
rs751287184 Chr.1: 16231902-16231902 on Build GRCh38
rs202169073 Chr.1: 16232013-16232013 on Build GRCh38
rs960699013 Chr.1: 16231913-16231913 on Build GRCh38
rs756140562 Chr.1: 16232025-16232025 on Build GRCh38
rs923387634 Chr.1: 16232062-16232062 on Build GRCh38
rs760867313 Chr.1: 16232068-16232074 on Build GRCh38
rs769928556 Chr.1: 16232056-16232056 on Build GRCh38
rs757589996 Chr.1: 16232042-16232042 on Build GRCh38
rs767466101 Chr.1: 16231932-16231932 on Build GRCh38
rs539532723 Chr.1: 16231933-16231933 on Build GRCh38
rs779939058 Chr.1: 16232117-16232117 on Build GRCh38
rs948117964 Chr.1: 16231935-16231935 on Build GRCh38
rs781443370 Chr.1: 16232043-16232043 on Build GRCh38
rs773516667 Chr.1: 16232019-16232020 on Build GRCh38
rs749270176 Chr.1: 16232073-16232073 on Build GRCh38
rs746078776 Chr.1: 16232048-16232048 on Build GRCh38
rs370375688 Chr.1: 16232002-16232002 on Build GRCh38
rs553153055 Chr.1: 16231972-16231972 on Build GRCh38
rs775366241 Chr.1: 16232061-16232061 on Build GRCh38
rs760333794 Chr.1: 16232107-16232107 on Build GRCh38
rs111505956 Chr.1: 16231905-16231905 on Build GRCh38
rs772438808 Chr.1: 16232017-16232017 on Build GRCh38
rs112234564 Chr.1: 16231904-16231904 on Build GRCh38
rs758777004 Chr.1: 16232020-16232020 on Build GRCh38
rs201135700 Chr.1: 16232012-16232012 on Build GRCh38
rs368888277 Chr.1: 16232098-16232098 on Build GRCh38

View Full Product Details