Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5059048-5059307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR52E2
Entrez Gene ID
119678
Gene Name
olfactory receptor family 52 subfamily E member 2
Gene Aliases
-
UniGene
Hs.553557
Gene Chromosome Location
Chr.11: 5058650-5059627 on Build GRCh38
Transcripts
NM_001005164.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4193212 c.472G>T p.V158F Chr.11: 5059156-5059156 on Build GRCh38
7133221 c.396C>T p.L132L Chr.11: 5059232-5059232 on Build GRCh38
4193210 c.487C>T p.L163F Chr.11: 5059141-5059141 on Build GRCh38
4193209 c.497T>A p.L166* Chr.11: 5059131-5059131 on Build GRCh38
1218732 c.575C>T p.A192V Chr.11: 5059053-5059053 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768543520 Chr.11: 5059235-5059235 on Build GRCh38
rs778319984 Chr.11: 5059149-5059149 on Build GRCh38
rs555360095 Chr.11: 5059222-5059222 on Build GRCh38
rs933132067 Chr.11: 5059221-5059221 on Build GRCh38
rs201816136 Chr.11: 5059269-5059269 on Build GRCh38
rs139088800 Chr.11: 5059255-5059255 on Build GRCh38
rs755313495 Chr.11: 5059205-5059205 on Build GRCh38
rs149502730 Chr.11: 5059135-5059135 on Build GRCh38
rs768625547 Chr.11: 5059170-5059170 on Build GRCh38
rs746712007 Chr.11: 5059226-5059226 on Build GRCh38
rs11035396 Chr.11: 5059129-5059129 on Build GRCh38
rs779563789 Chr.11: 5059224-5059224 on Build GRCh38
rs202028175 Chr.11: 5059254-5059254 on Build GRCh38
rs756634094 Chr.11: 5059217-5059217 on Build GRCh38
rs867909962 Chr.11: 5059168-5059168 on Build GRCh38
rs201562104 Chr.11: 5059073-5059073 on Build GRCh38
rs745479340 Chr.11: 5059156-5059156 on Build GRCh38
rs762937850 Chr.11: 5059265-5059265 on Build GRCh38
rs768909055 Chr.11: 5059082-5059082 on Build GRCh38
rs751866819 Chr.11: 5059204-5059204 on Build GRCh38
rs1004634666 Chr.11: 5059274-5059274 on Build GRCh38
rs776684305 Chr.11: 5059175-5059175 on Build GRCh38
rs114382179 Chr.11: 5059188-5059188 on Build GRCh38
rs934796412 Chr.11: 5059163-5059163 on Build GRCh38
rs1016097226 Chr.11: 5059091-5059091 on Build GRCh38
rs886183051 Chr.11: 5059257-5059257 on Build GRCh38
rs61746343 Chr.11: 5059107-5059107 on Build GRCh38
rs139722116 Chr.11: 5059157-5059157 on Build GRCh38
rs769633161 Chr.11: 5059253-5059253 on Build GRCh38
rs148255884 Chr.11: 5059176-5059176 on Build GRCh38
rs761795642 Chr.11: 5059250-5059250 on Build GRCh38
rs546949115 Chr.11: 5059080-5059080 on Build GRCh38
rs780415265 Chr.11: 5059128-5059128 on Build GRCh38
rs776601165 Chr.11: 5059249-5059249 on Build GRCh38
rs532542281 Chr.11: 5059208-5059208 on Build GRCh38
rs759152629 Chr.11: 5059087-5059087 on Build GRCh38
rs550308989 Chr.11: 5059117-5059117 on Build GRCh38
rs753160860 Chr.11: 5059209-5059209 on Build GRCh38
rs775173726 Chr.11: 5059098-5059098 on Build GRCh38
rs947283709 Chr.11: 5059187-5059187 on Build GRCh38
rs200599599 Chr.11: 5059167-5059167 on Build GRCh38
rs143140208 Chr.11: 5059169-5059169 on Build GRCh38
rs1038869417 Chr.11: 5059212-5059212 on Build GRCh38
rs74580557 Chr.11: 5059174-5059174 on Build GRCh38
rs371504143 Chr.11: 5059223-5059223 on Build GRCh38
rs573919932 Chr.11: 5059261-5059261 on Build GRCh38
rs866037161 Chr.11: 5059108-5059108 on Build GRCh38
rs1023109054 Chr.11: 5059136-5059136 on Build GRCh38
rs781656257 Chr.11: 5059134-5059134 on Build GRCh38
rs868023976 Chr.11: 5059150-5059150 on Build GRCh38
rs760636178 Chr.11: 5059103-5059103 on Build GRCh38
rs761967502 Chr.11: 5059113-5059113 on Build GRCh38
rs199913185 Chr.11: 5059200-5059200 on Build GRCh38
rs750625694 Chr.11: 5059121-5059121 on Build GRCh38
rs770398508 Chr.11: 5059144-5059144 on Build GRCh38
rs750489881 Chr.11: 5059183-5059183 on Build GRCh38
rs758578357 Chr.11: 5059127-5059127 on Build GRCh38
rs568665891 Chr.11: 5059106-5059106 on Build GRCh38
rs369203263 Chr.11: 5059071-5059071 on Build GRCh38
rs767305554 Chr.11: 5059095-5059095 on Build GRCh38
rs141829103 Chr.11: 5059186-5059186 on Build GRCh38
rs994601079 Chr.11: 5059239-5059239 on Build GRCh38

View Full Product Details