Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60891101-60891354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPF19
Entrez Gene ID
27339
Gene Name
pre-mRNA processing factor 19
Gene Aliases
NMP200, PRP19, PSO4, SNEV, UBOX4, hPSO4
UniGene
Hs.502705
Gene Chromosome Location
Chr.11: 60890547-60906589 on Build GRCh38
Transcripts
NM_014502.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6756541 c.1459G>A p.A487T Chr.11: 60891222-60891222 on Build GRCh38
5588730 c.1450G>T p.G484W Chr.11: 60891231-60891231 on Build GRCh38
467073 c.1458C>T p.H486H Chr.11: 60891223-60891223 on Build GRCh38
467072 c.1475C>T p.S492L Chr.11: 60891206-60891206 on Build GRCh38
6461924 c.1477A>T p.T493S Chr.11: 60891204-60891204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538227816 Chr.11: 60891165-60891165 on Build GRCh38
rs750597480 Chr.11: 60891129-60891129 on Build GRCh38
rs902243740 Chr.11: 60891323-60891323 on Build GRCh38
rs777394415 Chr.11: 60891217-60891217 on Build GRCh38
rs866783506 Chr.11: 60891200-60891200 on Build GRCh38
rs73491093 Chr.11: 60891326-60891326 on Build GRCh38
rs769182079 Chr.11: 60891255-60891255 on Build GRCh38
rs747422727 Chr.11: 60891240-60891240 on Build GRCh38
rs754825717 Chr.11: 60891151-60891151 on Build GRCh38
rs148949526 Chr.11: 60891259-60891259 on Build GRCh38
rs201221725 Chr.11: 60891228-60891228 on Build GRCh38
rs779945636 Chr.11: 60891271-60891272 on Build GRCh38
rs763129944 Chr.11: 60891288-60891288 on Build GRCh38
rs751325779 Chr.11: 60891223-60891223 on Build GRCh38
rs766432239 Chr.11: 60891147-60891147 on Build GRCh38
rs751386474 Chr.11: 60891148-60891148 on Build GRCh38
rs1046163865 Chr.11: 60891310-60891310 on Build GRCh38
rs976202914 Chr.11: 60891145-60891145 on Build GRCh38
rs140004740 Chr.11: 60891222-60891222 on Build GRCh38
rs923323582 Chr.11: 60891211-60891211 on Build GRCh38
rs149803092 Chr.11: 60891232-60891232 on Build GRCh38
rs565742443 Chr.11: 60891237-60891237 on Build GRCh38
rs747946929 Chr.11: 60891178-60891178 on Build GRCh38
rs145748444 Chr.11: 60891253-60891253 on Build GRCh38
rs146560356 Chr.11: 60891214-60891214 on Build GRCh38
rs574828394 Chr.11: 60891317-60891317 on Build GRCh38
rs370358090 Chr.11: 60891289-60891289 on Build GRCh38
rs370148247 Chr.11: 60891139-60891139 on Build GRCh38
rs754912835 Chr.11: 60891309-60891309 on Build GRCh38
rs780899009 Chr.11: 60891173-60891173 on Build GRCh38
rs377348006 Chr.11: 60891256-60891256 on Build GRCh38
rs375790221 Chr.11: 60891127-60891127 on Build GRCh38
rs761987547 Chr.11: 60891123-60891123 on Build GRCh38
rs768462314 Chr.11: 60891276-60891276 on Build GRCh38
rs748979010 Chr.11: 60891273-60891277 on Build GRCh38
rs773510292 Chr.11: 60891283-60891283 on Build GRCh38
rs751637973 Chr.11: 60891301-60891301 on Build GRCh38

View Full Product Details