Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 74099157-74099382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIMK1
Entrez Gene ID
3984
Gene Name
LIM domain kinase 1
Gene Aliases
LIMK, LIMK-1
UniGene
Hs.647035
Gene Chromosome Location
Chr.7: 74083777-74122525 on Build GRCh38
Transcripts
NM_001204426.1 NM_002314.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4613729 c.446_447insC p.H152fs*55 Chr.7: 74099178-74099179 on Build GRCh38
4613728 c.548_549insC p.H186fs*55 Chr.7: 74099178-74099179 on Build GRCh38
4551459 c.529G>A p.G177R Chr.7: 74099159-74099159 on Build GRCh38
3929173 c.535T>C p.S179P Chr.7: 74099165-74099165 on Build GRCh38
5351439 c.604C>T p.Q202* Chr.7: 74099234-74099234 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190112531 Chr.7: 74099351-74099351 on Build GRCh38
rs782170750 Chr.7: 74099215-74099215 on Build GRCh38
rs781958441 Chr.7: 74099230-74099230 on Build GRCh38
rs548918526 Chr.7: 74099265-74099265 on Build GRCh38
rs782340126 Chr.7: 74099259-74099259 on Build GRCh38
rs545304197 Chr.7: 74099182-74099182 on Build GRCh38
rs782226834 Chr.7: 74099208-74099208 on Build GRCh38
rs201807030 Chr.7: 74099249-74099249 on Build GRCh38
rs782103354 Chr.7: 74099185-74099185 on Build GRCh38
rs908667284 Chr.7: 74099183-74099183 on Build GRCh38
rs529365882 Chr.7: 74099269-74099269 on Build GRCh38
rs201008021 Chr.7: 74099209-74099209 on Build GRCh38
rs376215118 Chr.7: 74099283-74099283 on Build GRCh38
rs373252592 Chr.7: 74099278-74099278 on Build GRCh38
rs143025582 Chr.7: 74099197-74099197 on Build GRCh38
rs961541559 Chr.7: 74099332-74099332 on Build GRCh38
rs781917913 Chr.7: 74099181-74099181 on Build GRCh38
rs781996424 Chr.7: 74099285-74099285 on Build GRCh38
rs782668678 Chr.7: 74099255-74099255 on Build GRCh38
rs982615750 Chr.7: 74099339-74099339 on Build GRCh38
rs782691662 Chr.7: 74099231-74099231 on Build GRCh38
rs138470279 Chr.7: 74099189-74099189 on Build GRCh38
rs908692745 Chr.7: 74099317-74099317 on Build GRCh38
rs568492420 Chr.7: 74099266-74099266 on Build GRCh38
rs35827364 Chr.7: 74099199-74099199 on Build GRCh38
rs781853522 Chr.7: 74099245-74099245 on Build GRCh38
rs781800461 Chr.7: 74099250-74099250 on Build GRCh38
rs782415086 Chr.7: 74099224-74099224 on Build GRCh38
rs782488303 Chr.7: 74099248-74099248 on Build GRCh38
rs149379462 Chr.7: 74099237-74099237 on Build GRCh38
rs782366559 Chr.7: 74099228-74099228 on Build GRCh38
rs782509930 Chr.7: 74099257-74099257 on Build GRCh38
rs782262510 Chr.7: 74099322-74099322 on Build GRCh38
rs558423798 Chr.7: 74099356-74099356 on Build GRCh38
rs782443942 Chr.7: 74099254-74099254 on Build GRCh38
rs782587891 Chr.7: 74099210-74099210 on Build GRCh38
rs782775113 Chr.7: 74099239-74099239 on Build GRCh38
rs377130473 Chr.7: 74099256-74099256 on Build GRCh38
rs782321094 Chr.7: 74099180-74099180 on Build GRCh38
rs782631807 Chr.7: 74099205-74099205 on Build GRCh38
rs1054972138 Chr.7: 74099200-74099200 on Build GRCh38
rs568894545 Chr.7: 74099309-74099309 on Build GRCh38
rs782626523 Chr.7: 74099260-74099260 on Build GRCh38

View Full Product Details