Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 4653039-4653312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR51E1
Entrez Gene ID
143503
Gene Name
olfactory receptor family 51 subfamily E member 1
Gene Aliases
D-GPCR, DGPCR, GPR136, GPR164, OR51E1P, OR52A3P, POGR, PSGR2
UniGene
Hs.470038
Gene Chromosome Location
Chr.11: 4643926-4655488 on Build GRCh38
Transcripts
NM_152430.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3869199 c.522C>G p.S174S Chr.11: 4653048-4653048 on Build GRCh38
3869200 c.577G>A p.D193N Chr.11: 4653103-4653103 on Build GRCh38
50354 c.730G>A p.V244I Chr.11: 4653256-4653256 on Build GRCh38
6449069 c.618C>T p.I206I Chr.11: 4653144-4653144 on Build GRCh38
50352 c.560T>C p.V187A Chr.11: 4653086-4653086 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367935828 Chr.11: 4653224-4653224 on Build GRCh38
rs377651331 Chr.11: 4653112-4653112 on Build GRCh38
rs751967789 Chr.11: 4653138-4653138 on Build GRCh38
rs758870664 Chr.11: 4653120-4653120 on Build GRCh38
rs757719662 Chr.11: 4653239-4653239 on Build GRCh38
rs771264711 Chr.11: 4653081-4653081 on Build GRCh38
rs370898634 Chr.11: 4653151-4653151 on Build GRCh38
rs139501861 Chr.11: 4653113-4653113 on Build GRCh38
rs760197684 Chr.11: 4653123-4653123 on Build GRCh38
rs768628456 Chr.11: 4653165-4653165 on Build GRCh38
rs202077605 Chr.11: 4653124-4653124 on Build GRCh38
rs781489236 Chr.11: 4653084-4653084 on Build GRCh38
rs752366914 Chr.11: 4653113-4653113 on Build GRCh38
rs769797161 Chr.11: 4653148-4653148 on Build GRCh38
rs895645238 Chr.11: 4653156-4653156 on Build GRCh38
rs765818486 Chr.11: 4653204-4653204 on Build GRCh38
rs769820969 Chr.11: 4653092-4653092 on Build GRCh38
rs749981251 Chr.11: 4653121-4653121 on Build GRCh38
rs753374872 Chr.11: 4653205-4653205 on Build GRCh38
rs760306627 Chr.11: 4653183-4653183 on Build GRCh38
rs769296286 Chr.11: 4653238-4653238 on Build GRCh38
rs746040579 Chr.11: 4653144-4653144 on Build GRCh38
rs745449026 Chr.11: 4653208-4653208 on Build GRCh38
rs749385814 Chr.11: 4653265-4653265 on Build GRCh38
rs774202501 Chr.11: 4653166-4653166 on Build GRCh38
rs775313508 Chr.11: 4653104-4653104 on Build GRCh38
rs755337469 Chr.11: 4653085-4653085 on Build GRCh38
rs375308697 Chr.11: 4653174-4653174 on Build GRCh38
rs971561234 Chr.11: 4653259-4653259 on Build GRCh38
rs781544216 Chr.11: 4653140-4653140 on Build GRCh38
rs148787592 Chr.11: 4653256-4653256 on Build GRCh38
rs143466981 Chr.11: 4653119-4653119 on Build GRCh38
rs144348112 Chr.11: 4653255-4653255 on Build GRCh38
rs140408314 Chr.11: 4653158-4653158 on Build GRCh38
rs185465942 Chr.11: 4653139-4653139 on Build GRCh38
rs373676440 Chr.11: 4653269-4653269 on Build GRCh38
rs149601817 Chr.11: 4653223-4653223 on Build GRCh38
rs572635031 Chr.11: 4653173-4653173 on Build GRCh38
rs763435811 Chr.11: 4653212-4653212 on Build GRCh38
rs758880726 Chr.11: 4653132-4653132 on Build GRCh38
rs867460618 Chr.11: 4653080-4653080 on Build GRCh38
rs761569511 Chr.11: 4653172-4653172 on Build GRCh38
rs772879053 Chr.11: 4653175-4653175 on Build GRCh38
rs779623409 Chr.11: 4653274-4653274 on Build GRCh38
rs147018817 Chr.11: 4653106-4653106 on Build GRCh38
rs754916734 Chr.11: 4653266-4653266 on Build GRCh38
rs147163809 Chr.11: 4653150-4653150 on Build GRCh38
rs150540678 Chr.11: 4653107-4653107 on Build GRCh38

View Full Product Details