Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 171787586-171787859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A12
Entrez Gene ID
8604
Gene Name
solute carrier family 25 member 12
Gene Aliases
AGC1, ARALAR
UniGene
Hs.470608
Gene Chromosome Location
Chr.2: 171783405-171894306 on Build GRCh38
Transcripts
NM_003705.4 NR_047549.1 XM_011512070.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4305933 c.1756C>T p.R586* Chr.2: 171787650-171787650 on Build GRCh38
2893355 c.1705C>T p.R569W Chr.2: 171787828-171787828 on Build GRCh38
7210388 c.1744+1G>A p.? Chr.2: 171787788-171787788 on Build GRCh38
158729 c.1807C>T p.R603W Chr.2: 171787599-171787599 on Build GRCh38
4886208 c.1708G>T p.E570* Chr.2: 171787825-171787825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901376417 Chr.2: 171787835-171787835 on Build GRCh38
rs765417421 Chr.2: 171787665-171787665 on Build GRCh38
rs121434396 Chr.2: 171787637-171787637 on Build GRCh38
rs111527441 Chr.2: 171787743-171787743 on Build GRCh38
rs188048899 Chr.2: 171787796-171787796 on Build GRCh38
rs1059299 Chr.2: 171787608-171787608 on Build GRCh38
rs992670150 Chr.2: 171787712-171787712 on Build GRCh38
rs530436359 Chr.2: 171787817-171787817 on Build GRCh38
rs762212750 Chr.2: 171787692-171787693 on Build GRCh38
rs985659005 Chr.2: 171787721-171787721 on Build GRCh38
rs748887957 Chr.2: 171787795-171787795 on Build GRCh38
rs377004451 Chr.2: 171787784-171787784 on Build GRCh38
rs752119040 Chr.2: 171787701-171787701 on Build GRCh38
rs911319543 Chr.2: 171787816-171787816 on Build GRCh38
rs370060615 Chr.2: 171787778-171787778 on Build GRCh38
rs550978880 Chr.2: 171787756-171787756 on Build GRCh38
rs760496959 Chr.2: 171787652-171787652 on Build GRCh38
rs989416766 Chr.2: 171787812-171787812 on Build GRCh38
rs142891505 Chr.2: 171787612-171787612 on Build GRCh38
rs770199605 Chr.2: 171787705-171787705 on Build GRCh38
rs967612331 Chr.2: 171787730-171787730 on Build GRCh38
rs370973358 Chr.2: 171787650-171787650 on Build GRCh38
rs781717632 Chr.2: 171787744-171787744 on Build GRCh38
rs773781970 Chr.2: 171787827-171787827 on Build GRCh38
rs925847204 Chr.2: 171787750-171787750 on Build GRCh38
rs756465676 Chr.2: 171787781-171787781 on Build GRCh38
rs540738709 Chr.2: 171787687-171787687 on Build GRCh38
rs750636311 Chr.2: 171787675-171787675 on Build GRCh38
rs758575289 Chr.2: 171787678-171787678 on Build GRCh38
rs2292813 Chr.2: 171787719-171787719 on Build GRCh38

View Full Product Details