Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48592559-48592792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SULT2B1
Entrez Gene ID
6820
Gene Name
sulfotransferase family 2B member 1
Gene Aliases
HSST2
UniGene
Hs.369331
Gene Chromosome Location
Chr.19: 48552172-48599427 on Build GRCh38
Transcripts
NM_004605.2 NM_177973.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4730856 c.547C>T p.R183W Chr.19: 48592763-48592763 on Build GRCh38
1267021 c.547C>A p.R183R Chr.19: 48592763-48592763 on Build GRCh38
1267020 c.592C>A p.R198R Chr.19: 48592763-48592763 on Build GRCh38
4730855 c.592C>T p.R198W Chr.19: 48592763-48592763 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767344059 Chr.19: 48592744-48592744 on Build GRCh38
rs746137350 Chr.19: 48592698-48592698 on Build GRCh38
rs768407062 Chr.19: 48592718-48592718 on Build GRCh38
rs888269714 Chr.19: 48592634-48592634 on Build GRCh38
rs16982158 Chr.19: 48592742-48592742 on Build GRCh38
rs775812354 Chr.19: 48592702-48592702 on Build GRCh38
rs748694776 Chr.19: 48592672-48592672 on Build GRCh38
rs754737710 Chr.19: 48592679-48592679 on Build GRCh38
rs1006690221 Chr.19: 48592635-48592635 on Build GRCh38
rs769004684 Chr.19: 48592714-48592714 on Build GRCh38
rs1036294214 Chr.19: 48592597-48592597 on Build GRCh38
rs550646122 Chr.19: 48592619-48592619 on Build GRCh38
rs116135928 Chr.19: 48592682-48592682 on Build GRCh38
rs751343723 Chr.19: 48592678-48592678 on Build GRCh38
rs75015301 Chr.19: 48592651-48592651 on Build GRCh38
rs772731769 Chr.19: 48592732-48592732 on Build GRCh38
rs764776470 Chr.19: 48592726-48592726 on Build GRCh38
rs2302949 Chr.19: 48592614-48592614 on Build GRCh38
rs16982154 Chr.19: 48592741-48592741 on Build GRCh38
rs185763876 Chr.19: 48592668-48592668 on Build GRCh38
rs576779662 Chr.19: 48592755-48592755 on Build GRCh38
rs376235752 Chr.19: 48592764-48592764 on Build GRCh38
rs956380397 Chr.19: 48592696-48592696 on Build GRCh38
rs115108651 Chr.19: 48592701-48592701 on Build GRCh38
rs980940819 Chr.19: 48592716-48592716 on Build GRCh38
rs973267714 Chr.19: 48592659-48592659 on Build GRCh38
rs963114977 Chr.19: 48592654-48592654 on Build GRCh38
rs762376892 Chr.19: 48592720-48592720 on Build GRCh38
rs752620448 Chr.19: 48592748-48592748 on Build GRCh38
rs939618363 Chr.19: 48592586-48592586 on Build GRCh38
rs750141634 Chr.19: 48592673-48592673 on Build GRCh38
rs779108192 Chr.19: 48592691-48592691 on Build GRCh38
rs113644091 Chr.19: 48592645-48592645 on Build GRCh38
rs10403171 Chr.19: 48592641-48592641 on Build GRCh38
rs372325963 Chr.19: 48592683-48592683 on Build GRCh38
rs757596067 Chr.19: 48592606-48592606 on Build GRCh38
rs749794972 Chr.19: 48592603-48592603 on Build GRCh38
rs969549203 Chr.19: 48592705-48592705 on Build GRCh38
rs544329093 Chr.19: 48592763-48592763 on Build GRCh38
rs758023696 Chr.19: 48592676-48592676 on Build GRCh38
rs762411820 Chr.19: 48592737-48592737 on Build GRCh38
rs747337017 Chr.19: 48592710-48592710 on Build GRCh38
rs571218802 Chr.19: 48592610-48592610 on Build GRCh38

View Full Product Details