Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 71133004-71133233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WBSCR17
Entrez Gene ID
64409
Gene Name
Williams-Beuren syndrome chromosome region 17
Gene Aliases
GALNACT17, GALNT16, GALNT20, GALNTL3, GalNAc-T5L
UniGene
Hs.488591
Gene Chromosome Location
Chr.7: 71132414-71713601 on Build GRCh38
Transcripts
NM_022479.2 XM_011516467.2 XM_011516469.2 XM_017012521.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7251542 c.236A>C p.N79T Chr.7: 71133038-71133038 on Build GRCh38
3640190 c.220G>A p.V74M Chr.7: 71133022-71133022 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768442761 Chr.7: 71133122-71133122 on Build GRCh38
rs749995509 Chr.7: 71133027-71133027 on Build GRCh38
rs765820195 Chr.7: 71133091-71133091 on Build GRCh38
rs376660432 Chr.7: 71133086-71133086 on Build GRCh38
rs1036407800 Chr.7: 71133079-71133079 on Build GRCh38
rs969728538 Chr.7: 71133190-71133190 on Build GRCh38
rs746942359 Chr.7: 71133050-71133050 on Build GRCh38
rs757814002 Chr.7: 71133045-71133045 on Build GRCh38
rs773277842 Chr.7: 71133065-71133065 on Build GRCh38
rs774173914 Chr.7: 71133137-71133137 on Build GRCh38
rs1051538888 Chr.7: 71133140-71133140 on Build GRCh38
rs868364175 Chr.7: 71133186-71133186 on Build GRCh38
rs781201301 Chr.7: 71133058-71133058 on Build GRCh38
rs747944498 Chr.7: 71133060-71133060 on Build GRCh38
rs867090201 Chr.7: 71133123-71133123 on Build GRCh38
rs768003758 Chr.7: 71133077-71133077 on Build GRCh38
rs6976185 Chr.7: 71133146-71133146 on Build GRCh38
rs531018305 Chr.7: 71133127-71133127 on Build GRCh38
rs950515307 Chr.7: 71133173-71133173 on Build GRCh38
rs979962280 Chr.7: 71133099-71133099 on Build GRCh38
rs898689055 Chr.7: 71133152-71133152 on Build GRCh38
rs779575517 Chr.7: 71133049-71133049 on Build GRCh38
rs758868885 Chr.7: 71133144-71133144 on Build GRCh38
rs142677737 Chr.7: 71133056-71133056 on Build GRCh38
rs532058890 Chr.7: 71133169-71133169 on Build GRCh38
rs925809289 Chr.7: 71133114-71133114 on Build GRCh38
rs2903458 Chr.7: 71133073-71133073 on Build GRCh38
rs12673981 Chr.7: 71133118-71133118 on Build GRCh38
rs6976062 Chr.7: 71133085-71133085 on Build GRCh38
rs962842757 Chr.7: 71133048-71133048 on Build GRCh38
rs971714817 Chr.7: 71133202-71133202 on Build GRCh38
rs571293672 Chr.7: 71133159-71133159 on Build GRCh38
rs745434302 Chr.7: 71133081-71133081 on Build GRCh38
rs764852834 Chr.7: 71133021-71133021 on Build GRCh38
rs770191540 Chr.7: 71133062-71133062 on Build GRCh38
rs994321611 Chr.7: 71133161-71133161 on Build GRCh38
rs547015333 Chr.7: 71133184-71133184 on Build GRCh38
rs761001426 Chr.7: 71133083-71133083 on Build GRCh38
rs770940750 Chr.7: 71133072-71133072 on Build GRCh38
rs760044625 Chr.7: 71133075-71133075 on Build GRCh38
rs376053798 Chr.7: 71133068-71133068 on Build GRCh38
rs1053160700 Chr.7: 71133120-71133120 on Build GRCh38
rs1038608728 Chr.7: 71133147-71133147 on Build GRCh38
rs773420140 Chr.7: 71133183-71133183 on Build GRCh38

View Full Product Details