Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18010359-18010608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARRDC2
Entrez Gene ID
27106
Gene Name
arrestin domain containing 2
Gene Aliases
CLONE24945, PP2703
UniGene
Hs.515249
Gene Chromosome Location
Chr.19: 18001132-18014102 on Build GRCh38
Transcripts
NM_001025604.2 NM_001286826.1 NM_015683.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5964766 c.1027G>A p.D343N Chr.19: 18010601-18010601 on Build GRCh38
4075552 c.1020T>C p.P340P Chr.19: 18010579-18010579 on Build GRCh38
5520933 c.1048G>A p.E350K Chr.19: 18010607-18010607 on Build GRCh38
4075553 c.1005T>C p.P335P Chr.19: 18010579-18010579 on Build GRCh38
5964765 c.1042G>A p.D348N Chr.19: 18010601-18010601 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374287207 Chr.19: 18010393-18010393 on Build GRCh38
rs778237925 Chr.19: 18010527-18010527 on Build GRCh38
rs137999515 Chr.19: 18010468-18010468 on Build GRCh38
rs765115589 Chr.19: 18010443-18010443 on Build GRCh38
rs370881797 Chr.19: 18010543-18010543 on Build GRCh38
rs758877058 Chr.19: 18010523-18010523 on Build GRCh38
rs766203248 Chr.19: 18010389-18010389 on Build GRCh38
rs760251889 Chr.19: 18010388-18010388 on Build GRCh38
rs773279633 Chr.19: 18010572-18010572 on Build GRCh38
rs370553992 Chr.19: 18010588-18010588 on Build GRCh38
rs1006436071 Chr.19: 18010457-18010457 on Build GRCh38
rs113375710 Chr.19: 18010436-18010436 on Build GRCh38
rs772900553 Chr.19: 18010380-18010380 on Build GRCh38
rs566553001 Chr.19: 18010446-18010446 on Build GRCh38
rs967976671 Chr.19: 18010493-18010493 on Build GRCh38
rs374302335 Chr.19: 18010554-18010554 on Build GRCh38
rs546693309 Chr.19: 18010417-18010417 on Build GRCh38
rs376641323 Chr.19: 18010503-18010503 on Build GRCh38
rs753074527 Chr.19: 18010403-18010403 on Build GRCh38
rs755408714 Chr.19: 18010452-18010452 on Build GRCh38
rs768104130 Chr.19: 18010530-18010531 on Build GRCh38
rs977772145 Chr.19: 18010558-18010558 on Build GRCh38
rs560844846 Chr.19: 18010434-18010434 on Build GRCh38
rs1006823731 Chr.19: 18010437-18010437 on Build GRCh38
rs1048063935 Chr.19: 18010445-18010445 on Build GRCh38
rs781402327 Chr.19: 18010547-18010547 on Build GRCh38
rs367815858 Chr.19: 18010427-18010427 on Build GRCh38
rs1013335375 Chr.19: 18010491-18010491 on Build GRCh38
rs565228885 Chr.19: 18010551-18010551 on Build GRCh38
rs146969014 Chr.19: 18010429-18010429 on Build GRCh38
rs796944145 Chr.19: 18010482-18010482 on Build GRCh38
rs772108613 Chr.19: 18010377-18010377 on Build GRCh38
rs937577810 Chr.19: 18010510-18010510 on Build GRCh38
rs377112087 Chr.19: 18010563-18010563 on Build GRCh38
rs772196595 Chr.19: 18010564-18010564 on Build GRCh38
rs759959293 Chr.19: 18010400-18010400 on Build GRCh38
rs928846090 Chr.19: 18010506-18010506 on Build GRCh38
rs778616471 Chr.19: 18010559-18010559 on Build GRCh38
rs766469220 Chr.19: 18010418-18010418 on Build GRCh38
rs765470338 Chr.19: 18010402-18010402 on Build GRCh38
rs746523914 Chr.19: 18010587-18010587 on Build GRCh38

View Full Product Details