Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 52066517-52066762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF841
Entrez Gene ID
284371
Gene Name
zinc finger protein 841
Gene Aliases
-
UniGene
Hs.665751
Gene Chromosome Location
Chr.19: 52050028-52095780 on Build GRCh38
Transcripts
NM_001136499.1 NM_001321349.1 XM_011526833.2 XM_017026652.1 XM_017026653.1 XM_017026654.1 XM_017026655.1 XM_017026656.1 XM_017026657.1 XM_017026658.1 XM_017026659.1 XM_017026660.1 XM_017026661.1 XM_017026662.1 XM_017026663.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1590249 c.1278G>T p.K426N Chr.19: 52066604-52066604 on Build GRCh38
6341987 c.890A>G p.N297S Chr.19: 52066644-52066644 on Build GRCh38
5390552 c.1351C>T p.H451Y Chr.19: 52066531-52066531 on Build GRCh38
1590247 c.1270G>A p.A424T Chr.19: 52066612-52066612 on Build GRCh38
4459536 c.1129C>T p.R377* Chr.19: 52066753-52066753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771770702 Chr.19: 52066593-52066593 on Build GRCh38
rs866269325 Chr.19: 52066689-52066689 on Build GRCh38
rs772108090 Chr.19: 52066690-52066690 on Build GRCh38
rs753510497 Chr.19: 52066614-52066614 on Build GRCh38
rs755444222 Chr.19: 52066643-52066643 on Build GRCh38
rs761499237 Chr.19: 52066732-52066732 on Build GRCh38
rs1033296456 Chr.19: 52066674-52066674 on Build GRCh38
rs761440475 Chr.19: 52066616-52066616 on Build GRCh38
rs778008057 Chr.19: 52066651-52066651 on Build GRCh38
rs533288845 Chr.19: 52066606-52066606 on Build GRCh38
rs749467075 Chr.19: 52066669-52066669 on Build GRCh38
rs775425028 Chr.19: 52066595-52066595 on Build GRCh38
rs773440212 Chr.19: 52066560-52066560 on Build GRCh38
rs767942886 Chr.19: 52066543-52066544 on Build GRCh38
rs755277266 Chr.19: 52066548-52066548 on Build GRCh38
rs764754573 Chr.19: 52066620-52066620 on Build GRCh38
rs768531429 Chr.19: 52066713-52066713 on Build GRCh38
rs375970898 Chr.19: 52066543-52066543 on Build GRCh38
rs752019423 Chr.19: 52066640-52066640 on Build GRCh38
rs541053089 Chr.19: 52066566-52066566 on Build GRCh38
rs970795240 Chr.19: 52066646-52066646 on Build GRCh38
rs528358581 Chr.19: 52066557-52066557 on Build GRCh38
rs866597729 Chr.19: 52066723-52066723 on Build GRCh38
rs551467388 Chr.19: 52066647-52066647 on Build GRCh38
rs778988651 Chr.19: 52066680-52066680 on Build GRCh38
rs139155604 Chr.19: 52066571-52066571 on Build GRCh38
rs775268502 Chr.19: 52066697-52066697 on Build GRCh38
rs756444324 Chr.19: 52066648-52066648 on Build GRCh38
rs1054418419 Chr.19: 52066719-52066719 on Build GRCh38
rs747591636 Chr.19: 52066544-52066544 on Build GRCh38
rs774130131 Chr.19: 52066577-52066577 on Build GRCh38
rs150787624 Chr.19: 52066555-52066555 on Build GRCh38
rs745933415 Chr.19: 52066681-52066681 on Build GRCh38
rs371538802 Chr.19: 52066672-52066672 on Build GRCh38
rs776445579 Chr.19: 52066714-52066714 on Build GRCh38
rs760535914 Chr.19: 52066596-52066596 on Build GRCh38
rs759463219 Chr.19: 52066587-52066587 on Build GRCh38
rs781664063 Chr.19: 52066549-52066549 on Build GRCh38
rs781543951 Chr.19: 52066645-52066645 on Build GRCh38
rs1008251151 Chr.19: 52066615-52066615 on Build GRCh38
rs756046595 Chr.19: 52066564-52066564 on Build GRCh38
rs972060525 Chr.19: 52066623-52066623 on Build GRCh38
rs34626360 Chr.19: 52066607-52066608 on Build GRCh38

View Full Product Details