Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 41490726-41490953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA7
Entrez Gene ID
51125
Gene Name
golgin A7
Gene Aliases
GCP16, GOLGA3AP1, GOLGA7A, HSPC041
UniGene
Hs.654773
Gene Chromosome Location
Chr.8: 41490562-41510980 on Build GRCh38
Transcripts
NM_001002296.1 NM_001174124.1 NM_016099.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6396147 c.44A>G p.Q15R Chr.8: 41490898-41490898 on Build GRCh38
1728332 c.88T>C p.F30L Chr.8: 41490942-41490942 on Build GRCh38
7196234 c.50A>G p.D17G Chr.8: 41490904-41490904 on Build GRCh38
5816706 c.68G>T p.R23L Chr.8: 41490922-41490922 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs112499838 Chr.8: 41490820-41490820 on Build GRCh38
rs368262378 Chr.8: 41490821-41490821 on Build GRCh38
rs919580924 Chr.8: 41490783-41490783 on Build GRCh38
rs777664990 Chr.8: 41490872-41490872 on Build GRCh38
rs756265063 Chr.8: 41490851-41490851 on Build GRCh38
rs550090303 Chr.8: 41490782-41490782 on Build GRCh38
rs945718386 Chr.8: 41490794-41490794 on Build GRCh38
rs561594895 Chr.8: 41490749-41490749 on Build GRCh38
rs745848037 Chr.8: 41490874-41490874 on Build GRCh38
rs780405350 Chr.8: 41490829-41490829 on Build GRCh38
rs772063962 Chr.8: 41490881-41490881 on Build GRCh38
rs530314556 Chr.8: 41490757-41490757 on Build GRCh38
rs751864390 Chr.8: 41490835-41490835 on Build GRCh38
rs113202976 Chr.8: 41490808-41490808 on Build GRCh38
rs777029579 Chr.8: 41490825-41490825 on Build GRCh38
rs768563990 Chr.8: 41490893-41490893 on Build GRCh38
rs756595302 Chr.8: 41490854-41490854 on Build GRCh38
rs1051862434 Chr.8: 41490878-41490878 on Build GRCh38
rs552206338 Chr.8: 41490801-41490801 on Build GRCh38
rs139633998 Chr.8: 41490922-41490922 on Build GRCh38
rs1042981607 Chr.8: 41490889-41490889 on Build GRCh38
rs974121586 Chr.8: 41490779-41490779 on Build GRCh38
rs200358828 Chr.8: 41490886-41490886 on Build GRCh38
rs781461562 Chr.8: 41490845-41490845 on Build GRCh38
rs761449555 Chr.8: 41490818-41490818 on Build GRCh38
rs775434447 Chr.8: 41490882-41490882 on Build GRCh38
rs372140953 Chr.8: 41490885-41490885 on Build GRCh38
rs758795317 Chr.8: 41490827-41490827 on Build GRCh38
rs3780024 Chr.8: 41490799-41490799 on Build GRCh38
rs534980888 Chr.8: 41490823-41490823 on Build GRCh38
rs755324077 Chr.8: 41490844-41490844 on Build GRCh38
rs112876183 Chr.8: 41490787-41490787 on Build GRCh38
rs961661909 Chr.8: 41490763-41490763 on Build GRCh38
rs748285531 Chr.8: 41490848-41490848 on Build GRCh38

View Full Product Details