Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 51744677-51744930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSPT2
Entrez Gene ID
23708
Gene Name
G1 to S phase transition 2
Gene Aliases
ERF3B, GST2
UniGene
Hs.59523
Gene Chromosome Location
Chr.X: 51743385-51746232 on Build GRCh38
Transcripts
NM_018094.4

SNP Detail

SNP ID Chromosome Location
rs797044501 Chr.X: 51744784-51744784 on Build GRCh38
rs782551231 Chr.X: 51744834-51744834 on Build GRCh38
rs782790209 Chr.X: 51744836-51744836 on Build GRCh38
rs782713927 Chr.X: 51744707-51744707 on Build GRCh38
rs782492592 Chr.X: 51744857-51744857 on Build GRCh38
rs202017797 Chr.X: 51744871-51744871 on Build GRCh38
rs140733143 Chr.X: 51744900-51744900 on Build GRCh38
rs782287535 Chr.X: 51744906-51744906 on Build GRCh38
rs782454145 Chr.X: 51744890-51744890 on Build GRCh38
rs376744022 Chr.X: 51744808-51744808 on Build GRCh38
rs929569177 Chr.X: 51744859-51744859 on Build GRCh38
rs369786409 Chr.X: 51744862-51744862 on Build GRCh38

View Full Product Details