Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12800691-12800964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRDX2
Entrez Gene ID
7001
Gene Name
peroxiredoxin 2
Gene Aliases
HEL-S-2a, NKEF-B, NKEFB, PRP, PRX2, PRXII, PTX1, TDPX1, TPX1, TSA
UniGene
Hs.432121
Gene Chromosome Location
Chr.19: 12796820-12801910 on Build GRCh38
Transcripts
NM_005809.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5537574 c.229G>A p.V77M Chr.19: 12800944-12800944 on Build GRCh38
3796655 c.227C>T p.S76L Chr.19: 12800946-12800946 on Build GRCh38
4453735 c.340A>C p.N114H Chr.19: 12800833-12800833 on Build GRCh38
1720122 c.301C>T p.P101S Chr.19: 12800872-12800872 on Build GRCh38
4385046 c.253G>A p.A85T Chr.19: 12800920-12800920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768812837 Chr.19: 12800800-12800800 on Build GRCh38
rs573045025 Chr.19: 12800765-12800765 on Build GRCh38
rs368264855 Chr.19: 12800764-12800764 on Build GRCh38
rs780809158 Chr.19: 12800879-12800879 on Build GRCh38
rs761905421 Chr.19: 12800845-12800845 on Build GRCh38
rs376098373 Chr.19: 12800945-12800945 on Build GRCh38
rs777613891 Chr.19: 12800817-12800817 on Build GRCh38
rs916807986 Chr.19: 12800824-12800824 on Build GRCh38
rs777270227 Chr.19: 12800920-12800920 on Build GRCh38
rs372357619 Chr.19: 12800718-12800718 on Build GRCh38
rs748793958 Chr.19: 12800921-12800921 on Build GRCh38
rs909204901 Chr.19: 12800734-12800734 on Build GRCh38
rs761015596 Chr.19: 12800779-12800779 on Build GRCh38
rs949532017 Chr.19: 12800834-12800834 on Build GRCh38
rs755088051 Chr.19: 12800784-12800784 on Build GRCh38
rs34577345 Chr.19: 12800895-12800895 on Build GRCh38
rs765369515 Chr.19: 12800846-12800846 on Build GRCh38
rs749402992 Chr.19: 12800724-12800724 on Build GRCh38
rs868512766 Chr.19: 12800865-12800865 on Build GRCh38
rs541462105 Chr.19: 12800821-12800821 on Build GRCh38
rs754770332 Chr.19: 12800868-12800868 on Build GRCh38
rs755665687 Chr.19: 12800897-12800897 on Build GRCh38
rs766403350 Chr.19: 12800856-12800856 on Build GRCh38
rs947336612 Chr.19: 12800831-12800831 on Build GRCh38
rs777514193 Chr.19: 12800935-12800936 on Build GRCh38
rs751448343 Chr.19: 12800859-12800859 on Build GRCh38
rs763048288 Chr.19: 12800853-12800853 on Build GRCh38
rs371004936 Chr.19: 12800851-12800851 on Build GRCh38

View Full Product Details