Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103532611-103532876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMT61A
Entrez Gene ID
115708
Gene Name
tRNA methyltransferase 61A
Gene Aliases
C14orf172, GCD14, Gcd14p, TRM61, hTRM61
UniGene
Hs.525610
Gene Chromosome Location
Chr.14: 103529172-103537073 on Build GRCh38
Transcripts
NM_152307.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6304272 c.478C>T p.R160C Chr.14: 103532728-103532728 on Build GRCh38
6809059 c.384G>A p.T128T Chr.14: 103532634-103532634 on Build GRCh38
3689906 c.495C>T p.C165C Chr.14: 103532745-103532745 on Build GRCh38
953641 c.564C>A p.A188A Chr.14: 103532814-103532814 on Build GRCh38
6347426 c.464G>T p.R155L Chr.14: 103532714-103532714 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747508008 Chr.14: 103532713-103532713 on Build GRCh38
rs370257424 Chr.14: 103532832-103532832 on Build GRCh38
rs559412988 Chr.14: 103532761-103532761 on Build GRCh38
rs763916586 Chr.14: 103532739-103532739 on Build GRCh38
rs772491014 Chr.14: 103532633-103532633 on Build GRCh38
rs544887297 Chr.14: 103532778-103532778 on Build GRCh38
rs764882612 Chr.14: 103532674-103532674 on Build GRCh38
rs758770178 Chr.14: 103532705-103532705 on Build GRCh38
rs542902305 Chr.14: 103532856-103532856 on Build GRCh38
rs563404267 Chr.14: 103532844-103532844 on Build GRCh38
rs373210459 Chr.14: 103532762-103532762 on Build GRCh38
rs537191569 Chr.14: 103532680-103532680 on Build GRCh38
rs997419700 Chr.14: 103532669-103532669 on Build GRCh38
rs767496789 Chr.14: 103532747-103532747 on Build GRCh38
rs765359264 Chr.14: 103532833-103532833 on Build GRCh38
rs371544924 Chr.14: 103532640-103532640 on Build GRCh38
rs373651893 Chr.14: 103532684-103532684 on Build GRCh38
rs557025159 Chr.14: 103532857-103532857 on Build GRCh38
rs1057390847 Chr.14: 103532811-103532811 on Build GRCh38
rs569343687 Chr.14: 103532845-103532845 on Build GRCh38
rs748964962 Chr.14: 103532795-103532795 on Build GRCh38
rs746076593 Chr.14: 103532826-103532826 on Build GRCh38
rs749946605 Chr.14: 103532724-103532725 on Build GRCh38
rs778183539 Chr.14: 103532776-103532776 on Build GRCh38
rs978954240 Chr.14: 103532766-103532766 on Build GRCh38
rs376660789 Chr.14: 103532704-103532704 on Build GRCh38
rs762704130 Chr.14: 103532735-103532735 on Build GRCh38
rs373346624 Chr.14: 103532782-103532782 on Build GRCh38
rs756301434 Chr.14: 103532688-103532688 on Build GRCh38
rs779610934 Chr.14: 103532787-103532787 on Build GRCh38
rs772161793 Chr.14: 103532824-103532824 on Build GRCh38
rs769325334 Chr.14: 103532783-103532783 on Build GRCh38
rs369292682 Chr.14: 103532668-103532668 on Build GRCh38
rs761662772 Chr.14: 103532818-103532818 on Build GRCh38
rs757761969 Chr.14: 103532779-103532779 on Build GRCh38
rs376240553 Chr.14: 103532649-103532649 on Build GRCh38
rs573048606 Chr.14: 103532770-103532770 on Build GRCh38
rs1043196830 Chr.14: 103532631-103532631 on Build GRCh38
rs931727832 Chr.14: 103532707-103532707 on Build GRCh38
rs761857212 Chr.14: 103532744-103532744 on Build GRCh38
rs763960359 Chr.14: 103532683-103532683 on Build GRCh38
rs1014858135 Chr.14: 103532850-103532850 on Build GRCh38
rs1001612874 Chr.14: 103532803-103532803 on Build GRCh38
rs781632633 Chr.14: 103532781-103532781 on Build GRCh38
rs753912375 Chr.14: 103532693-103532693 on Build GRCh38
rs901137937 Chr.14: 103532663-103532663 on Build GRCh38
rs745307285 Chr.14: 103532725-103532725 on Build GRCh38
rs558288621 Chr.14: 103532634-103532634 on Build GRCh38
rs764801368 Chr.14: 103532669-103532669 on Build GRCh38
rs774489533 Chr.14: 103532740-103532740 on Build GRCh38
rs769362508 Chr.14: 103532728-103532728 on Build GRCh38
rs201889259 Chr.14: 103532814-103532814 on Build GRCh38
rs980236035 Chr.14: 103532733-103532733 on Build GRCh38
rs886314548 Chr.14: 103532682-103532682 on Build GRCh38
rs200065262 Chr.14: 103532696-103532696 on Build GRCh38
rs542612707 Chr.14: 103532714-103532714 on Build GRCh38
rs905664657 Chr.14: 103532849-103532849 on Build GRCh38
rs754003512 Chr.14: 103532760-103532760 on Build GRCh38
rs200737456 Chr.14: 103532646-103532646 on Build GRCh38
rs369894031 Chr.14: 103532775-103532775 on Build GRCh38
rs947583538 Chr.14: 103532765-103532765 on Build GRCh38
rs530594871 Chr.14: 103532823-103532823 on Build GRCh38
rs778902763 Chr.14: 103532648-103532648 on Build GRCh38
rs376392701 Chr.14: 103532815-103532815 on Build GRCh38
rs1047674008 Chr.14: 103532825-103532825 on Build GRCh38
rs557071436 Chr.14: 103532692-103532692 on Build GRCh38
rs775159038 Chr.14: 103532732-103532732 on Build GRCh38

View Full Product Details