Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184354006-184354267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCN2
Entrez Gene ID
1181
Gene Name
chloride voltage-gated channel 2
Gene Aliases
CIC-2, CLC2, ECA2, ECA3, EGI11, EGI3, EGMA, EJM6, EJM8, LKPAT, clC-2
UniGene
Hs.436847
Gene Chromosome Location
Chr.3: 184346185-184361651 on Build GRCh38
Transcripts
NM_001171087.2 NM_001171088.2 NM_001171089.2 NM_004366.5 XM_006713489.1 XM_006713490.2 XM_011512401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4672622 c.1715G>A p.R572H Chr.3: 184354107-184354107 on Build GRCh38
4115772 c.1586C>T p.A529V Chr.3: 184354236-184354236 on Build GRCh38
4886268 c.1692C>A p.Y564* Chr.3: 184354130-184354130 on Build GRCh38
4149703 c.1672C>T p.R558* Chr.3: 184354150-184354150 on Build GRCh38
3331328 c.1673G>A p.R558Q Chr.3: 184354149-184354149 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777315003 Chr.3: 184354068-184354068 on Build GRCh38
rs761825582 Chr.3: 184354217-184354217 on Build GRCh38
rs748646018 Chr.3: 184354055-184354055 on Build GRCh38
rs966004094 Chr.3: 184354061-184354061 on Build GRCh38
rs201330912 Chr.3: 184354113-184354113 on Build GRCh38
rs776493182 Chr.3: 184354214-184354214 on Build GRCh38
rs745479097 Chr.3: 184354195-184354195 on Build GRCh38
rs774433405 Chr.3: 184354226-184354226 on Build GRCh38
rs1042484029 Chr.3: 184354223-184354223 on Build GRCh38
rs746824041 Chr.3: 184354211-184354211 on Build GRCh38
rs1018940878 Chr.3: 184354030-184354030 on Build GRCh38
rs115803480 Chr.3: 184354196-184354196 on Build GRCh38
rs781539184 Chr.3: 184354054-184354054 on Build GRCh38
rs377007160 Chr.3: 184354141-184354141 on Build GRCh38
rs374265846 Chr.3: 184354097-184354097 on Build GRCh38
rs572504135 Chr.3: 184354151-184354151 on Build GRCh38
rs756767463 Chr.3: 184354190-184354190 on Build GRCh38
rs756597060 Chr.3: 184354059-184354059 on Build GRCh38
rs756612780 Chr.3: 184354169-184354169 on Build GRCh38
rs199539697 Chr.3: 184354149-184354149 on Build GRCh38
rs200337116 Chr.3: 184354222-184354222 on Build GRCh38
rs774202980 Chr.3: 184354084-184354084 on Build GRCh38
rs201085080 Chr.3: 184354077-184354077 on Build GRCh38
rs200332081 Chr.3: 184354096-184354096 on Build GRCh38
rs538550244 Chr.3: 184354076-184354076 on Build GRCh38
rs1016560160 Chr.3: 184354041-184354041 on Build GRCh38
rs137978189 Chr.3: 184354189-184354189 on Build GRCh38
rs796175896 Chr.3: 184354025-184354025 on Build GRCh38
rs745531587 Chr.3: 184354095-184354095 on Build GRCh38
rs779876583 Chr.3: 184354198-184354198 on Build GRCh38
rs927645302 Chr.3: 184354228-184354228 on Build GRCh38
rs753047668 Chr.3: 184354160-184354160 on Build GRCh38
rs150362392 Chr.3: 184354150-184354150 on Build GRCh38
rs768638424 Chr.3: 184354213-184354213 on Build GRCh38
rs770702381 Chr.3: 184354078-184354078 on Build GRCh38
rs748454023 Chr.3: 184354105-184354107 on Build GRCh38
rs745559622 Chr.3: 184354173-184354173 on Build GRCh38
rs372735937 Chr.3: 184354118-184354118 on Build GRCh38
rs760613157 Chr.3: 184354108-184354108 on Build GRCh38
rs199889662 Chr.3: 184354117-184354117 on Build GRCh38
rs766576851 Chr.3: 184354124-184354124 on Build GRCh38
rs34658600 Chr.3: 184354046-184354046 on Build GRCh38

View Full Product Details