Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 3755434-3755707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC27
Entrez Gene ID
148870
Gene Name
coiled-coil domain containing 27
Gene Aliases
-
UniGene
Hs.348700
Gene Chromosome Location
Chr.1: 3752398-3771645 on Build GRCh38
Transcripts
NM_152492.2 XM_011540780.1 XM_017000402.1 XM_017000403.1 XM_017000404.1 XM_017000405.1 XM_017000406.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6880486 c.472G>A p.E158K Chr.1: 3755486-3755486 on Build GRCh38
319279 c.486C>A p.S162R Chr.1: 3755500-3755500 on Build GRCh38
5342380 c.489G>A p.S163S Chr.1: 3755503-3755503 on Build GRCh38
4669886 c.505A>G p.T169A Chr.1: 3755519-3755519 on Build GRCh38
2081186 c.511G>T p.D171Y Chr.1: 3755525-3755525 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983895723 Chr.1: 3755647-3755647 on Build GRCh38
rs761428776 Chr.1: 3755483-3755483 on Build GRCh38
rs963449751 Chr.1: 3755520-3755520 on Build GRCh38
rs138353020 Chr.1: 3755535-3755535 on Build GRCh38
rs781522400 Chr.1: 3755462-3755462 on Build GRCh38
rs769266294 Chr.1: 3755482-3755482 on Build GRCh38
rs200546551 Chr.1: 3755561-3755561 on Build GRCh38
rs757376567 Chr.1: 3755511-3755511 on Build GRCh38
rs771615605 Chr.1: 3755552-3755552 on Build GRCh38
rs756184696 Chr.1: 3755585-3755585 on Build GRCh38
rs571918762 Chr.1: 3755557-3755557 on Build GRCh38
rs199671732 Chr.1: 3755577-3755577 on Build GRCh38
rs139820400 Chr.1: 3755468-3755468 on Build GRCh38
rs752552695 Chr.1: 3755510-3755510 on Build GRCh38
rs754683379 Chr.1: 3755686-3755686 on Build GRCh38
rs753168734 Chr.1: 3755474-3755474 on Build GRCh38
rs772763352 Chr.1: 3755614-3755614 on Build GRCh38
rs779108779 Chr.1: 3755607-3755607 on Build GRCh38
rs755392937 Chr.1: 3755596-3755596 on Build GRCh38
rs367634151 Chr.1: 3755505-3755505 on Build GRCh38
rs756274642 Chr.1: 3755521-3755521 on Build GRCh38
rs773021597 Chr.1: 3755487-3755487 on Build GRCh38
rs779617326 Chr.1: 3755655-3755655 on Build GRCh38
rs758206536 Chr.1: 3755567-3755567 on Build GRCh38
rs545859943 Chr.1: 3755668-3755668 on Build GRCh38
rs760335993 Chr.1: 3755490-3755490 on Build GRCh38
rs562932128 Chr.1: 3755682-3755682 on Build GRCh38
rs372310361 Chr.1: 3755609-3755609 on Build GRCh38
rs149439180 Chr.1: 3755560-3755560 on Build GRCh38
rs201434850 Chr.1: 3755544-3755544 on Build GRCh38
rs766290829 Chr.1: 3755497-3755497 on Build GRCh38
rs902714278 Chr.1: 3755533-3755533 on Build GRCh38
rs866088672 Chr.1: 3755504-3755504 on Build GRCh38
rs149911658 Chr.1: 3755463-3755463 on Build GRCh38
rs936397108 Chr.1: 3755640-3755640 on Build GRCh38
rs377290099 Chr.1: 3755669-3755669 on Build GRCh38
rs146403079 Chr.1: 3755484-3755484 on Build GRCh38
rs267598583 Chr.1: 3755512-3755512 on Build GRCh38
rs538197172 Chr.1: 3755672-3755672 on Build GRCh38
rs775768173 Chr.1: 3755473-3755473 on Build GRCh38
rs768872260 Chr.1: 3755537-3755537 on Build GRCh38
rs749722445 Chr.1: 3755536-3755536 on Build GRCh38
rs752708476 Chr.1: 3755566-3755566 on Build GRCh38
rs909596366 Chr.1: 3755481-3755481 on Build GRCh38
rs139363014 Chr.1: 3755556-3755556 on Build GRCh38
rs774973075 Chr.1: 3755543-3755543 on Build GRCh38
rs892647149 Chr.1: 3755665-3755665 on Build GRCh38
rs760469532 Chr.1: 3755464-3755467 on Build GRCh38
rs143598266 Chr.1: 3755503-3755503 on Build GRCh38
rs756545169 Chr.1: 3755517-3755517 on Build GRCh38
rs779325883 Chr.1: 3755673-3755673 on Build GRCh38
rs150439655 Chr.1: 3755518-3755518 on Build GRCh38
rs199649998 Chr.1: 3755502-3755502 on Build GRCh38
rs778417322 Chr.1: 3755617-3755617 on Build GRCh38
rs746556620 Chr.1: 3755618-3755618 on Build GRCh38
rs879017005 Chr.1: 3755638-3755638 on Build GRCh38
rs4648555 Chr.1: 3755592-3755592 on Build GRCh38
rs767602641 Chr.1: 3755575-3755575 on Build GRCh38
rs746036284 Chr.1: 3755465-3755465 on Build GRCh38

View Full Product Details