Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9755057-9755314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD69
Entrez Gene ID
969
Gene Name
CD69 molecule
Gene Aliases
AIM, BL-AC/P26, CLEC2C, EA1, GP32/28, MLR-3
UniGene
Hs.208854
Gene Chromosome Location
Chr.12: 9752486-9760901 on Build GRCh38
Transcripts
NM_001781.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2047243 c.251C>T p.S84L Chr.12: 9755198-9755198 on Build GRCh38
4045600 c.249T>G p.S83S Chr.12: 9755200-9755200 on Build GRCh38
944900 c.387+2T>C p.? Chr.12: 9755060-9755060 on Build GRCh38
6138713 c.332A>T p.N111I Chr.12: 9755117-9755117 on Build GRCh38
4045598 c.290A>G p.Y97C Chr.12: 9755159-9755159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200018429 Chr.12: 9755272-9755272 on Build GRCh38
rs758461109 Chr.12: 9755151-9755151 on Build GRCh38
rs996490771 Chr.12: 9755223-9755223 on Build GRCh38
rs759377891 Chr.12: 9755134-9755134 on Build GRCh38
rs150634267 Chr.12: 9755102-9755102 on Build GRCh38
rs149340487 Chr.12: 9755239-9755239 on Build GRCh38
rs777850345 Chr.12: 9755153-9755153 on Build GRCh38
rs370079360 Chr.12: 9755137-9755137 on Build GRCh38
rs3176798 Chr.12: 9755190-9755190 on Build GRCh38
rs773715256 Chr.12: 9755085-9755085 on Build GRCh38
rs746297540 Chr.12: 9755189-9755189 on Build GRCh38
rs780572690 Chr.12: 9755196-9755196 on Build GRCh38
rs761428857 Chr.12: 9755093-9755093 on Build GRCh38
rs771554433 Chr.12: 9755225-9755225 on Build GRCh38
rs753264576 Chr.12: 9755183-9755183 on Build GRCh38
rs766211104 Chr.12: 9755110-9755110 on Build GRCh38
rs138724350 Chr.12: 9755177-9755177 on Build GRCh38
rs772853305 Chr.12: 9755229-9755229 on Build GRCh38
rs778343041 Chr.12: 9755263-9755263 on Build GRCh38
rs770763886 Chr.12: 9755241-9755241 on Build GRCh38
rs759413895 Chr.12: 9755251-9755251 on Build GRCh38
rs201332184 Chr.12: 9755244-9755244 on Build GRCh38
rs747767453 Chr.12: 9755088-9755088 on Build GRCh38
rs753752547 Chr.12: 9755127-9755127 on Build GRCh38
rs771515061 Chr.12: 9755094-9755094 on Build GRCh38
rs762807243 Chr.12: 9755273-9755273 on Build GRCh38
rs751719214 Chr.12: 9755155-9755155 on Build GRCh38
rs764091481 Chr.12: 9755275-9755275 on Build GRCh38
rs1028945720 Chr.12: 9755269-9755269 on Build GRCh38
rs770172111 Chr.12: 9755193-9755193 on Build GRCh38
rs765267342 Chr.12: 9755136-9755136 on Build GRCh38
rs749782838 Chr.12: 9755210-9755210 on Build GRCh38
rs868331664 Chr.12: 9755116-9755116 on Build GRCh38

View Full Product Details