Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 84529069-84529318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH6
Entrez Gene ID
1768
Gene Name
dynein axonemal heavy chain 6
Gene Aliases
DNHL1, Dnahc6, HL-2, HL2
UniGene
Hs.249972
Gene Chromosome Location
Chr.2: 84459588-84819589 on Build GRCh38
Transcripts
NM_001370.1 XM_006711956.2 XM_011532649.2 XM_011532650.2 XM_011532652.1 XM_011532653.1 XM_011532654.1 XM_011532655.2 XM_011532657.1 XM_011532660.1 XM_017003521.1 XM_017003522.1 XM_017003523.1 XM_017003524.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7316802 c.612G>A p.M204I Chr.2: 84529116-84529116 on Build GRCh38
3001081 c.591A>C p.E197D Chr.2: 84529095-84529095 on Build GRCh38
6227193 c.575T>C p.I192T Chr.2: 84529079-84529079 on Build GRCh38
6227194 c.575T>C p.I192T Chr.2: 84529079-84529079 on Build GRCh38
4828757 c.589G>A p.E197K Chr.2: 84529093-84529093 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772845816 Chr.2: 84529196-84529196 on Build GRCh38
rs771607358 Chr.2: 84529164-84529164 on Build GRCh38
rs140020163 Chr.2: 84529193-84529193 on Build GRCh38
rs748641131 Chr.2: 84529153-84529153 on Build GRCh38
rs983131406 Chr.2: 84529253-84529255 on Build GRCh38
rs1012671740 Chr.2: 84529129-84529129 on Build GRCh38
rs35141655 Chr.2: 84529229-84529230 on Build GRCh38
rs1008916901 Chr.2: 84529157-84529157 on Build GRCh38
rs759341656 Chr.2: 84529158-84529162 on Build GRCh38
rs767093648 Chr.2: 84529163-84529164 on Build GRCh38
rs868818078 Chr.2: 84529169-84529169 on Build GRCh38
rs544613390 Chr.2: 84529223-84529223 on Build GRCh38
rs569300923 Chr.2: 84529176-84529176 on Build GRCh38
rs4832090 Chr.2: 84529217-84529217 on Build GRCh38
rs749623052 Chr.2: 84529112-84529112 on Build GRCh38
rs747846955 Chr.2: 84529122-84529122 on Build GRCh38
rs988784330 Chr.2: 84529221-84529221 on Build GRCh38
rs774899113 Chr.2: 84529141-84529141 on Build GRCh38
rs769299206 Chr.2: 84529155-84529155 on Build GRCh38
rs768921357 Chr.2: 84529131-84529131 on Build GRCh38

View Full Product Details