Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 45366370-45366627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATM
Entrez Gene ID
2628
Gene Name
glycine amidinotransferase
Gene Aliases
AGAT, AT, CCDS3
UniGene
Hs.560354
Gene Chromosome Location
Chr.15: 45361124-45402317 on Build GRCh38
Transcripts
NM_001321015.1 NM_001482.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
433966 c.775A>T p.I259F Chr.15: 45366409-45366409 on Build GRCh38
6669649 c.781G>T p.A261S Chr.15: 45366403-45366403 on Build GRCh38
5514681 c.729A>G p.K243K Chr.15: 45366455-45366455 on Build GRCh38
962248 c.739A>G p.T247A Chr.15: 45366445-45366445 on Build GRCh38
6580502 c.676-8C>G p.? Chr.15: 45366516-45366516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764877849 Chr.15: 45366474-45366474 on Build GRCh38
rs758138751 Chr.15: 45366484-45366484 on Build GRCh38
rs1038653266 Chr.15: 45366562-45366562 on Build GRCh38
rs778983865 Chr.15: 45366531-45366531 on Build GRCh38
rs754791975 Chr.15: 45366497-45366497 on Build GRCh38
rs764653380 Chr.15: 45366452-45366452 on Build GRCh38
rs775638076 Chr.15: 45366548-45366548 on Build GRCh38
rs748127418 Chr.15: 45366522-45366522 on Build GRCh38
rs570670146 Chr.15: 45366499-45366499 on Build GRCh38
rs768949946 Chr.15: 45366551-45366551 on Build GRCh38
rs761087555 Chr.15: 45366549-45366549 on Build GRCh38
rs779648078 Chr.15: 45366485-45366485 on Build GRCh38
rs746029716 Chr.15: 45366535-45366535 on Build GRCh38
rs202225656 Chr.15: 45366492-45366492 on Build GRCh38
rs775933965 Chr.15: 45366406-45366406 on Build GRCh38
rs761016596 Chr.15: 45366428-45366428 on Build GRCh38
rs146057680 Chr.15: 45366483-45366483 on Build GRCh38
rs761384746 Chr.15: 45366467-45366467 on Build GRCh38
rs754453345 Chr.15: 45366456-45366456 on Build GRCh38
rs772339429 Chr.15: 45366539-45366539 on Build GRCh38

View Full Product Details