Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 5453133-5453360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NET1
Entrez Gene ID
10276
Gene Name
neuroepithelial cell transforming 1
Gene Aliases
ARHGEF8, NET1A
UniGene
Hs.25155
Gene Chromosome Location
Chr.10: 5412551-5459056 on Build GRCh38
Transcripts
NM_001047160.2 NM_005863.4 NR_073040.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4014627 c.510T>C p.S170S Chr.10: 5453327-5453327 on Build GRCh38
367021 c.475G>C p.E159Q Chr.10: 5453292-5453292 on Build GRCh38
33509 c.442G>A p.D148N Chr.10: 5453259-5453259 on Build GRCh38
4706498 c.500A>G p.D167G Chr.10: 5453317-5453317 on Build GRCh38
4706497 c.662A>G p.D221G Chr.10: 5453317-5453317 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11253185 Chr.10: 5453194-5453194 on Build GRCh38
rs749433117 Chr.10: 5453279-5453279 on Build GRCh38
rs757735786 Chr.10: 5453223-5453223 on Build GRCh38
rs4567355 Chr.10: 5453255-5453255 on Build GRCh38
rs752849409 Chr.10: 5453217-5453217 on Build GRCh38
rs771925361 Chr.10: 5453329-5453329 on Build GRCh38
rs746950171 Chr.10: 5453331-5453331 on Build GRCh38
rs780188156 Chr.10: 5453276-5453276 on Build GRCh38
rs1013431228 Chr.10: 5453174-5453174 on Build GRCh38
rs759260902 Chr.10: 5453202-5453202 on Build GRCh38
rs770384396 Chr.10: 5453334-5453334 on Build GRCh38
rs377599173 Chr.10: 5453217-5453217 on Build GRCh38
rs370239800 Chr.10: 5453330-5453330 on Build GRCh38
rs768947309 Chr.10: 5453286-5453286 on Build GRCh38
rs894380059 Chr.10: 5453266-5453266 on Build GRCh38
rs935924513 Chr.10: 5453243-5453243 on Build GRCh38
rs747947249 Chr.10: 5453312-5453312 on Build GRCh38
rs373998179 Chr.10: 5453176-5453179 on Build GRCh38
rs779055914 Chr.10: 5453307-5453307 on Build GRCh38
rs373341552 Chr.10: 5453201-5453201 on Build GRCh38
rs78935709 Chr.10: 5453156-5453156 on Build GRCh38
rs1033789804 Chr.10: 5453157-5453157 on Build GRCh38
rs924273339 Chr.10: 5453228-5453228 on Build GRCh38
rs756205462 Chr.10: 5453257-5453257 on Build GRCh38
rs781763319 Chr.10: 5453236-5453236 on Build GRCh38
rs895231786 Chr.10: 5453164-5453164 on Build GRCh38
rs966669758 Chr.10: 5453224-5453224 on Build GRCh38

View Full Product Details