Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 239989975-239990232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFA10
Entrez Gene ID
4705
Gene Name
NADH:ubiquinone oxidoreductase subunit A10
Gene Aliases
CI-42KD, CI-42k
UniGene
Hs.277677
Gene Chromosome Location
Chr.2: 239892450-240025402 on Build GRCh38
Transcripts
NM_001322019.1 NM_001322020.1 NM_004544.3 NR_136155.1 NR_136156.1 NR_136157.1 NR_136158.1 XM_011511228.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
720944 c.999+1G>A p.? Chr.2: 239990073-239990073 on Build GRCh38
3579815 c.943C>T p.P315S Chr.2: 239990130-239990130 on Build GRCh38
3579816 c.940C>T p.L314F Chr.2: 239990133-239990133 on Build GRCh38
3709531 c.947A>G p.E316G Chr.2: 239990126-239990126 on Build GRCh38
6723671 c.900T>A p.D300E Chr.2: 239990173-239990173 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778359023 Chr.2: 239990070-239990070 on Build GRCh38
rs762245412 Chr.2: 239990097-239990097 on Build GRCh38
rs772226484 Chr.2: 239990169-239990169 on Build GRCh38
rs1047895963 Chr.2: 239990200-239990200 on Build GRCh38
rs777314877 Chr.2: 239990095-239990095 on Build GRCh38
rs772406797 Chr.2: 239990174-239990174 on Build GRCh38
rs750583575 Chr.2: 239990046-239990046 on Build GRCh38
rs754831388 Chr.2: 239990061-239990061 on Build GRCh38
rs1009837787 Chr.2: 239990199-239990199 on Build GRCh38
rs781782535 Chr.2: 239990077-239990077 on Build GRCh38
rs199902163 Chr.2: 239990161-239990161 on Build GRCh38
rs766358570 Chr.2: 239990198-239990198 on Build GRCh38
rs769146731 Chr.2: 239990020-239990020 on Build GRCh38
rs549261468 Chr.2: 239990007-239990007 on Build GRCh38
rs780552550 Chr.2: 239990139-239990139 on Build GRCh38
rs770237200 Chr.2: 239990120-239990120 on Build GRCh38
rs995602943 Chr.2: 239990015-239990015 on Build GRCh38
rs769063751 Chr.2: 239990151-239990151 on Build GRCh38
rs199987605 Chr.2: 239990141-239990141 on Build GRCh38
rs561645519 Chr.2: 239990029-239990029 on Build GRCh38
rs547496868 Chr.2: 239990177-239990177 on Build GRCh38
rs927525207 Chr.2: 239990067-239990067 on Build GRCh38
rs760855270 Chr.2: 239990197-239990197 on Build GRCh38
rs373080713 Chr.2: 239990051-239990051 on Build GRCh38
rs756093084 Chr.2: 239990050-239990050 on Build GRCh38
rs531918393 Chr.2: 239990034-239990034 on Build GRCh38
rs929063799 Chr.2: 239990127-239990127 on Build GRCh38
rs764167121 Chr.2: 239990025-239990025 on Build GRCh38
rs758042753 Chr.2: 239990079-239990079 on Build GRCh38
rs980415625 Chr.2: 239990059-239990059 on Build GRCh38
rs774806560 Chr.2: 239990042-239990042 on Build GRCh38
rs149015032 Chr.2: 239990153-239990153 on Build GRCh38
rs747830311 Chr.2: 239990076-239990076 on Build GRCh38

View Full Product Details