Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 121460041-121460298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLQ
Entrez Gene ID
10721
Gene Name
DNA polymerase theta
Gene Aliases
PRO0327
UniGene
Hs.241517
Gene Chromosome Location
Chr.3: 121431420-121546006 on Build GRCh38
Transcripts
NM_199420.3 XM_011512343.2 XM_011512347.2 XM_011512348.2 XM_011512354.2 XM_017005565.1

SNP Detail

SNP ID Chromosome Location
rs762653253 Chr.3: 121460096-121460096 on Build GRCh38
rs769839271 Chr.3: 121460181-121460181 on Build GRCh38
rs3218638 Chr.3: 121460265-121460265 on Build GRCh38
rs766459523 Chr.3: 121460215-121460215 on Build GRCh38
rs754004818 Chr.3: 121460216-121460216 on Build GRCh38
rs912055934 Chr.3: 121460170-121460170 on Build GRCh38
rs771303558 Chr.3: 121460176-121460176 on Build GRCh38
rs756208896 Chr.3: 121460138-121460138 on Build GRCh38
rs768392117 Chr.3: 121460194-121460194 on Build GRCh38
rs762786939 Chr.3: 121460190-121460190 on Build GRCh38
rs751274563 Chr.3: 121460266-121460266 on Build GRCh38
rs746607693 Chr.3: 121460174-121460174 on Build GRCh38
rs754305359 Chr.3: 121460160-121460160 on Build GRCh38
rs759527088 Chr.3: 121460221-121460221 on Build GRCh38
rs746425164 Chr.3: 121460152-121460152 on Build GRCh38
rs756207967 Chr.3: 121460264-121460265 on Build GRCh38
rs764957894 Chr.3: 121460071-121460071 on Build GRCh38
rs78788065 Chr.3: 121460165-121460165 on Build GRCh38
rs745359419 Chr.3: 121460180-121460180 on Build GRCh38
rs775415107 Chr.3: 121460093-121460093 on Build GRCh38
rs773454524 Chr.3: 121460199-121460199 on Build GRCh38
rs767768260 Chr.3: 121460108-121460108 on Build GRCh38
rs776938206 Chr.3: 121460178-121460178 on Build GRCh38
rs748246811 Chr.3: 121460098-121460098 on Build GRCh38
rs750623071 Chr.3: 121460131-121460131 on Build GRCh38
rs149262016 Chr.3: 121460086-121460086 on Build GRCh38
rs550897860 Chr.3: 121460091-121460091 on Build GRCh38
rs753118922 Chr.3: 121460244-121460244 on Build GRCh38
rs778220870 Chr.3: 121460260-121460260 on Build GRCh38
rs777614615 Chr.3: 121460166-121460166 on Build GRCh38
rs772616659 Chr.3: 121460246-121460246 on Build GRCh38
rs370791975 Chr.3: 121460214-121460214 on Build GRCh38
rs143352658 Chr.3: 121460162-121460162 on Build GRCh38
rs760819269 Chr.3: 121460204-121460204 on Build GRCh38
rs1000284210 Chr.3: 121460107-121460107 on Build GRCh38
rs765809823 Chr.3: 121460223-121460223 on Build GRCh38
rs775261164 Chr.3: 121460186-121460186 on Build GRCh38
rs376597667 Chr.3: 121460163-121460163 on Build GRCh38

View Full Product Details