Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 79642711-79642970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAPD4
Entrez Gene ID
167153
Gene Name
PAP associated domain containing 4
Gene Aliases
GLD2, TUT2
UniGene
Hs.418198
Gene Chromosome Location
Chr.5: 79612416-79686648 on Build GRCh38
Transcripts
NM_001114393.1 NM_001114394.1 NM_001297744.1 NM_001297745.1 NM_173797.3 XM_011543219.2 XM_011543220.2 XM_011543221.2 XM_011543222.2 XM_011543223.2 XM_011543224.2 XM_011543225.2 XM_011543226.2 XM_011543227.2 XM_011543228.2 XM_011543229.2 XM_011543230.2 XM_011543231.2 XM_011543232.2 XM_011543233.2 XM_017009151.1 XM_017009152.1 XM_017009153.1 XM_017009154.1 XM_017009155.1 XM_017009156.1 XM_017009157.1 XM_017009158.1 XM_017009159.1 XM_017009160.1 XM_017009161.1 XM_017009162.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6220900 c.675_676insT p.Q228fs*7 Chr.5: 79642834-79642835 on Build GRCh38
1070439 c.675T>C p.C225C Chr.5: 79642834-79642834 on Build GRCh38
3074864 c.674_675insT p.Q228fs*7 Chr.5: 79642833-79642834 on Build GRCh38
6737372 c.675delT p.Q228fs*2 Chr.5: 79642834-79642834 on Build GRCh38
163246 c.738C>A p.F246L Chr.5: 79642897-79642897 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs939708859 Chr.5: 79642757-79642757 on Build GRCh38
rs751555715 Chr.5: 79642924-79642924 on Build GRCh38
rs370926509 Chr.5: 79642826-79642826 on Build GRCh38
rs769183115 Chr.5: 79642744-79642744 on Build GRCh38
rs765704956 Chr.5: 79642833-79642834 on Build GRCh38
rs1049890325 Chr.5: 79642841-79642841 on Build GRCh38
rs777530109 Chr.5: 79642902-79642902 on Build GRCh38
rs762319646 Chr.5: 79642817-79642821 on Build GRCh38
rs768988602 Chr.5: 79642829-79642829 on Build GRCh38
rs374745213 Chr.5: 79642845-79642845 on Build GRCh38
rs745568964 Chr.5: 79642811-79642811 on Build GRCh38
rs766710866 Chr.5: 79642877-79642877 on Build GRCh38
rs538260056 Chr.5: 79642771-79642771 on Build GRCh38
rs1036773587 Chr.5: 79642769-79642769 on Build GRCh38
rs575615109 Chr.5: 79642763-79642763 on Build GRCh38
rs780791495 Chr.5: 79642810-79642810 on Build GRCh38
rs374718741 Chr.5: 79642935-79642935 on Build GRCh38
rs138982884 Chr.5: 79642796-79642796 on Build GRCh38
rs774946578 Chr.5: 79642830-79642830 on Build GRCh38
rs753799892 Chr.5: 79642878-79642878 on Build GRCh38
rs761041537 Chr.5: 79642870-79642870 on Build GRCh38
rs755106579 Chr.5: 79642879-79642879 on Build GRCh38
rs774930513 Chr.5: 79642822-79642822 on Build GRCh38
rs768037314 Chr.5: 79642865-79642865 on Build GRCh38
rs112990902 Chr.5: 79642751-79642752 on Build GRCh38
rs770742008 Chr.5: 79642805-79642805 on Build GRCh38
rs886596402 Chr.5: 79642835-79642835 on Build GRCh38
rs142138126 Chr.5: 79642866-79642866 on Build GRCh38

View Full Product Details