Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33263729-33264002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS52
Entrez Gene ID
6293
Gene Name
VPS52, GARP complex subunit
Gene Aliases
ARE1, SAC2, SACM2L, dJ1033B10.5
UniGene
Hs.480356
Gene Chromosome Location
Chr.6: 33250272-33271965 on Build GRCh38
Transcripts
NM_001289174.1 NM_001289175.1 NM_001289176.1 NM_022553.5 XM_011514797.1 XM_011514798.2 XM_011514799.1 XM_017011177.1 XM_017011178.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3873113 c.1709T>C p.M570T Chr.6: 33263791-33263791 on Build GRCh38
1634751 c.1653A>G p.A551A Chr.6: 33263847-33263847 on Build GRCh38
108053 c.1675G>T p.E559* Chr.6: 33263825-33263825 on Build GRCh38
483928 c.1621-1G>A p.? Chr.6: 33263880-33263880 on Build GRCh38
1077976 c.1680G>T p.Q560H Chr.6: 33263820-33263820 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752481839 Chr.6: 33263816-33263816 on Build GRCh38
rs1001899236 Chr.6: 33263939-33263939 on Build GRCh38
rs200423674 Chr.6: 33263762-33263762 on Build GRCh38
rs777759336 Chr.6: 33263833-33263833 on Build GRCh38
rs763983027 Chr.6: 33263976-33263976 on Build GRCh38
rs766072967 Chr.6: 33263789-33263789 on Build GRCh38
rs772513714 Chr.6: 33263862-33263862 on Build GRCh38
rs760052833 Chr.6: 33263975-33263975 on Build GRCh38
rs757600726 Chr.6: 33263981-33263981 on Build GRCh38
rs568396778 Chr.6: 33263924-33263924 on Build GRCh38
rs768003075 Chr.6: 33263766-33263766 on Build GRCh38
rs917763099 Chr.6: 33263942-33263942 on Build GRCh38
rs772935565 Chr.6: 33263832-33263832 on Build GRCh38
rs773748398 Chr.6: 33263904-33263904 on Build GRCh38
rs748543921 Chr.6: 33263857-33263857 on Build GRCh38
rs752106075 Chr.6: 33263833-33263835 on Build GRCh38
rs942222946 Chr.6: 33263799-33263799 on Build GRCh38
rs751478385 Chr.6: 33263981-33263981 on Build GRCh38
rs772666947 Chr.6: 33263774-33263774 on Build GRCh38
rs758270114 Chr.6: 33263826-33263826 on Build GRCh38
rs779431856 Chr.6: 33263855-33263855 on Build GRCh38
rs765039780 Chr.6: 33263805-33263805 on Build GRCh38
rs753457819 Chr.6: 33263792-33263792 on Build GRCh38
rs760144016 Chr.6: 33263910-33263910 on Build GRCh38
rs61730290 Chr.6: 33263795-33263795 on Build GRCh38
rs768950108 Chr.6: 33263851-33263851 on Build GRCh38
rs764554006 Chr.6: 33263754-33263756 on Build GRCh38
rs139356284 Chr.6: 33263825-33263825 on Build GRCh38
rs764805606 Chr.6: 33263962-33263962 on Build GRCh38
rs199625038 Chr.6: 33263835-33263835 on Build GRCh38
rs780463214 Chr.6: 33263845-33263845 on Build GRCh38
rs762533125 Chr.6: 33263919-33263919 on Build GRCh38
rs1055269288 Chr.6: 33263949-33263949 on Build GRCh38
rs752499446 Chr.6: 33263964-33263964 on Build GRCh38
rs760222846 Chr.6: 33263780-33263780 on Build GRCh38
rs749552156 Chr.6: 33263849-33263849 on Build GRCh38
rs369656887 Chr.6: 33263761-33263761 on Build GRCh38
rs756236120 Chr.6: 33263838-33263838 on Build GRCh38
rs937957835 Chr.6: 33263786-33263786 on Build GRCh38
rs148314382 Chr.6: 33263921-33263921 on Build GRCh38
rs770463796 Chr.6: 33263916-33263916 on Build GRCh38

View Full Product Details