Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 76353966-76354227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCAPER
Entrez Gene ID
49855
Gene Name
S-phase cyclin A associated protein in the ER
Gene Aliases
MSTP063, ZNF291, Zfp291
UniGene
Hs.458986
Gene Chromosome Location
Chr.15: 76348185-76905455 on Build GRCh38
Transcripts
NM_001145923.1 NM_020843.2 XM_005254417.2 XM_005254419.2 XM_011521650.1 XM_011521652.2 XM_011521653.2 XM_011521654.2 XM_011521656.2 XM_017022266.1 XM_017022267.1 XM_017022268.1 XM_017022269.1 XM_017022270.1 XM_017022271.1 XM_017022272.1 XM_017022273.1 XM_017022275.1 XM_017022276.1 XM_017022277.1 XM_017022278.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4492115 c.3915C>T p.F1305F Chr.15: 76354081-76354081 on Build GRCh38
6274089 c.3196C>A p.R1066R Chr.15: 76354062-76354062 on Build GRCh38
6274090 c.3934C>A p.R1312R Chr.15: 76354062-76354062 on Build GRCh38
3504177 c.3861C>T p.I1287I Chr.15: 76354135-76354135 on Build GRCh38
3504176 c.3123C>T p.I1041I Chr.15: 76354135-76354135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759288244 Chr.15: 76354119-76354119 on Build GRCh38
rs758607112 Chr.15: 76354122-76354122 on Build GRCh38
rs767705751 Chr.15: 76354111-76354111 on Build GRCh38
rs780336984 Chr.15: 76354159-76354159 on Build GRCh38
rs752906897 Chr.15: 76354121-76354121 on Build GRCh38
rs775650987 Chr.15: 76354177-76354177 on Build GRCh38
rs903059003 Chr.15: 76353991-76353991 on Build GRCh38
rs547563819 Chr.15: 76354165-76354165 on Build GRCh38
rs58388611 Chr.15: 76354114-76354114 on Build GRCh38
rs774619387 Chr.15: 76354015-76354015 on Build GRCh38
rs777730048 Chr.15: 76354029-76354029 on Build GRCh38
rs34110326 Chr.15: 76354144-76354144 on Build GRCh38
rs192286287 Chr.15: 76354178-76354178 on Build GRCh38
rs755168803 Chr.15: 76354012-76354012 on Build GRCh38
rs757819578 Chr.15: 76354134-76354134 on Build GRCh38
rs765023104 Chr.15: 76354120-76354120 on Build GRCh38
rs772755467 Chr.15: 76354172-76354172 on Build GRCh38
rs565954952 Chr.15: 76354173-76354173 on Build GRCh38
rs571838126 Chr.15: 76354070-76354070 on Build GRCh38
rs758438275 Chr.15: 76354018-76354018 on Build GRCh38
rs770310196 Chr.15: 76354061-76354061 on Build GRCh38
rs770461586 Chr.15: 76354174-76354174 on Build GRCh38
rs764234123 Chr.15: 76354126-76354126 on Build GRCh38
rs771636723 Chr.15: 76354032-76354032 on Build GRCh38
rs1013743089 Chr.15: 76354115-76354115 on Build GRCh38
rs761380569 Chr.15: 76354066-76354066 on Build GRCh38
rs746864872 Chr.15: 76354189-76354192 on Build GRCh38
rs781747592 Chr.15: 76354135-76354135 on Build GRCh38
rs773983887 Chr.15: 76354064-76354064 on Build GRCh38
rs746286985 Chr.15: 76354050-76354050 on Build GRCh38
rs187434753 Chr.15: 76354030-76354030 on Build GRCh38
rs781668546 Chr.15: 76354047-76354047 on Build GRCh38
rs756485410 Chr.15: 76354148-76354148 on Build GRCh38
rs780046376 Chr.15: 76354012-76354014 on Build GRCh38
rs369331107 Chr.15: 76353996-76353996 on Build GRCh38
rs747832101 Chr.15: 76354163-76354163 on Build GRCh38
rs779059483 Chr.15: 76354014-76354014 on Build GRCh38
rs760574113 Chr.15: 76354071-76354071 on Build GRCh38

View Full Product Details