Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2853662-2853923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRSS22
Entrez Gene ID
64063
Gene Name
protease, serine 22
Gene Aliases
BSSP-4, SP001LA, hBSSP-4
UniGene
Hs.459709
Gene Chromosome Location
Chr.16: 2852727-2858630 on Build GRCh38
Transcripts
NM_022119.3 XM_005255473.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6451382 c.660C>A p.P220P Chr.16: 2853922-2853922 on Build GRCh38
4556638 c.698G>A p.G233E Chr.16: 2853884-2853884 on Build GRCh38
5521838 c.659C>T p.P220L Chr.16: 2853923-2853923 on Build GRCh38
3110595 c.685G>A p.G229S Chr.16: 2853897-2853897 on Build GRCh38
6757158 c.676C>T p.L226L Chr.16: 2853906-2853906 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368817861 Chr.16: 2853898-2853898 on Build GRCh38
rs760100029 Chr.16: 2853879-2853879 on Build GRCh38
rs1031558754 Chr.16: 2853779-2853779 on Build GRCh38
rs769860950 Chr.16: 2853869-2853869 on Build GRCh38
rs534999567 Chr.16: 2853724-2853724 on Build GRCh38
rs1000508984 Chr.16: 2853754-2853754 on Build GRCh38
rs372408132 Chr.16: 2853895-2853895 on Build GRCh38
rs536551659 Chr.16: 2853842-2853842 on Build GRCh38
rs956169584 Chr.16: 2853808-2853808 on Build GRCh38
rs866529605 Chr.16: 2853896-2853896 on Build GRCh38
rs773485346 Chr.16: 2853871-2853871 on Build GRCh38
rs1024568532 Chr.16: 2853846-2853846 on Build GRCh38
rs112285893 Chr.16: 2853849-2853849 on Build GRCh38
rs935855822 Chr.16: 2853769-2853769 on Build GRCh38
rs980892932 Chr.16: 2853897-2853897 on Build GRCh38
rs748109853 Chr.16: 2853865-2853865 on Build GRCh38
rs746997160 Chr.16: 2853853-2853853 on Build GRCh38
rs753227088 Chr.16: 2853825-2853825 on Build GRCh38
rs567539355 Chr.16: 2853794-2853794 on Build GRCh38
rs534560165 Chr.16: 2853780-2853780 on Build GRCh38
rs375766638 Chr.16: 2853899-2853899 on Build GRCh38
rs201654205 Chr.16: 2853884-2853884 on Build GRCh38
rs779961901 Chr.16: 2853852-2853852 on Build GRCh38
rs904219078 Chr.16: 2853707-2853707 on Build GRCh38
rs781348470 Chr.16: 2853857-2853857 on Build GRCh38
rs763143895 Chr.16: 2853873-2853873 on Build GRCh38
rs774661731 Chr.16: 2853878-2853878 on Build GRCh38
rs768430637 Chr.16: 2853856-2853856 on Build GRCh38
rs78025696 Chr.16: 2853705-2853705 on Build GRCh38
rs771328335 Chr.16: 2853876-2853876 on Build GRCh38
rs755410566 Chr.16: 2853822-2853822 on Build GRCh38
rs186938140 Chr.16: 2853732-2853732 on Build GRCh38
rs2074294 Chr.16: 2853844-2853844 on Build GRCh38
rs370625385 Chr.16: 2853770-2853770 on Build GRCh38

View Full Product Details