Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150695503-150695748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM22
Entrez Gene ID
55696
Gene Name
RNA binding motif protein 22
Gene Aliases
Cwc2, ZC3H16, fSAP47
UniGene
Hs.713564
Gene Chromosome Location
Chr.5: 150690790-150701107 on Build GRCh38
Transcripts
NM_018047.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6241795 c.433G>T p.A145S Chr.5: 150695672-150695672 on Build GRCh38
5867884 c.514C>T p.P172S Chr.5: 150695591-150695591 on Build GRCh38
4404685 c.597A>C p.K199N Chr.5: 150695655-150695655 on Build GRCh38
6406620 c.721G>T p.D241Y Chr.5: 150695531-150695531 on Build GRCh38
5867886 c.502G>A p.A168T Chr.5: 150695603-150695603 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373419831 Chr.5: 150695605-150695605 on Build GRCh38
rs747815625 Chr.5: 150695723-150695726 on Build GRCh38
rs747724385 Chr.5: 150695643-150695643 on Build GRCh38
rs771338991 Chr.5: 150695693-150695693 on Build GRCh38
rs1031906532 Chr.5: 150695716-150695716 on Build GRCh38
rs376547079 Chr.5: 150695652-150695652 on Build GRCh38
rs369569574 Chr.5: 150695574-150695574 on Build GRCh38
rs764227456 Chr.5: 150695539-150695539 on Build GRCh38
rs780680359 Chr.5: 150695567-150695567 on Build GRCh38
rs758349183 Chr.5: 150695552-150695552 on Build GRCh38
rs751302951 Chr.5: 150695559-150695559 on Build GRCh38
rs766340552 Chr.5: 150695553-150695553 on Build GRCh38
rs753986489 Chr.5: 150695543-150695543 on Build GRCh38
rs977258600 Chr.5: 150695714-150695714 on Build GRCh38
rs746194675 Chr.5: 150695718-150695718 on Build GRCh38
rs1804920 Chr.5: 150695700-150695700 on Build GRCh38
rs867171275 Chr.5: 150695637-150695637 on Build GRCh38
rs774980920 Chr.5: 150695710-150695710 on Build GRCh38
rs375056561 Chr.5: 150695561-150695561 on Build GRCh38

View Full Product Details