Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 63594170-63594443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINB13
Entrez Gene ID
5275
Gene Name
serpin family B member 13
Gene Aliases
HSHUR7SEQ, HUR7, PI13, headpin
UniGene
Hs.241407
Gene Chromosome Location
Chr.18: 63587206-63599199 on Build GRCh38
Transcripts
NM_001307923.1 NM_012397.3 XM_005266707.3 XM_011526029.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7389389 c.496G>T p.D166Y Chr.18: 63594378-63594378 on Build GRCh38
3891241 c.473-1G>A p.? Chr.18: 63594354-63594354 on Build GRCh38
212122 c.482A>T p.K161M Chr.18: 63594364-63594364 on Build GRCh38
6235003 c.507T>A p.I169I Chr.18: 63594389-63594389 on Build GRCh38
1680396 c.503C>T p.S168F Chr.18: 63594385-63594385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373011810 Chr.18: 63594413-63594413 on Build GRCh38
rs748273316 Chr.18: 63594256-63594256 on Build GRCh38
rs747280817 Chr.18: 63594356-63594356 on Build GRCh38
rs769727914 Chr.18: 63594366-63594366 on Build GRCh38
rs914830313 Chr.18: 63594298-63594298 on Build GRCh38
rs536289864 Chr.18: 63594375-63594375 on Build GRCh38
rs117476956 Chr.18: 63594273-63594273 on Build GRCh38
rs538336644 Chr.18: 63594324-63594324 on Build GRCh38
rs754326375 Chr.18: 63594198-63594198 on Build GRCh38
rs368996053 Chr.18: 63594346-63594346 on Build GRCh38
rs759511075 Chr.18: 63594282-63594282 on Build GRCh38
rs924920850 Chr.18: 63594307-63594307 on Build GRCh38
rs777152514 Chr.18: 63594305-63594306 on Build GRCh38
rs754361059 Chr.18: 63594330-63594330 on Build GRCh38
rs538280384 Chr.18: 63594197-63594197 on Build GRCh38
rs372688280 Chr.18: 63594351-63594351 on Build GRCh38
rs370000053 Chr.18: 63594387-63594387 on Build GRCh38
rs746616561 Chr.18: 63594307-63594308 on Build GRCh38
rs751225311 Chr.18: 63594311-63594311 on Build GRCh38
rs749856118 Chr.18: 63594230-63594230 on Build GRCh38
rs545075987 Chr.18: 63594299-63594299 on Build GRCh38
rs764678087 Chr.18: 63594286-63594286 on Build GRCh38
rs772819902 Chr.18: 63594271-63594271 on Build GRCh38
rs775079263 Chr.18: 63594287-63594287 on Build GRCh38
rs750002811 Chr.18: 63594354-63594355 on Build GRCh38
rs144903442 Chr.18: 63594414-63594414 on Build GRCh38
rs201275072 Chr.18: 63594317-63594317 on Build GRCh38
rs762507739 Chr.18: 63594374-63594374 on Build GRCh38
rs770513475 Chr.18: 63594310-63594311 on Build GRCh38
rs577063790 Chr.18: 63594363-63594363 on Build GRCh38
rs779404417 Chr.18: 63594266-63594266 on Build GRCh38
rs11152394 Chr.18: 63594281-63594281 on Build GRCh38
rs779169225 Chr.18: 63594242-63594242 on Build GRCh38
rs544319890 Chr.18: 63594394-63594394 on Build GRCh38
rs188869995 Chr.18: 63594200-63594200 on Build GRCh38
rs774021426 Chr.18: 63594380-63594380 on Build GRCh38
rs761585476 Chr.18: 63594386-63594386 on Build GRCh38
rs934189508 Chr.18: 63594284-63594284 on Build GRCh38
rs11152395 Chr.18: 63594306-63594306 on Build GRCh38
rs199653578 Chr.18: 63594371-63594371 on Build GRCh38
rs1014746759 Chr.18: 63594216-63594216 on Build GRCh38
rs1017433424 Chr.18: 63594223-63594223 on Build GRCh38
rs11152396 Chr.18: 63594331-63594331 on Build GRCh38
rs770682594 Chr.18: 63594357-63594357 on Build GRCh38
rs760057833 Chr.18: 63594410-63594410 on Build GRCh38
rs757705436 Chr.18: 63594219-63594219 on Build GRCh38
rs181614529 Chr.18: 63594288-63594288 on Build GRCh38
rs755073818 Chr.18: 63594233-63594233 on Build GRCh38
rs143961207 Chr.18: 63594365-63594365 on Build GRCh38
rs776529578 Chr.18: 63594405-63594406 on Build GRCh38
rs766886009 Chr.18: 63594320-63594320 on Build GRCh38
rs267605226 Chr.18: 63594362-63594362 on Build GRCh38
rs867235066 Chr.18: 63594218-63594218 on Build GRCh38

View Full Product Details