Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 43068355-43068628 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTLL1
Entrez Gene ID
25809
Gene Name
tubulin tyrosine ligase like 1
Gene Aliases
C22orf7, HS323M22B
UniGene
Hs.660298
Gene Chromosome Location
Chr.22: 43039516-43089428 on Build GRCh38
Transcripts
NM_012263.4 NR_027779.1 XM_006724225.3 XM_011530108.2 XM_011530110.2 XM_017028751.1 XM_017028752.1 XM_017028753.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3363687 c.405G>T p.W135C Chr.22: 43068508-43068508 on Build GRCh38
1682309 c.355G>A p.D119N Chr.22: 43068558-43068558 on Build GRCh38
5499962 c.453C>T p.I151I Chr.22: 43068460-43068460 on Build GRCh38
330406 c.441C>T p.G147G Chr.22: 43068472-43068472 on Build GRCh38
6811408 c.392C>T p.P131L Chr.22: 43068521-43068521 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754256538 Chr.22: 43068521-43068521 on Build GRCh38
rs371528419 Chr.22: 43068609-43068609 on Build GRCh38
rs1031350089 Chr.22: 43068415-43068415 on Build GRCh38
rs779881916 Chr.22: 43068413-43068413 on Build GRCh38
rs138993027 Chr.22: 43068503-43068503 on Build GRCh38
rs149115561 Chr.22: 43068430-43068430 on Build GRCh38
rs140558809 Chr.22: 43068529-43068529 on Build GRCh38
rs767793669 Chr.22: 43068588-43068588 on Build GRCh38
rs752825372 Chr.22: 43068500-43068500 on Build GRCh38
rs746695944 Chr.22: 43068422-43068422 on Build GRCh38
rs762768609 Chr.22: 43068455-43068455 on Build GRCh38
rs768431621 Chr.22: 43068550-43068550 on Build GRCh38
rs940690448 Chr.22: 43068398-43068398 on Build GRCh38
rs767721476 Chr.22: 43068487-43068487 on Build GRCh38
rs747715920 Chr.22: 43068554-43068554 on Build GRCh38
rs557734562 Chr.22: 43068600-43068601 on Build GRCh38
rs748079631 Chr.22: 43068594-43068595 on Build GRCh38
rs938973807 Chr.22: 43068452-43068453 on Build GRCh38
rs752704073 Chr.22: 43068380-43068380 on Build GRCh38
rs768413237 Chr.22: 43068427-43068427 on Build GRCh38
rs776273674 Chr.22: 43068429-43068429 on Build GRCh38
rs138233602 Chr.22: 43068584-43068584 on Build GRCh38
rs1000819271 Chr.22: 43068384-43068384 on Build GRCh38
rs761741217 Chr.22: 43068442-43068442 on Build GRCh38
rs533160676 Chr.22: 43068402-43068411 on Build GRCh38
rs756086352 Chr.22: 43068518-43068518 on Build GRCh38
rs149584241 Chr.22: 43068585-43068585 on Build GRCh38
rs775521985 Chr.22: 43068596-43068596 on Build GRCh38
rs780653917 Chr.22: 43068551-43068551 on Build GRCh38
rs373336576 Chr.22: 43068561-43068561 on Build GRCh38
rs770840187 Chr.22: 43068565-43068565 on Build GRCh38
rs759809722 Chr.22: 43068482-43068482 on Build GRCh38
rs774475059 Chr.22: 43068463-43068466 on Build GRCh38
rs768741672 Chr.22: 43068388-43068389 on Build GRCh38
rs766204003 Chr.22: 43068464-43068464 on Build GRCh38
rs6003030 Chr.22: 43068410-43068410 on Build GRCh38
rs146662174 Chr.22: 43068409-43068409 on Build GRCh38
rs769788157 Chr.22: 43068449-43068449 on Build GRCh38
rs868842293 Chr.22: 43068413-43068414 on Build GRCh38
rs370165605 Chr.22: 43068520-43068520 on Build GRCh38
rs145162777 Chr.22: 43068469-43068469 on Build GRCh38
rs201616202 Chr.22: 43068538-43068538 on Build GRCh38
rs150448437 Chr.22: 43068562-43068562 on Build GRCh38
rs375050767 Chr.22: 43068391-43068391 on Build GRCh38
rs370182099 Chr.22: 43068414-43068414 on Build GRCh38
rs756711124 Chr.22: 43068389-43068389 on Build GRCh38
rs779480978 Chr.22: 43068531-43068531 on Build GRCh38
rs1011678488 Chr.22: 43068517-43068517 on Build GRCh38
rs903837151 Chr.22: 43068383-43068383 on Build GRCh38
rs772974940 Chr.22: 43068454-43068454 on Build GRCh38

View Full Product Details