Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 45703311-45703572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SACM1L
Entrez Gene ID
22908
Gene Name
SAC1 suppressor of actin mutations 1-like (yeast)
Gene Aliases
SAC1
UniGene
Hs.156509
Gene Chromosome Location
Chr.3: 45689241-45745425 on Build GRCh38
Transcripts
NM_001319072.1 NM_001319073.1 NM_014016.4 XM_011533500.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4117821 c.103C>G p.R35G Chr.3: 45703508-45703508 on Build GRCh38
4462497 c.124C>T p.L42F Chr.3: 45703529-45703529 on Build GRCh38
4920282 c.63A>G p.E21E Chr.3: 45703468-45703468 on Build GRCh38
6258967 c.108G>T p.V36V Chr.3: 45703513-45703513 on Build GRCh38
1045172 c.33-1G>T p.? Chr.3: 45703437-45703437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746985236 Chr.3: 45703400-45703400 on Build GRCh38
rs374190458 Chr.3: 45703500-45703500 on Build GRCh38
rs779486295 Chr.3: 45703480-45703480 on Build GRCh38
rs753237930 Chr.3: 45703433-45703433 on Build GRCh38
rs141930534 Chr.3: 45703358-45703358 on Build GRCh38
rs894649862 Chr.3: 45703386-45703400 on Build GRCh38
rs983489169 Chr.3: 45703496-45703496 on Build GRCh38
rs754189139 Chr.3: 45703449-45703449 on Build GRCh38
rs1018922406 Chr.3: 45703508-45703508 on Build GRCh38
rs1032516791 Chr.3: 45703395-45703395 on Build GRCh38
rs756922702 Chr.3: 45703451-45703451 on Build GRCh38
rs767530817 Chr.3: 45703430-45703430 on Build GRCh38
rs759586930 Chr.3: 45703429-45703429 on Build GRCh38
rs763094491 Chr.3: 45703419-45703419 on Build GRCh38
rs757979090 Chr.3: 45703477-45703477 on Build GRCh38
rs774811718 Chr.3: 45703440-45703440 on Build GRCh38
rs188211739 Chr.3: 45703484-45703484 on Build GRCh38
rs781348073 Chr.3: 45703406-45703409 on Build GRCh38
rs764531339 Chr.3: 45703519-45703519 on Build GRCh38
rs778686870 Chr.3: 45703464-45703464 on Build GRCh38
rs747955868 Chr.3: 45703493-45703493 on Build GRCh38
rs138379651 Chr.3: 45703514-45703514 on Build GRCh38
rs935946700 Chr.3: 45703394-45703394 on Build GRCh38
rs766615536 Chr.3: 45703425-45703425 on Build GRCh38
rs748211519 Chr.3: 45703534-45703534 on Build GRCh38
rs764457057 Chr.3: 45703448-45703448 on Build GRCh38
rs907922200 Chr.3: 45703509-45703509 on Build GRCh38
rs371274265 Chr.3: 45703441-45703441 on Build GRCh38
rs1053432549 Chr.3: 45703413-45703413 on Build GRCh38
rs904331550 Chr.3: 45703359-45703359 on Build GRCh38
rs776446200 Chr.3: 45703492-45703492 on Build GRCh38
rs774361689 Chr.3: 45703426-45703426 on Build GRCh38
rs1030276974 Chr.3: 45703472-45703472 on Build GRCh38
rs754447001 Chr.3: 45703529-45703529 on Build GRCh38
rs767579328 Chr.3: 45703502-45703502 on Build GRCh38
rs745558204 Chr.3: 45703476-45703476 on Build GRCh38
rs768483093 Chr.3: 45703402-45703402 on Build GRCh38
rs1001410632 Chr.3: 45703393-45703397 on Build GRCh38
rs776303104 Chr.3: 45703409-45703409 on Build GRCh38
rs768481910 Chr.3: 45703489-45703489 on Build GRCh38
rs759817847 Chr.3: 45703501-45703501 on Build GRCh38
rs775468178 Chr.3: 45703396-45703396 on Build GRCh38
rs761208104 Chr.3: 45703515-45703515 on Build GRCh38

View Full Product Details