Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 10464867-10465094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ROPN1L
Entrez Gene ID
83853
Gene Name
rhophilin associated tail protein 1 like
Gene Aliases
ASP, RSPH11
UniGene
Hs.381089
Gene Chromosome Location
Chr.5: 10441862-10482807 on Build GRCh38
Transcripts
NM_001201466.1 NM_031916.4 XM_006714503.2 XM_006714504.2 XM_017009946.1 XM_017009947.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
735131 c.679G>T p.D227Y Chr.5: 10464933-10464933 on Build GRCh38
6168583 c.636C>A p.F212L Chr.5: 10464890-10464890 on Build GRCh38
5401896 c.679G>A p.D227N Chr.5: 10464933-10464933 on Build GRCh38
448400 c.622C>G p.L208V Chr.5: 10464876-10464876 on Build GRCh38
88655 c.667G>C p.E223Q Chr.5: 10464921-10464921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770280679 Chr.5: 10464936-10464936 on Build GRCh38
rs1051157321 Chr.5: 10465015-10465015 on Build GRCh38
rs142401850 Chr.5: 10464910-10464910 on Build GRCh38
rs528006489 Chr.5: 10465005-10465005 on Build GRCh38
rs754827777 Chr.5: 10464901-10464901 on Build GRCh38
rs773523032 Chr.5: 10464969-10464969 on Build GRCh38
rs559060316 Chr.5: 10464960-10464960 on Build GRCh38
rs374649146 Chr.5: 10464988-10464988 on Build GRCh38
rs146235615 Chr.5: 10464951-10464951 on Build GRCh38
rs748897594 Chr.5: 10464933-10464933 on Build GRCh38
rs772777023 Chr.5: 10464915-10464915 on Build GRCh38
rs563081436 Chr.5: 10464919-10464919 on Build GRCh38
rs142858482 Chr.5: 10464939-10464939 on Build GRCh38
rs574244668 Chr.5: 10464909-10464909 on Build GRCh38
rs189726346 Chr.5: 10465032-10465032 on Build GRCh38
rs769075460 Chr.5: 10464986-10464987 on Build GRCh38
rs34596820 Chr.5: 10464930-10464931 on Build GRCh38
rs745468405 Chr.5: 10464942-10464942 on Build GRCh38
rs939837714 Chr.5: 10465034-10465034 on Build GRCh38
rs533667087 Chr.5: 10464973-10464973 on Build GRCh38
rs890967778 Chr.5: 10465018-10465018 on Build GRCh38
rs765796257 Chr.5: 10464963-10464963 on Build GRCh38
rs762148367 Chr.5: 10464962-10464962 on Build GRCh38
rs763104402 Chr.5: 10464970-10464970 on Build GRCh38

View Full Product Details