Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44861098-44861333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTTG1IP
Entrez Gene ID
754
Gene Name
pituitary tumor-transforming 1 interacting protein
Gene Aliases
C21orf1, C21orf3, PBF
UniGene
Hs.474010
Gene Chromosome Location
Chr.21: 44849585-44873903 on Build GRCh38
Transcripts
NM_001286822.1 NM_004339.3 NR_104597.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5858764 c.211G>A p.V71I Chr.21: 44861229-44861229 on Build GRCh38
7154435 c.276G>T p.W92C Chr.21: 44861164-44861164 on Build GRCh38
5733565 c.268G>A p.V90I Chr.21: 44861172-44861172 on Build GRCh38
6161568 c.176_177GG>TC p.W59F Chr.21: 44861263-44861264 on Build GRCh38
1414448 c.259C>T p.R87C Chr.21: 44861181-44861181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780746972 Chr.21: 44861152-44861152 on Build GRCh38
rs777140471 Chr.21: 44861302-44861302 on Build GRCh38
rs574813996 Chr.21: 44861180-44861180 on Build GRCh38
rs774766842 Chr.21: 44861130-44861130 on Build GRCh38
rs778849261 Chr.21: 44861278-44861278 on Build GRCh38
rs758032410 Chr.21: 44861154-44861154 on Build GRCh38
rs751093743 Chr.21: 44861241-44861241 on Build GRCh38
rs780790893 Chr.21: 44861182-44861182 on Build GRCh38
rs995818281 Chr.21: 44861181-44861181 on Build GRCh38
rs772696653 Chr.21: 44861218-44861218 on Build GRCh38
rs759252090 Chr.21: 44861147-44861147 on Build GRCh38
rs773729193 Chr.21: 44861141-44861141 on Build GRCh38
rs763445809 Chr.21: 44861240-44861240 on Build GRCh38
rs369693004 Chr.21: 44861296-44861296 on Build GRCh38
rs761164176 Chr.21: 44861221-44861221 on Build GRCh38
rs749344323 Chr.21: 44861202-44861202 on Build GRCh38
rs779842845 Chr.21: 44861268-44861268 on Build GRCh38
rs144668992 Chr.21: 44861255-44861255 on Build GRCh38
rs762401175 Chr.21: 44861136-44861136 on Build GRCh38
rs1017735492 Chr.21: 44861223-44861223 on Build GRCh38
rs771383132 Chr.21: 44861298-44861298 on Build GRCh38
rs555793895 Chr.21: 44861127-44861127 on Build GRCh38
rs767712471 Chr.21: 44861139-44861139 on Build GRCh38
rs749467135 Chr.21: 44861271-44861271 on Build GRCh38
rs778474487 Chr.21: 44861292-44861292 on Build GRCh38
rs200698166 Chr.21: 44861220-44861220 on Build GRCh38
rs188877011 Chr.21: 44861210-44861210 on Build GRCh38
rs772430025 Chr.21: 44861289-44861289 on Build GRCh38
rs540961273 Chr.21: 44861239-44861239 on Build GRCh38
rs780292790 Chr.21: 44861198-44861198 on Build GRCh38
rs11537961 Chr.21: 44861242-44861242 on Build GRCh38
rs768565617 Chr.21: 44861209-44861209 on Build GRCh38
rs756323234 Chr.21: 44861192-44861192 on Build GRCh38
rs142345359 Chr.21: 44861227-44861227 on Build GRCh38
rs572176295 Chr.21: 44861228-44861228 on Build GRCh38
rs776852385 Chr.21: 44861286-44861287 on Build GRCh38
rs755949360 Chr.21: 44861264-44861264 on Build GRCh38
rs775235646 Chr.21: 44861231-44861231 on Build GRCh38
rs755245744 Chr.21: 44861272-44861272 on Build GRCh38
rs769172206 Chr.21: 44861128-44861128 on Build GRCh38
rs376599030 Chr.21: 44861145-44861145 on Build GRCh38
rs745718441 Chr.21: 44861188-44861188 on Build GRCh38
rs755774187 Chr.21: 44861288-44861288 on Build GRCh38
rs867604734 Chr.21: 44861168-44861168 on Build GRCh38
rs752225430 Chr.21: 44861153-44861153 on Build GRCh38
rs763682648 Chr.21: 44861155-44861155 on Build GRCh38
rs757127661 Chr.21: 44861254-44861254 on Build GRCh38

View Full Product Details