Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65096136-65096375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS51
Entrez Gene ID
738
Gene Name
VPS51, GARP complex subunit
Gene Aliases
ANG2, ANG3, C11orf2, C11orf3, FFR
UniGene
Hs.277517
Gene Chromosome Location
Chr.11: 65096115-65111860 on Build GRCh38
Transcripts
NM_013265.3 NR_073519.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4891565 c.91C>T p.R31W Chr.11: 65096341-65096341 on Build GRCh38
5460651 c.21C>A p.A7A Chr.11: 65096271-65096271 on Build GRCh38
4567700 c.81_82GG>AA p.E28K Chr.11: 65096331-65096332 on Build GRCh38
1289603 c.46delG p.D17fs*21 Chr.11: 65096296-65096296 on Build GRCh38
4614681 c.69delG p.E25fs*13 Chr.11: 65096319-65096319 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764092824 Chr.11: 65096294-65096294 on Build GRCh38
rs757532613 Chr.11: 65096284-65096284 on Build GRCh38
rs184045929 Chr.11: 65096319-65096319 on Build GRCh38
rs575107564 Chr.11: 65096210-65096210 on Build GRCh38
rs768484102 Chr.11: 65096305-65096305 on Build GRCh38
rs962847102 Chr.11: 65096269-65096269 on Build GRCh38
rs771429081 Chr.11: 65096345-65096345 on Build GRCh38
rs544525817 Chr.11: 65096214-65096214 on Build GRCh38
rs748143996 Chr.11: 65096335-65096335 on Build GRCh38
rs1044034908 Chr.11: 65096188-65096188 on Build GRCh38
rs777921734 Chr.11: 65096281-65096281 on Build GRCh38
rs747134833 Chr.11: 65096282-65096282 on Build GRCh38
rs974578367 Chr.11: 65096313-65096313 on Build GRCh38
rs771431896 Chr.11: 65096207-65096207 on Build GRCh38
rs760376454 Chr.11: 65096259-65096259 on Build GRCh38
rs933115589 Chr.11: 65096213-65096213 on Build GRCh38
rs921803718 Chr.11: 65096314-65096314 on Build GRCh38
rs749061638 Chr.11: 65096293-65096293 on Build GRCh38
rs765807256 Chr.11: 65096270-65096270 on Build GRCh38
rs11548900 Chr.11: 65096295-65096295 on Build GRCh38
rs758537539 Chr.11: 65096276-65096276 on Build GRCh38
rs1015601126 Chr.11: 65096283-65096283 on Build GRCh38
rs999913169 Chr.11: 65096231-65096231 on Build GRCh38
rs533209939 Chr.11: 65096349-65096349 on Build GRCh38
rs755439481 Chr.11: 65096264-65096264 on Build GRCh38
rs753322601 Chr.11: 65096275-65096275 on Build GRCh38
rs762838125 Chr.11: 65096187-65096187 on Build GRCh38
rs976787069 Chr.11: 65096338-65096338 on Build GRCh38
rs767125286 Chr.11: 65096228-65096228 on Build GRCh38
rs896544707 Chr.11: 65096246-65096246 on Build GRCh38
rs954176017 Chr.11: 65096343-65096343 on Build GRCh38
rs772730076 Chr.11: 65096232-65096232 on Build GRCh38
rs116522417 Chr.11: 65096157-65096157 on Build GRCh38
rs774271870 Chr.11: 65096324-65096324 on Build GRCh38
rs781201726 Chr.11: 65096314-65096328 on Build GRCh38
rs900430609 Chr.11: 65096180-65096180 on Build GRCh38
rs766610118 Chr.11: 65096261-65096261 on Build GRCh38
rs1030120733 Chr.11: 65096340-65096340 on Build GRCh38
rs36115087 Chr.11: 65096296-65096296 on Build GRCh38
rs760193484 Chr.11: 65096351-65096351 on Build GRCh38
rs761285445 Chr.11: 65096222-65096222 on Build GRCh38
rs779664684 Chr.11: 65096285-65096285 on Build GRCh38
rs934422727 Chr.11: 65096260-65096260 on Build GRCh38
rs910448379 Chr.11: 65096227-65096227 on Build GRCh38
rs768603549 Chr.11: 65096201-65096201 on Build GRCh38
rs754247915 Chr.11: 65096263-65096263 on Build GRCh38
rs923122012 Chr.11: 65096202-65096202 on Build GRCh38

View Full Product Details